Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 238923209-238923737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ43879
Entrez Gene ID
401039
Gene Name
FLJ43879 protein
Gene Aliases
-
UniGene
Hs.516791
Gene Chromosome Location
Chr.2: 238919302-238926269 on Build GRCh38
Transcripts
NR_034162.1

SNP Detail

SNP ID Chromosome Location
rs200928061 Chr.2: 238923267-238923268 on Build GRCh38
rs948336667 Chr.2: 238923242-238923242 on Build GRCh38
rs1033864794 Chr.2: 238923571-238923571 on Build GRCh38
rs192649972 Chr.2: 238923563-238923563 on Build GRCh38
rs892472045 Chr.2: 238923532-238923532 on Build GRCh38
rs541340102 Chr.2: 238923608-238923608 on Build GRCh38
rs939356267 Chr.2: 238923264-238923264 on Build GRCh38
rs941441017 Chr.2: 238923278-238923278 on Build GRCh38
rs374711161 Chr.2: 238923302-238923302 on Build GRCh38
rs985115155 Chr.2: 238923411-238923411 on Build GRCh38
rs928229803 Chr.2: 238923489-238923489 on Build GRCh38
rs1002414166 Chr.2: 238923332-238923332 on Build GRCh38
rs879704523 Chr.2: 238923328-238923329 on Build GRCh38
rs766918019 Chr.2: 238923618-238923618 on Build GRCh38
rs562899532 Chr.2: 238923395-238923395 on Build GRCh38
rs773632411 Chr.2: 238923609-238923609 on Build GRCh38
rs537797445 Chr.2: 238923259-238923259 on Build GRCh38
rs980917273 Chr.2: 238923476-238923476 on Build GRCh38
rs549945399 Chr.2: 238923699-238923699 on Build GRCh38
rs572127853 Chr.2: 238923375-238923375 on Build GRCh38
rs1016330696 Chr.2: 238923648-238923648 on Build GRCh38
rs182552526 Chr.2: 238923402-238923402 on Build GRCh38
rs960466637 Chr.2: 238923351-238923352 on Build GRCh38
rs576138977 Chr.2: 238923434-238923434 on Build GRCh38
rs960940895 Chr.2: 238923494-238923494 on Build GRCh38
rs1002436779 Chr.2: 238923567-238923567 on Build GRCh38
rs564928139 Chr.2: 238923587-238923587 on Build GRCh38
rs1022473143 Chr.2: 238923453-238923453 on Build GRCh38
rs908500458 Chr.2: 238923565-238923565 on Build GRCh38
rs950378954 Chr.2: 238923388-238923388 on Build GRCh38
rs112191674 Chr.2: 238923337-238923337 on Build GRCh38
rs79946209 Chr.2: 238923682-238923682 on Build GRCh38
rs368170951 Chr.2: 238923457-238923457 on Build GRCh38
rs964621828 Chr.2: 238923681-238923681 on Build GRCh38
rs1047081105 Chr.2: 238923266-238923266 on Build GRCh38
rs1050283449 Chr.2: 238923371-238923371 on Build GRCh38
rs188122627 Chr.2: 238923416-238923416 on Build GRCh38
rs923090673 Chr.2: 238923460-238923460 on Build GRCh38
rs1030620231 Chr.2: 238923385-238923385 on Build GRCh38
rs762525214 Chr.2: 238923250-238923250 on Build GRCh38
rs1051101135 Chr.2: 238923507-238923507 on Build GRCh38
rs145913359 Chr.2: 238923277-238923277 on Build GRCh38
rs74463556 Chr.2: 238923593-238923593 on Build GRCh38
rs943273561 Chr.2: 238923446-238923446 on Build GRCh38
rs1011036634 Chr.2: 238923428-238923428 on Build GRCh38
rs141491775 Chr.2: 238923628-238923628 on Build GRCh38
rs754308410 Chr.2: 238923644-238923644 on Build GRCh38
rs1008726748 Chr.2: 238923633-238923633 on Build GRCh38
rs538550512 Chr.2: 238923319-238923319 on Build GRCh38
rs189025975 Chr.2: 238923712-238923712 on Build GRCh38
rs945360305 Chr.2: 238923547-238923547 on Build GRCh38
rs1010249116 Chr.2: 238923288-238923288 on Build GRCh38
rs934097027 Chr.2: 238923475-238923475 on Build GRCh38
rs890356675 Chr.2: 238923372-238923372 on Build GRCh38
rs147821890 Chr.2: 238923374-238923374 on Build GRCh38
rs112978100 Chr.2: 238923700-238923700 on Build GRCh38
rs1050207124 Chr.2: 238923616-238923616 on Build GRCh38
rs1033832374 Chr.2: 238923345-238923345 on Build GRCh38
rs117084578 Chr.2: 238923301-238923301 on Build GRCh38
rs184545433 Chr.2: 238923698-238923698 on Build GRCh38
rs915355360 Chr.2: 238923240-238923240 on Build GRCh38
rs969936850 Chr.2: 238923454-238923454 on Build GRCh38
rs377588407 Chr.2: 238923254-238923254 on Build GRCh38
rs140832008 Chr.2: 238923248-238923248 on Build GRCh38
rs368509242 Chr.2: 238923617-238923617 on Build GRCh38
rs548807235 Chr.2: 238923664-238923664 on Build GRCh38

View Full Product Details