Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 238923611-238924111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ43879
Entrez Gene ID
401039
Gene Name
FLJ43879 protein
Gene Aliases
-
UniGene
Hs.516791
Gene Chromosome Location
Chr.2: 238919302-238926269 on Build GRCh38
Transcripts
NR_034162.1

SNP Detail

SNP ID Chromosome Location
rs549564272 Chr.2: 238924083-238924085 on Build GRCh38
rs34292287 Chr.2: 238923867-238923868 on Build GRCh38
rs914031256 Chr.2: 238923767-238923767 on Build GRCh38
rs921380459 Chr.2: 238923909-238923909 on Build GRCh38
rs1006365649 Chr.2: 238924002-238924002 on Build GRCh38
rs868286042 Chr.2: 238923832-238923832 on Build GRCh38
rs537132887 Chr.2: 238923997-238923997 on Build GRCh38
rs113190768 Chr.2: 238923893-238923893 on Build GRCh38
rs536019503 Chr.2: 238923863-238923863 on Build GRCh38
rs375551377 Chr.2: 238923934-238923934 on Build GRCh38
rs908555085 Chr.2: 238923897-238923897 on Build GRCh38
rs963011823 Chr.2: 238923902-238923902 on Build GRCh38
rs1035185014 Chr.2: 238923864-238923864 on Build GRCh38
rs1043754851 Chr.2: 238923822-238923822 on Build GRCh38
rs549945399 Chr.2: 238923699-238923699 on Build GRCh38
rs974641298 Chr.2: 238924018-238924018 on Build GRCh38
rs1016330696 Chr.2: 238923648-238923648 on Build GRCh38
rs944637780 Chr.2: 238923963-238923963 on Build GRCh38
rs79946209 Chr.2: 238923682-238923682 on Build GRCh38
rs969152060 Chr.2: 238923766-238923766 on Build GRCh38
rs752768140 Chr.2: 238924012-238924012 on Build GRCh38
rs964621828 Chr.2: 238923681-238923681 on Build GRCh38
rs985927239 Chr.2: 238923927-238923927 on Build GRCh38
rs982999865 Chr.2: 238923895-238923895 on Build GRCh38
rs886219830 Chr.2: 238923965-238923965 on Build GRCh38
rs946402886 Chr.2: 238924027-238924027 on Build GRCh38
rs964318650 Chr.2: 238924015-238924015 on Build GRCh38
rs74888197 Chr.2: 238924022-238924022 on Build GRCh38
rs34325667 Chr.2: 238923762-238923762 on Build GRCh38
rs904981075 Chr.2: 238924038-238924038 on Build GRCh38
rs938256187 Chr.2: 238923892-238923892 on Build GRCh38
rs73097401 Chr.2: 238924023-238924023 on Build GRCh38
rs114249901 Chr.2: 238923911-238923911 on Build GRCh38
rs754308410 Chr.2: 238923644-238923644 on Build GRCh38
rs1008726748 Chr.2: 238923633-238923633 on Build GRCh38
rs1022486604 Chr.2: 238923737-238923737 on Build GRCh38
rs370961758 Chr.2: 238923800-238923800 on Build GRCh38
rs559753556 Chr.2: 238924052-238924052 on Build GRCh38
rs1043390979 Chr.2: 238924037-238924037 on Build GRCh38
rs547347018 Chr.2: 238923777-238923777 on Build GRCh38
rs558653277 Chr.2: 238924010-238924010 on Build GRCh38
rs189025975 Chr.2: 238923712-238923712 on Build GRCh38
rs945447299 Chr.2: 238923768-238923768 on Build GRCh38
rs112978100 Chr.2: 238923700-238923700 on Build GRCh38
rs974084764 Chr.2: 238923907-238923907 on Build GRCh38
rs184545433 Chr.2: 238923698-238923698 on Build GRCh38
rs527908625 Chr.2: 238924039-238924039 on Build GRCh38
rs548807235 Chr.2: 238923664-238923664 on Build GRCh38
rs201497552 Chr.2: 238923851-238923851 on Build GRCh38

View Full Product Details