Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 58850384-58850921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAS
Entrez Gene ID
2778
Gene Name
GNAS complex locus
Gene Aliases
AHO, C20orf45, GNAS1, GPSA, GSA, GSP, NESP, POH, SCG6, SgVI
UniGene
Hs.125898
Gene Chromosome Location
Chr.20: 58839681-58911196 on Build GRCh38
Transcripts
NM_001309861.1 NM_016592.3 XM_017027815.1 XM_017027821.1 XM_017027822.1 NR_002785.2

Gene Symbol
GNAS-AS1
Entrez Gene ID
149775
Gene Name
GNAS antisense RNA 1
Gene Aliases
GNAS-AS, GNAS1AS, GNASAS, NCRNA00075, NESP-AS, NESPAS, SANG
UniGene
Hs.122718
Gene Chromosome Location
Chr.20: 58818918-58850903 on Build GRCh38
Transcripts
NM_001309861.1 NM_016592.3 XM_017027815.1 XM_017027821.1 XM_017027822.1 NR_002785.2

SNP Detail

SNP ID Chromosome Location
rs1027531412 Chr.20: 58850890-58850890 on Build GRCh38
rs1047632511 Chr.20: 58850439-58850439 on Build GRCh38
rs527929829 Chr.20: 58850441-58850441 on Build GRCh38
rs928531341 Chr.20: 58850768-58850768 on Build GRCh38
rs902642015 Chr.20: 58850526-58850526 on Build GRCh38
rs975620050 Chr.20: 58850415-58850415 on Build GRCh38
rs561382585 Chr.20: 58850487-58850487 on Build GRCh38
rs573544132 Chr.20: 58850845-58850845 on Build GRCh38
rs953001595 Chr.20: 58850892-58850892 on Build GRCh38
rs935491046 Chr.20: 58850812-58850812 on Build GRCh38
rs879566047 Chr.20: 58850869-58850869 on Build GRCh38
rs749604101 Chr.20: 58850508-58850508 on Build GRCh38
rs956495397 Chr.20: 58850594-58850594 on Build GRCh38
rs1019316056 Chr.20: 58850876-58850876 on Build GRCh38
rs937282779 Chr.20: 58850791-58850791 on Build GRCh38
rs530438616 Chr.20: 58850703-58850703 on Build GRCh38
rs575054931 Chr.20: 58850461-58850461 on Build GRCh38
rs192776054 Chr.20: 58850795-58850795 on Build GRCh38
rs931520358 Chr.20: 58850756-58850756 on Build GRCh38
rs951801805 Chr.20: 58850438-58850438 on Build GRCh38
rs964222310 Chr.20: 58850885-58850885 on Build GRCh38
rs982809062 Chr.20: 58850788-58850788 on Build GRCh38
rs1000111099 Chr.20: 58850573-58850573 on Build GRCh38
rs550172967 Chr.20: 58850810-58850810 on Build GRCh38
rs964169391 Chr.20: 58850609-58850609 on Build GRCh38
rs6128447 Chr.20: 58850433-58850433 on Build GRCh38
rs1028694462 Chr.20: 58850615-58850615 on Build GRCh38
rs981694327 Chr.20: 58850751-58850751 on Build GRCh38
rs1019165153 Chr.20: 58850595-58850595 on Build GRCh38
rs975672388 Chr.20: 58850627-58850627 on Build GRCh38
rs1043785163 Chr.20: 58850805-58850805 on Build GRCh38
rs534623373 Chr.20: 58850518-58850518 on Build GRCh38
rs981279708 Chr.20: 58850896-58850896 on Build GRCh38
rs1001161762 Chr.20: 58850839-58850840 on Build GRCh38
rs564494963 Chr.20: 58850838-58850838 on Build GRCh38
rs887401052 Chr.20: 58850858-58850858 on Build GRCh38
rs543667430 Chr.20: 58850801-58850801 on Build GRCh38
rs563960988 Chr.20: 58850847-58850847 on Build GRCh38
rs1010973728 Chr.20: 58850500-58850500 on Build GRCh38
rs547972515 Chr.20: 58850448-58850448 on Build GRCh38
rs563353511 Chr.20: 58850889-58850889 on Build GRCh38
rs896347854 Chr.20: 58850797-58850797 on Build GRCh38
rs76501839 Chr.20: 58850440-58850440 on Build GRCh38
rs116335020 Chr.20: 58850871-58850871 on Build GRCh38
rs939325525 Chr.20: 58850478-58850478 on Build GRCh38
rs574559187 Chr.20: 58850607-58850607 on Build GRCh38
rs908508041 Chr.20: 58850790-58850790 on Build GRCh38
rs529489816 Chr.20: 58850882-58850882 on Build GRCh38
rs577465140 Chr.20: 58850886-58850886 on Build GRCh38
rs1045426430 Chr.20: 58850808-58850808 on Build GRCh38
rs984594103 Chr.20: 58850697-58850697 on Build GRCh38
rs1055065526 Chr.20: 58850484-58850484 on Build GRCh38
rs960014312 Chr.20: 58850896-58850897 on Build GRCh38
rs919951595 Chr.20: 58850429-58850429 on Build GRCh38
rs951770688 Chr.20: 58850625-58850625 on Build GRCh38
rs552712421 Chr.20: 58850597-58850597 on Build GRCh38
rs940597954 Chr.20: 58850454-58850454 on Build GRCh38
rs753729444 Chr.20: 58850752-58850752 on Build GRCh38
rs1031052040 Chr.20: 58850540-58850540 on Build GRCh38
rs962119215 Chr.20: 58850891-58850891 on Build GRCh38
rs938765803 Chr.20: 58850792-58850792 on Build GRCh38
rs938727948 Chr.20: 58850444-58850444 on Build GRCh38
rs1028658417 Chr.20: 58850848-58850848 on Build GRCh38
rs1045611109 Chr.20: 58850460-58850460 on Build GRCh38
rs774600843 Chr.20: 58850474-58850474 on Build GRCh38
rs961869771 Chr.20: 58850734-58850734 on Build GRCh38
rs916620440 Chr.20: 58850800-58850800 on Build GRCh38
rs112469794 Chr.20: 58850407-58850407 on Build GRCh38
rs908563159 Chr.20: 58850893-58850893 on Build GRCh38

View Full Product Details