Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62546448-62546986 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf166-AS1
Entrez Gene ID
253868
Gene Name
C20orf166 antisense RNA 1
Gene Aliases
C20orf200, NCRNA00335
UniGene
Hs.337024
Gene Chromosome Location
Chr.20: 62543070-62551561 on Build GRCh38
Transcripts
NR_033263.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3963755 c.28G>T p.A10S Chr.20: 62546613-62546613 on Build GRCh38
3963754 c.128G>A p.S43N Chr.20: 62546513-62546513 on Build GRCh38
7281362 c.1A>G p.M1V Chr.20: 62546640-62546640 on Build GRCh38
3707914 c.14C>A p.A5D Chr.20: 62546627-62546627 on Build GRCh38
724867 c.157C>T p.L53L Chr.20: 62546484-62546484 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778292955 Chr.20: 62546854-62546854 on Build GRCh38
rs372930214 Chr.20: 62546908-62546908 on Build GRCh38
rs371643075 Chr.20: 62546502-62546502 on Build GRCh38
rs782467424 Chr.20: 62546473-62546473 on Build GRCh38
rs782194003 Chr.20: 62546630-62546630 on Build GRCh38
rs782636248 Chr.20: 62546635-62546635 on Build GRCh38
rs531999696 Chr.20: 62546705-62546705 on Build GRCh38
rs558894025 Chr.20: 62546831-62546831 on Build GRCh38
rs898618219 Chr.20: 62546920-62546920 on Build GRCh38
rs111651569 Chr.20: 62546612-62546612 on Build GRCh38
rs782203353 Chr.20: 62546613-62546613 on Build GRCh38
rs139545857 Chr.20: 62546556-62546556 on Build GRCh38
rs782635774 Chr.20: 62546565-62546565 on Build GRCh38
rs3934573 Chr.20: 62546738-62546738 on Build GRCh38
rs373427978 Chr.20: 62546607-62546607 on Build GRCh38
rs138465692 Chr.20: 62546503-62546503 on Build GRCh38
rs781942067 Chr.20: 62546661-62546661 on Build GRCh38
rs782717866 Chr.20: 62546514-62546514 on Build GRCh38
rs782455902 Chr.20: 62546614-62546614 on Build GRCh38
rs782032677 Chr.20: 62546510-62546510 on Build GRCh38
rs565494161 Chr.20: 62546520-62546520 on Build GRCh38
rs782763373 Chr.20: 62546536-62546536 on Build GRCh38
rs1004443183 Chr.20: 62546712-62546712 on Build GRCh38
rs782553146 Chr.20: 62546488-62546488 on Build GRCh38
rs782497169 Chr.20: 62546523-62546523 on Build GRCh38
rs1034151948 Chr.20: 62546935-62546935 on Build GRCh38
rs960673604 Chr.20: 62546796-62546796 on Build GRCh38
rs782790670 Chr.20: 62546487-62546488 on Build GRCh38
rs782362944 Chr.20: 62546495-62546495 on Build GRCh38
rs148327724 Chr.20: 62546519-62546519 on Build GRCh38
rs782233583 Chr.20: 62546655-62546655 on Build GRCh38
rs368086453 Chr.20: 62546472-62546472 on Build GRCh38
rs532376824 Chr.20: 62546742-62546742 on Build GRCh38
rs147120745 Chr.20: 62546485-62546485 on Build GRCh38
rs144283995 Chr.20: 62546518-62546518 on Build GRCh38
rs1043937496 Chr.20: 62546639-62546639 on Build GRCh38
rs540323240 Chr.20: 62546584-62546584 on Build GRCh38
rs574510320 Chr.20: 62546954-62546954 on Build GRCh38
rs923034984 Chr.20: 62546847-62546847 on Build GRCh38
rs782102926 Chr.20: 62546866-62546866 on Build GRCh38
rs943113085 Chr.20: 62546650-62546650 on Build GRCh38
rs543235753 Chr.20: 62546702-62546702 on Build GRCh38
rs782461710 Chr.20: 62546470-62546470 on Build GRCh38
rs1002394961 Chr.20: 62546925-62546925 on Build GRCh38
rs1038623830 Chr.20: 62546910-62546910 on Build GRCh38
rs45612436 Chr.20: 62546744-62546744 on Build GRCh38
rs374833108 Chr.20: 62546674-62546674 on Build GRCh38
rs782234966 Chr.20: 62546569-62546569 on Build GRCh38
rs782646866 Chr.20: 62546492-62546492 on Build GRCh38
rs186612346 Chr.20: 62546845-62546845 on Build GRCh38
rs368157249 Chr.20: 62546682-62546682 on Build GRCh38
rs191155016 Chr.20: 62546855-62546855 on Build GRCh38
rs782679584 Chr.20: 62546618-62546618 on Build GRCh38
rs781904672 Chr.20: 62546493-62546494 on Build GRCh38
rs887344907 Chr.20: 62546711-62546711 on Build GRCh38
rs782085141 Chr.20: 62546603-62546603 on Build GRCh38
rs753087150 Chr.20: 62546841-62546841 on Build GRCh38
rs140092114 Chr.20: 62546620-62546620 on Build GRCh38
rs1033771952 Chr.20: 62546787-62546787 on Build GRCh38
rs903710202 Chr.20: 62546643-62546643 on Build GRCh38
rs782734089 Chr.20: 62546909-62546909 on Build GRCh38
rs782573666 Chr.20: 62546480-62546480 on Build GRCh38
rs576312365 Chr.20: 62546573-62546573 on Build GRCh38
rs1047366315 Chr.20: 62546882-62546882 on Build GRCh38
rs782215554 Chr.20: 62546602-62546603 on Build GRCh38
rs782450421 Chr.20: 62546549-62546549 on Build GRCh38
rs763129135 Chr.20: 62546487-62546487 on Build GRCh38
rs782428314 Chr.20: 62546574-62546574 on Build GRCh38
rs960768651 Chr.20: 62546936-62546936 on Build GRCh38
rs886144340 Chr.20: 62546898-62546898 on Build GRCh38
rs782755547 Chr.20: 62546690-62546690 on Build GRCh38
rs782342391 Chr.20: 62546658-62546658 on Build GRCh38
rs782400148 Chr.20: 62546624-62546624 on Build GRCh38
rs45553940 Chr.20: 62546805-62546805 on Build GRCh38
rs913933557 Chr.20: 62546798-62546798 on Build GRCh38
rs782649513 Chr.20: 62546602-62546602 on Build GRCh38
rs782565004 Chr.20: 62546551-62546551 on Build GRCh38
rs144142894 Chr.20: 62546598-62546598 on Build GRCh38
rs781925442 Chr.20: 62546515-62546515 on Build GRCh38
rs141396933 Chr.20: 62546548-62546548 on Build GRCh38
rs782243583 Chr.20: 62546494-62546494 on Build GRCh38
rs267606044 Chr.20: 62546507-62546507 on Build GRCh38
rs782601411 Chr.20: 62546500-62546500 on Build GRCh38
rs782028004 Chr.20: 62546580-62546580 on Build GRCh38
rs6062279 Chr.20: 62546736-62546736 on Build GRCh38
rs561942090 Chr.20: 62546809-62546809 on Build GRCh38
rs202060553 Chr.20: 62546570-62546570 on Build GRCh38
rs782506758 Chr.20: 62546499-62546499 on Build GRCh38
rs529826317 Chr.20: 62546802-62546802 on Build GRCh38
rs537324410 Chr.20: 62546813-62546813 on Build GRCh38
rs782016126 Chr.20: 62546675-62546675 on Build GRCh38
rs182690704 Chr.20: 62546797-62546797 on Build GRCh38

View Full Product Details