Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 13845705-13846204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4F29P
Entrez Gene ID
54055
Gene Name
cytochrome P450 family 4 subfamily F member 29, pseudogene
Gene Aliases
4F-se4[6:7:8], C21orf15, CYP4F-se4[6:7:8], CYP4F3LP
UniGene
Hs.580910
Gene Chromosome Location
Chr.21: 13843133-13848364 on Build GRCh38
Transcripts
NR_026755.1

SNP Detail

SNP ID Chromosome Location
rs200326347 Chr.21: 13846177-13846178 on Build GRCh38
rs975242093 Chr.21: 13846112-13846112 on Build GRCh38
rs981811647 Chr.21: 13845877-13845877 on Build GRCh38
rs3117465 Chr.21: 13846095-13846095 on Build GRCh38
rs112133877 Chr.21: 13846141-13846141 on Build GRCh38
rs9975091 Chr.21: 13845871-13845871 on Build GRCh38
rs8133062 Chr.21: 13845782-13845782 on Build GRCh38
rs536510307 Chr.21: 13846000-13846000 on Build GRCh38
rs920886469 Chr.21: 13846155-13846155 on Build GRCh38
rs1022204916 Chr.21: 13846079-13846079 on Build GRCh38
rs141923031 Chr.21: 13846052-13846052 on Build GRCh38
rs2334716 Chr.21: 13846158-13846158 on Build GRCh38
rs145024135 Chr.21: 13845854-13845854 on Build GRCh38
rs948183110 Chr.21: 13846165-13846165 on Build GRCh38
rs113320689 Chr.21: 13846059-13846059 on Build GRCh38
rs1016402996 Chr.21: 13845859-13845859 on Build GRCh38
rs535563733 Chr.21: 13845813-13845813 on Build GRCh38
rs922009450 Chr.21: 13846114-13846114 on Build GRCh38
rs546793098 Chr.21: 13846030-13846030 on Build GRCh38
rs397843084 Chr.21: 13845807-13845807 on Build GRCh38
rs933360044 Chr.21: 13845836-13845836 on Build GRCh38
rs551454455 Chr.21: 13845752-13845752 on Build GRCh38
rs886451397 Chr.21: 13846019-13846019 on Build GRCh38
rs1023396689 Chr.21: 13845872-13845872 on Build GRCh38
rs374779477 Chr.21: 13845953-13845953 on Build GRCh38
rs60989409 Chr.21: 13846066-13846066 on Build GRCh38
rs899347611 Chr.21: 13845862-13845862 on Build GRCh38
rs138803058 Chr.21: 13845858-13845858 on Build GRCh38
rs753712910 Chr.21: 13845760-13845760 on Build GRCh38
rs573777148 Chr.21: 13845895-13845895 on Build GRCh38
rs2334714 Chr.21: 13846104-13846104 on Build GRCh38
rs2775976 Chr.21: 13845901-13845901 on Build GRCh38
rs560409236 Chr.21: 13846048-13846048 on Build GRCh38
rs942547722 Chr.21: 13845932-13845932 on Build GRCh38
rs371717148 Chr.21: 13846173-13846173 on Build GRCh38
rs3929682 Chr.21: 13845914-13845914 on Build GRCh38
rs555034718 Chr.21: 13845823-13845823 on Build GRCh38
rs536970062 Chr.21: 13845789-13845789 on Build GRCh38
rs2092890 Chr.21: 13846070-13846070 on Build GRCh38
rs2878250 Chr.21: 13846087-13846087 on Build GRCh38
rs4046605 Chr.21: 13846178-13846179 on Build GRCh38
rs2942534 Chr.21: 13845808-13845808 on Build GRCh38
rs750810111 Chr.21: 13846041-13846041 on Build GRCh38
rs35082157 Chr.21: 13845966-13845967 on Build GRCh38
rs371180233 Chr.21: 13845915-13845915 on Build GRCh38
rs546854467 Chr.21: 13845781-13845781 on Build GRCh38
rs907828959 Chr.21: 13846130-13846130 on Build GRCh38
rs533297551 Chr.21: 13846027-13846027 on Build GRCh38
rs35564392 Chr.21: 13845744-13845744 on Build GRCh38
rs533156209 Chr.21: 13845767-13845767 on Build GRCh38
rs940693748 Chr.21: 13846137-13846137 on Build GRCh38
rs576395906 Chr.21: 13845973-13845973 on Build GRCh38
rs947451159 Chr.21: 13845820-13845820 on Build GRCh38
rs954762363 Chr.21: 13846127-13846127 on Build GRCh38
rs2334715 Chr.21: 13846115-13846115 on Build GRCh38
rs182215673 Chr.21: 13846065-13846065 on Build GRCh38
rs1038359867 Chr.21: 13846020-13846020 on Build GRCh38
rs555484579 Chr.21: 13846058-13846058 on Build GRCh38
rs989823672 Chr.21: 13845911-13845911 on Build GRCh38
rs568850635 Chr.21: 13845830-13845830 on Build GRCh38
rs989358470 Chr.21: 13845806-13845806 on Build GRCh38
rs189033877 Chr.21: 13845855-13845855 on Build GRCh38
rs779830869 Chr.21: 13846107-13846107 on Build GRCh38
rs1012454618 Chr.21: 13846036-13846036 on Build GRCh38
rs149481432 Chr.21: 13846129-13846129 on Build GRCh38
rs564361948 Chr.21: 13846002-13846002 on Build GRCh38
rs397843083 Chr.21: 13845803-13845803 on Build GRCh38
rs145869071 Chr.21: 13845904-13845904 on Build GRCh38
rs915073993 Chr.21: 13845809-13845809 on Build GRCh38
rs553981973 Chr.21: 13845873-13845873 on Build GRCh38

View Full Product Details