Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 13847948-13848448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4F29P
Entrez Gene ID
54055
Gene Name
cytochrome P450 family 4 subfamily F member 29, pseudogene
Gene Aliases
4F-se4[6:7:8], C21orf15, CYP4F-se4[6:7:8], CYP4F3LP
UniGene
Hs.580910
Gene Chromosome Location
Chr.21: 13843133-13848364 on Build GRCh38
Transcripts
NR_026755.1

SNP Detail

SNP ID Chromosome Location
rs1020188023 Chr.21: 13848055-13848055 on Build GRCh38
rs1027045736 Chr.21: 13847996-13847996 on Build GRCh38
rs937526906 Chr.21: 13848214-13848214 on Build GRCh38
rs9975493 Chr.21: 13848240-13848240 on Build GRCh38
rs949798318 Chr.21: 13848395-13848395 on Build GRCh38
rs192704387 Chr.21: 13848388-13848388 on Build GRCh38
rs371245258 Chr.21: 13847979-13847979 on Build GRCh38
rs376997388 Chr.21: 13848023-13848023 on Build GRCh38
rs528567543 Chr.21: 13848104-13848104 on Build GRCh38
rs569602832 Chr.21: 13848227-13848227 on Build GRCh38
rs865869440 Chr.21: 13848362-13848362 on Build GRCh38
rs967239648 Chr.21: 13848077-13848077 on Build GRCh38
rs373029095 Chr.21: 13848399-13848399 on Build GRCh38
rs985556097 Chr.21: 13848112-13848112 on Build GRCh38
rs558305545 Chr.21: 13848249-13848249 on Build GRCh38
rs1007491272 Chr.21: 13848425-13848425 on Build GRCh38
rs911337259 Chr.21: 13848118-13848118 on Build GRCh38
rs965386086 Chr.21: 13848125-13848125 on Build GRCh38
rs548394524 Chr.21: 13848115-13848115 on Build GRCh38
rs1007831899 Chr.21: 13848266-13848266 on Build GRCh38
rs550997372 Chr.21: 13848173-13848173 on Build GRCh38
rs553949670 Chr.21: 13848207-13848207 on Build GRCh38
rs895475235 Chr.21: 13848014-13848014 on Build GRCh38
rs151314334 Chr.21: 13848132-13848132 on Build GRCh38
rs1048678652 Chr.21: 13848202-13848202 on Build GRCh38
rs1048563595 Chr.21: 13847976-13847976 on Build GRCh38
rs766809053 Chr.21: 13848201-13848201 on Build GRCh38
rs552788810 Chr.21: 13847990-13847990 on Build GRCh38
rs186938655 Chr.21: 13848398-13848398 on Build GRCh38
rs1018345281 Chr.21: 13848290-13848290 on Build GRCh38
rs367929431 Chr.21: 13848401-13848401 on Build GRCh38
rs111310465 Chr.21: 13848042-13848042 on Build GRCh38
rs984363360 Chr.21: 13848365-13848365 on Build GRCh38
rs780823769 Chr.21: 13848260-13848260 on Build GRCh38
rs969903507 Chr.21: 13847997-13847997 on Build GRCh38
rs183715012 Chr.21: 13848389-13848389 on Build GRCh38
rs1055959196 Chr.21: 13848008-13848008 on Build GRCh38
rs951367544 Chr.21: 13848357-13848357 on Build GRCh38
rs796418222 Chr.21: 13848113-13848113 on Build GRCh38
rs554359466 Chr.21: 13848328-13848328 on Build GRCh38
rs796302847 Chr.21: 13848119-13848119 on Build GRCh38
rs889063659 Chr.21: 13848412-13848412 on Build GRCh38
rs116024964 Chr.21: 13848250-13848250 on Build GRCh38
rs763432576 Chr.21: 13848165-13848165 on Build GRCh38
rs113330813 Chr.21: 13848295-13848299 on Build GRCh38
rs751226920 Chr.21: 13848238-13848238 on Build GRCh38
rs191578427 Chr.21: 13848419-13848419 on Build GRCh38
rs918607379 Chr.21: 13848154-13848154 on Build GRCh38
rs112067023 Chr.21: 13848052-13848052 on Build GRCh38
rs78268950 Chr.21: 13848346-13848346 on Build GRCh38
rs756542681 Chr.21: 13848263-13848263 on Build GRCh38
rs568400024 Chr.21: 13848124-13848124 on Build GRCh38
rs765174381 Chr.21: 13848375-13848375 on Build GRCh38
rs1000088475 Chr.21: 13848095-13848095 on Build GRCh38
rs796985096 Chr.21: 13848102-13848102 on Build GRCh38
rs949843615 Chr.21: 13848232-13848232 on Build GRCh38
rs73150992 Chr.21: 13848074-13848074 on Build GRCh38
rs534676121 Chr.21: 13848322-13848322 on Build GRCh38
rs7283493 Chr.21: 13848035-13848035 on Build GRCh38
rs980376728 Chr.21: 13848004-13848004 on Build GRCh38
rs73150993 Chr.21: 13848092-13848092 on Build GRCh38
rs528686013 Chr.21: 13848426-13848426 on Build GRCh38
rs532883235 Chr.21: 13847974-13847974 on Build GRCh38
rs560003682 Chr.21: 13848050-13848050 on Build GRCh38
rs1054280388 Chr.21: 13848402-13848402 on Build GRCh38
rs796132208 Chr.21: 13848108-13848108 on Build GRCh38
rs901680640 Chr.21: 13848428-13848428 on Build GRCh38
rs1041754359 Chr.21: 13848238-13848238 on Build GRCh38
rs910174471 Chr.21: 13848370-13848370 on Build GRCh38

View Full Product Details