Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 17885352-17885859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHODL
Entrez Gene ID
140578
Gene Name
chondrolectin
Gene Aliases
C21orf68, MT75, PRED12
UniGene
Hs.283725
Gene Chromosome Location
Chr.21: 17819329-18267371 on Build GRCh38
Transcripts
XM_011529457.2 NR_024354.1

Gene Symbol
CHODL-AS1
Entrez Gene ID
54075
Gene Name
CHODL antisense RNA 1
Gene Aliases
C21orf39, NCRNA00157
UniGene
Hs.404306
Gene Chromosome Location
Chr.21: 17835672-17885608 on Build GRCh38
Transcripts
XM_011529457.2 NR_024354.1

SNP Detail

SNP ID Chromosome Location
rs911593058 Chr.21: 17885764-17885764 on Build GRCh38
rs781440809 Chr.21: 17885629-17885631 on Build GRCh38
rs966797452 Chr.21: 17885541-17885541 on Build GRCh38
rs1053919382 Chr.21: 17885626-17885626 on Build GRCh38
rs902844333 Chr.21: 17885672-17885672 on Build GRCh38
rs757301481 Chr.21: 17885620-17885620 on Build GRCh38
rs931214165 Chr.21: 17885429-17885429 on Build GRCh38
rs1047396014 Chr.21: 17885395-17885395 on Build GRCh38
rs528925723 Chr.21: 17885757-17885757 on Build GRCh38
rs139243368 Chr.21: 17885498-17885498 on Build GRCh38
rs542944235 Chr.21: 17885741-17885741 on Build GRCh38
rs542997272 Chr.21: 17885689-17885689 on Build GRCh38
rs570834141 Chr.21: 17885732-17885732 on Build GRCh38
rs186394574 Chr.21: 17885583-17885583 on Build GRCh38
rs919759922 Chr.21: 17885376-17885376 on Build GRCh38
rs946542434 Chr.21: 17885795-17885795 on Build GRCh38
rs1035020576 Chr.21: 17885591-17885591 on Build GRCh38
rs190438945 Chr.21: 17885775-17885775 on Build GRCh38
rs982703189 Chr.21: 17885759-17885759 on Build GRCh38
rs75659899 Chr.21: 17885779-17885779 on Build GRCh38
rs4347933 Chr.21: 17885469-17885469 on Build GRCh38
rs1047458438 Chr.21: 17885444-17885444 on Build GRCh38
rs1047384518 Chr.21: 17885735-17885735 on Build GRCh38
rs561471928 Chr.21: 17885724-17885724 on Build GRCh38
rs1054313563 Chr.21: 17885812-17885812 on Build GRCh38
rs1035338072 Chr.21: 17885683-17885683 on Build GRCh38
rs932689284 Chr.21: 17885792-17885792 on Build GRCh38
rs932738559 Chr.21: 17885586-17885586 on Build GRCh38
rs150015789 Chr.21: 17885744-17885744 on Build GRCh38
rs893970711 Chr.21: 17885832-17885832 on Build GRCh38
rs1021664016 Chr.21: 17885671-17885671 on Build GRCh38
rs999491902 Chr.21: 17885545-17885545 on Build GRCh38
rs1009813948 Chr.21: 17885657-17885657 on Build GRCh38
rs879266337 Chr.21: 17885609-17885609 on Build GRCh38
rs779063513 Chr.21: 17885633-17885633 on Build GRCh38
rs566329130 Chr.21: 17885674-17885674 on Build GRCh38
rs551286880 Chr.21: 17885714-17885714 on Build GRCh38
rs547208247 Chr.21: 17885766-17885766 on Build GRCh38
rs545634698 Chr.21: 17885540-17885540 on Build GRCh38
rs181959335 Chr.21: 17885505-17885505 on Build GRCh38
rs991209554 Chr.21: 17885723-17885723 on Build GRCh38
rs555938802 Chr.21: 17885428-17885428 on Build GRCh38
rs574166910 Chr.21: 17885443-17885443 on Build GRCh38
rs1026813542 Chr.21: 17885753-17885753 on Build GRCh38
rs79743285 Chr.21: 17885784-17885784 on Build GRCh38
rs943947806 Chr.21: 17885769-17885769 on Build GRCh38
rs908540221 Chr.21: 17885447-17885447 on Build GRCh38

View Full Product Details