Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 32732769-32733271 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAXBP1-AS1
Entrez Gene ID
100506215
Gene Name
PAXBP1 antisense RNA 1
Gene Aliases
GCFC1-AS1
UniGene
Hs.657123
Gene Chromosome Location
Chr.21: 32728114-32743122 on Build GRCh38
Transcripts
NR_038879.1 NR_038880.1

SNP Detail

SNP ID Chromosome Location
rs558723766 Chr.21: 32732921-32732921 on Build GRCh38
rs931044988 Chr.21: 32732935-32732935 on Build GRCh38
rs948827301 Chr.21: 32733202-32733202 on Build GRCh38
rs950784352 Chr.21: 32733019-32733019 on Build GRCh38
rs189169831 Chr.21: 32733014-32733014 on Build GRCh38
rs939133414 Chr.21: 32733244-32733244 on Build GRCh38
rs147305194 Chr.21: 32732836-32732836 on Build GRCh38
rs576645683 Chr.21: 32732835-32732835 on Build GRCh38
rs1027216624 Chr.21: 32733226-32733226 on Build GRCh38
rs892455415 Chr.21: 32733142-32733142 on Build GRCh38
rs1000885859 Chr.21: 32733120-32733120 on Build GRCh38
rs964001922 Chr.21: 32732892-32732892 on Build GRCh38
rs926443942 Chr.21: 32733224-32733224 on Build GRCh38
rs964281281 Chr.21: 32733208-32733208 on Build GRCh38
rs1007755704 Chr.21: 32732937-32732937 on Build GRCh38
rs985194903 Chr.21: 32733098-32733102 on Build GRCh38
rs7283541 Chr.21: 32732972-32732972 on Build GRCh38
rs919788118 Chr.21: 32732928-32732928 on Build GRCh38
rs1056828273 Chr.21: 32733022-32733022 on Build GRCh38
rs1031082517 Chr.21: 32733141-32733141 on Build GRCh38
rs896693964 Chr.21: 32733033-32733033 on Build GRCh38
rs956726798 Chr.21: 32732798-32732798 on Build GRCh38
rs575144976 Chr.21: 32732893-32732893 on Build GRCh38
rs1017743197 Chr.21: 32732969-32732969 on Build GRCh38
rs1045614524 Chr.21: 32733097-32733097 on Build GRCh38
rs976307264 Chr.21: 32733002-32733002 on Build GRCh38
rs1022082657 Chr.21: 32732905-32732905 on Build GRCh38
rs904047691 Chr.21: 32733100-32733100 on Build GRCh38
rs574749447 Chr.21: 32733112-32733112 on Build GRCh38
rs1019613488 Chr.21: 32733178-32733178 on Build GRCh38
rs1008149340 Chr.21: 32733173-32733173 on Build GRCh38
rs1003478099 Chr.21: 32732794-32732794 on Build GRCh38
rs892144625 Chr.21: 32732869-32732869 on Build GRCh38
rs908205024 Chr.21: 32732986-32732986 on Build GRCh38
rs980156626 Chr.21: 32733221-32733221 on Build GRCh38
rs541929928 Chr.21: 32733201-32733201 on Build GRCh38
rs975368779 Chr.21: 32733215-32733215 on Build GRCh38
rs559492972 Chr.21: 32733086-32733086 on Build GRCh38
rs909325329 Chr.21: 32733182-32733183 on Build GRCh38

View Full Product Details