Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 37220557-37221081 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSCR9
Entrez Gene ID
257203
Gene Name
Down syndrome critical region 9 (non-protein coding)
Gene Aliases
NCRNA00038
UniGene
Hs.505159
Gene Chromosome Location
Chr.21: 37208503-37221736 on Build GRCh38
Transcripts
NR_026719.2

SNP Detail

SNP ID Chromosome Location
rs1009569532 Chr.21: 37220949-37220949 on Build GRCh38
rs369326809 Chr.21: 37220962-37220962 on Build GRCh38
rs375226086 Chr.21: 37220790-37220790 on Build GRCh38
rs62226495 Chr.21: 37220856-37220856 on Build GRCh38
rs73220508 Chr.21: 37220623-37220623 on Build GRCh38
rs896515229 Chr.21: 37220932-37220932 on Build GRCh38
rs768662600 Chr.21: 37220774-37220774 on Build GRCh38
rs985809235 Chr.21: 37220698-37220698 on Build GRCh38
rs988115459 Chr.21: 37220933-37220933 on Build GRCh38
rs937172152 Chr.21: 37220993-37220993 on Build GRCh38
rs1014204108 Chr.21: 37221042-37221042 on Build GRCh38
rs953245618 Chr.21: 37220695-37220695 on Build GRCh38
rs373205819 Chr.21: 37220680-37220680 on Build GRCh38
rs1056401865 Chr.21: 37220921-37220921 on Build GRCh38
rs966140423 Chr.21: 37221056-37221056 on Build GRCh38
rs922152403 Chr.21: 37220599-37220599 on Build GRCh38
rs774469536 Chr.21: 37221016-37221016 on Build GRCh38
rs1032682074 Chr.21: 37220822-37220822 on Build GRCh38
rs930836587 Chr.21: 37220780-37220780 on Build GRCh38
rs376455439 Chr.21: 37221049-37221049 on Build GRCh38
rs911507384 Chr.21: 37220707-37220707 on Build GRCh38
rs1032956043 Chr.21: 37220673-37220673 on Build GRCh38
rs117403436 Chr.21: 37220972-37220972 on Build GRCh38
rs978571849 Chr.21: 37220634-37220634 on Build GRCh38
rs944479890 Chr.21: 37220729-37220729 on Build GRCh38
rs1007448532 Chr.21: 37221032-37221032 on Build GRCh38
rs1019975150 Chr.21: 37220809-37220809 on Build GRCh38
rs905471338 Chr.21: 37220832-37220832 on Build GRCh38
rs903923165 Chr.21: 37220965-37220965 on Build GRCh38
rs907904378 Chr.21: 37220957-37220957 on Build GRCh38
rs938225768 Chr.21: 37220916-37220916 on Build GRCh38
rs1055614693 Chr.21: 37221003-37221003 on Build GRCh38
rs974966563 Chr.21: 37220582-37220582 on Build GRCh38
rs953276219 Chr.21: 37221021-37221021 on Build GRCh38
rs73905443 Chr.21: 37220738-37220738 on Build GRCh38
rs1049192026 Chr.21: 37220794-37220794 on Build GRCh38
rs184802249 Chr.21: 37220885-37220885 on Build GRCh38
rs1034277529 Chr.21: 37220742-37220742 on Build GRCh38
rs967438700 Chr.21: 37220812-37220812 on Build GRCh38
rs780096554 Chr.21: 37220661-37220661 on Build GRCh38
rs886420522 Chr.21: 37220662-37220662 on Build GRCh38
rs978518373 Chr.21: 37220821-37220821 on Build GRCh38

View Full Product Details