Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 40382974-40383476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSCAM
Entrez Gene ID
1826
Gene Name
DS cell adhesion molecule
Gene Aliases
CHD2, CHD2-42, CHD2-52
UniGene
Hs.397800
Gene Chromosome Location
Chr.21: 40010999-40847113 on Build GRCh38
Transcripts
NM_001271534.1 NM_001389.3 NR_073202.1 NR_038896.1 NR_038898.1 NR_038899.1 NR_038900.1

Gene Symbol
DSCAM-AS1
Entrez Gene ID
100506492
Gene Name
DSCAM antisense RNA 1
Gene Aliases
M41
UniGene
Hs.667053
Gene Chromosome Location
Chr.21: 40383083-40385358 on Build GRCh38
Transcripts
NM_001271534.1 NM_001389.3 NR_073202.1 NR_038896.1 NR_038898.1 NR_038899.1 NR_038900.1

SNP Detail

SNP ID Chromosome Location
rs918815753 Chr.21: 40383139-40383139 on Build GRCh38
rs747821923 Chr.21: 40383099-40383099 on Build GRCh38
rs1026407000 Chr.21: 40383351-40383351 on Build GRCh38
rs1011138030 Chr.21: 40383205-40383205 on Build GRCh38
rs943425244 Chr.21: 40383171-40383171 on Build GRCh38
rs879218548 Chr.21: 40383384-40383384 on Build GRCh38
rs78310895 Chr.21: 40383426-40383426 on Build GRCh38
rs990616237 Chr.21: 40383311-40383311 on Build GRCh38
rs535448874 Chr.21: 40383447-40383447 on Build GRCh38
rs934248419 Chr.21: 40383338-40383338 on Build GRCh38
rs1024355514 Chr.21: 40383051-40383051 on Build GRCh38
rs372431319 Chr.21: 40383295-40383295 on Build GRCh38
rs1054940791 Chr.21: 40383183-40383183 on Build GRCh38
rs749211381 Chr.21: 40383389-40383389 on Build GRCh38
rs935920219 Chr.21: 40383232-40383232 on Build GRCh38
rs2017995 Chr.21: 40383304-40383304 on Build GRCh38
rs912016091 Chr.21: 40383379-40383379 on Build GRCh38
rs959135656 Chr.21: 40383071-40383071 on Build GRCh38
rs981428254 Chr.21: 40383105-40383105 on Build GRCh38
rs987961079 Chr.21: 40383375-40383375 on Build GRCh38
rs1021288702 Chr.21: 40383014-40383014 on Build GRCh38
rs532885945 Chr.21: 40383368-40383368 on Build GRCh38
rs573197164 Chr.21: 40383145-40383145 on Build GRCh38
rs546324423 Chr.21: 40383126-40383126 on Build GRCh38
rs1052908770 Chr.21: 40383235-40383235 on Build GRCh38
rs747797160 Chr.21: 40383341-40383345 on Build GRCh38
rs902453989 Chr.21: 40383252-40383252 on Build GRCh38
rs973639184 Chr.21: 40383341-40383350 on Build GRCh38
rs546678118 Chr.21: 40383425-40383425 on Build GRCh38
rs1054569691 Chr.21: 40383412-40383412 on Build GRCh38
rs779114125 Chr.21: 40383397-40383397 on Build GRCh38
rs911844315 Chr.21: 40383166-40383166 on Build GRCh38
rs893680474 Chr.21: 40383200-40383200 on Build GRCh38
rs777302834 Chr.21: 40383344-40383344 on Build GRCh38
rs369632545 Chr.21: 40383404-40383404 on Build GRCh38
rs1034621449 Chr.21: 40383047-40383047 on Build GRCh38
rs941514947 Chr.21: 40383111-40383111 on Build GRCh38
rs973298155 Chr.21: 40383329-40383329 on Build GRCh38
rs963499504 Chr.21: 40383132-40383132 on Build GRCh38
rs551181620 Chr.21: 40383253-40383253 on Build GRCh38
rs542122299 Chr.21: 40383150-40383150 on Build GRCh38
rs1017090171 Chr.21: 40383320-40383320 on Build GRCh38
rs918729321 Chr.21: 40383332-40383332 on Build GRCh38
rs911261259 Chr.21: 40383395-40383395 on Build GRCh38
rs943353622 Chr.21: 40383383-40383383 on Build GRCh38
rs946125832 Chr.21: 40383427-40383427 on Build GRCh38
rs780774157 Chr.21: 40383076-40383076 on Build GRCh38
rs981731118 Chr.21: 40383039-40383039 on Build GRCh38
rs539757997 Chr.21: 40383017-40383017 on Build GRCh38
rs987581955 Chr.21: 40383157-40383157 on Build GRCh38
rs962933381 Chr.21: 40383325-40383325 on Build GRCh38
rs778703115 Chr.21: 40383109-40383109 on Build GRCh38
rs1039357050 Chr.21: 40383180-40383180 on Build GRCh38
rs537627520 Chr.21: 40383013-40383013 on Build GRCh38
rs142116786 Chr.21: 40383144-40383144 on Build GRCh38
rs562387571 Chr.21: 40383156-40383156 on Build GRCh38
rs1020902707 Chr.21: 40383210-40383210 on Build GRCh38
rs1033875899 Chr.21: 40383319-40383319 on Build GRCh38
rs531262110 Chr.21: 40383187-40383187 on Build GRCh38
rs368872994 Chr.21: 40383062-40383062 on Build GRCh38

View Full Product Details