Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 45419562-45420058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL18A1
Entrez Gene ID
80781
Gene Name
collagen type XVIII alpha 1 chain
Gene Aliases
KNO, KNO1, KS
UniGene
Hs.517356
Gene Chromosome Location
Chr.21: 45405137-45513720 on Build GRCh38
Transcripts
NM_130445.3 NR_027498.1 NR_028082.1

Gene Symbol
COL18A1-AS1
Entrez Gene ID
378832
Gene Name
COL18A1 antisense RNA 1
Gene Aliases
C21orf123, NCRNA00175, PRED80
UniGene
Hs.517353
Gene Chromosome Location
Chr.21: 45419716-45425070 on Build GRCh38
Transcripts
NM_130445.3 NR_027498.1 NR_028082.1

SNP Detail

SNP ID Chromosome Location
rs542357171 Chr.21: 45419641-45419641 on Build GRCh38
rs955711395 Chr.21: 45419696-45419696 on Build GRCh38
rs897536110 Chr.21: 45420008-45420008 on Build GRCh38
rs2838915 Chr.21: 45419655-45419655 on Build GRCh38
rs946013541 Chr.21: 45419885-45419885 on Build GRCh38
rs985205856 Chr.21: 45419984-45419984 on Build GRCh38
rs771323306 Chr.21: 45419936-45419936 on Build GRCh38
rs971644751 Chr.21: 45419747-45419747 on Build GRCh38
rs879273804 Chr.21: 45419753-45419753 on Build GRCh38
rs930781714 Chr.21: 45419595-45419595 on Build GRCh38
rs567780003 Chr.21: 45419839-45419839 on Build GRCh38
rs986041212 Chr.21: 45419847-45419847 on Build GRCh38
rs994452098 Chr.21: 45419832-45419832 on Build GRCh38
rs544641483 Chr.21: 45419802-45419802 on Build GRCh38
rs1022828647 Chr.21: 45419739-45419739 on Build GRCh38
rs374986778 Chr.21: 45419944-45419944 on Build GRCh38
rs372212379 Chr.21: 45420021-45420021 on Build GRCh38
rs919704189 Chr.21: 45419592-45419592 on Build GRCh38
rs578204807 Chr.21: 45419620-45419620 on Build GRCh38
rs759243904 Chr.21: 45419988-45419988 on Build GRCh38
rs553633562 Chr.21: 45419919-45419919 on Build GRCh38
rs774914859 Chr.21: 45419962-45419962 on Build GRCh38
rs998827186 Chr.21: 45419649-45419649 on Build GRCh38
rs887528094 Chr.21: 45419820-45419820 on Build GRCh38
rs147390985 Chr.21: 45419937-45419937 on Build GRCh38
rs913266595 Chr.21: 45419872-45419872 on Build GRCh38
rs192485646 Chr.21: 45419829-45419829 on Build GRCh38
rs1014503788 Chr.21: 45419630-45419630 on Build GRCh38
rs1018404896 Chr.21: 45419865-45419865 on Build GRCh38
rs957463177 Chr.21: 45419866-45419866 on Build GRCh38
rs183966161 Chr.21: 45419912-45419912 on Build GRCh38
rs1003131737 Chr.21: 45419917-45419917 on Build GRCh38
rs966226869 Chr.21: 45419990-45419990 on Build GRCh38
rs551744075 Chr.21: 45419825-45419825 on Build GRCh38
rs773711538 Chr.21: 45419867-45419867 on Build GRCh38
rs749764181 Chr.21: 45419928-45419928 on Build GRCh38
rs945960731 Chr.21: 45419797-45419797 on Build GRCh38
rs150091879 Chr.21: 45419818-45419818 on Build GRCh38
rs1054423070 Chr.21: 45419852-45419852 on Build GRCh38
rs928548878 Chr.21: 45419924-45419924 on Build GRCh38
rs1015537238 Chr.21: 45419789-45419789 on Build GRCh38
rs78934945 Chr.21: 45419836-45419836 on Build GRCh38
rs571214018 Chr.21: 45419723-45419723 on Build GRCh38
rs553670575 Chr.21: 45419594-45419594 on Build GRCh38
rs936036402 Chr.21: 45419740-45419740 on Build GRCh38
rs188889615 Chr.21: 45419795-45419795 on Build GRCh38
rs2838917 Chr.21: 45419989-45419989 on Build GRCh38
rs907010862 Chr.21: 45419916-45419916 on Build GRCh38
rs563169776 Chr.21: 45419626-45419626 on Build GRCh38
rs985603175 Chr.21: 45419702-45419702 on Build GRCh38
rs1046048361 Chr.21: 45419798-45419798 on Build GRCh38
rs575286723 Chr.21: 45419629-45419629 on Build GRCh38
rs138612388 Chr.21: 45419830-45419830 on Build GRCh38
rs367601118 Chr.21: 45419867-45419867 on Build GRCh38
rs944890151 Chr.21: 45419835-45419835 on Build GRCh38
rs753745942 Chr.21: 45419618-45419618 on Build GRCh38
rs556911571 Chr.21: 45419943-45419943 on Build GRCh38
rs865913347 Chr.21: 45419799-45419799 on Build GRCh38
rs1002753306 Chr.21: 45419784-45419784 on Build GRCh38
rs560652820 Chr.21: 45419654-45419654 on Build GRCh38
rs769784920 Chr.21: 45419841-45419841 on Build GRCh38
rs2838916 Chr.21: 45419710-45419710 on Build GRCh38
rs578184259 Chr.21: 45419925-45419925 on Build GRCh38
rs1013160172 Chr.21: 45419733-45419733 on Build GRCh38
rs140588754 Chr.21: 45419622-45419622 on Build GRCh38
rs577484521 Chr.21: 45419762-45419762 on Build GRCh38
rs1044876795 Chr.21: 45419913-45419913 on Build GRCh38

View Full Product Details