Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 45422304-45422797 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL18A1
Entrez Gene ID
80781
Gene Name
collagen type XVIII alpha 1 chain
Gene Aliases
KNO, KNO1, KS
UniGene
Hs.517356
Gene Chromosome Location
Chr.21: 45405137-45513720 on Build GRCh38
Transcripts
NM_130445.3 NR_027498.1 NR_028082.1

Gene Symbol
COL18A1-AS1
Entrez Gene ID
378832
Gene Name
COL18A1 antisense RNA 1
Gene Aliases
C21orf123, NCRNA00175, PRED80
UniGene
Hs.517353
Gene Chromosome Location
Chr.21: 45419716-45425070 on Build GRCh38
Transcripts
NM_130445.3 NR_027498.1 NR_028082.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6026681 c.35G>T p.G12V Chr.21: 45422530-45422530 on Build GRCh38
6632608 c.40A>G p.R14G Chr.21: 45422525-45422525 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758279418 Chr.21: 45422510-45422510 on Build GRCh38
rs371332266 Chr.21: 45422576-45422576 on Build GRCh38
rs904054294 Chr.21: 45422637-45422637 on Build GRCh38
rs932252588 Chr.21: 45422626-45422626 on Build GRCh38
rs765563449 Chr.21: 45422470-45422470 on Build GRCh38
rs1024854981 Chr.21: 45422335-45422335 on Build GRCh38
rs973717565 Chr.21: 45422623-45422623 on Build GRCh38
rs375981940 Chr.21: 45422490-45422490 on Build GRCh38
rs374608012 Chr.21: 45422519-45422519 on Build GRCh38
rs778844035 Chr.21: 45422502-45422502 on Build GRCh38
rs75558821 Chr.21: 45422561-45422561 on Build GRCh38
rs764185605 Chr.21: 45422532-45422532 on Build GRCh38
rs902651551 Chr.21: 45422701-45422701 on Build GRCh38
rs749827643 Chr.21: 45422612-45422612 on Build GRCh38
rs746342887 Chr.21: 45422527-45422527 on Build GRCh38
rs111384117 Chr.21: 45422526-45422526 on Build GRCh38
rs573657261 Chr.21: 45422665-45422665 on Build GRCh38
rs1001064821 Chr.21: 45422712-45422712 on Build GRCh38
rs369109321 Chr.21: 45422501-45422501 on Build GRCh38
rs1052992531 Chr.21: 45422647-45422647 on Build GRCh38
rs749190851 Chr.21: 45422405-45422405 on Build GRCh38
rs780431650 Chr.21: 45422698-45422698 on Build GRCh38
rs919731430 Chr.21: 45422367-45422367 on Build GRCh38
rs533124017 Chr.21: 45422374-45422374 on Build GRCh38
rs770916362 Chr.21: 45422422-45422422 on Build GRCh38
rs747224795 Chr.21: 45422520-45422520 on Build GRCh38
rs373383111 Chr.21: 45422387-45422387 on Build GRCh38
rs1015821481 Chr.21: 45422713-45422713 on Build GRCh38
rs541049381 Chr.21: 45422666-45422666 on Build GRCh38
rs980590304 Chr.21: 45422361-45422361 on Build GRCh38
rs779984995 Chr.21: 45422402-45422402 on Build GRCh38
rs902100594 Chr.21: 45422433-45422433 on Build GRCh38
rs1012512312 Chr.21: 45422776-45422776 on Build GRCh38
rs765895502 Chr.21: 45422544-45422544 on Build GRCh38
rs966905140 Chr.21: 45422557-45422557 on Build GRCh38
rs761077950 Chr.21: 45422365-45422365 on Build GRCh38
rs769106430 Chr.21: 45422478-45422478 on Build GRCh38
rs367968714 Chr.21: 45422611-45422611 on Build GRCh38
rs976487819 Chr.21: 45422729-45422729 on Build GRCh38
rs773148513 Chr.21: 45422420-45422420 on Build GRCh38
rs920957924 Chr.21: 45422740-45422740 on Build GRCh38
rs775911134 Chr.21: 45422602-45422602 on Build GRCh38
rs1014057510 Chr.21: 45422652-45422652 on Build GRCh38
rs751362004 Chr.21: 45422417-45422417 on Build GRCh38
rs748010039 Chr.21: 45422509-45422509 on Build GRCh38
rs8130384 Chr.21: 45422352-45422352 on Build GRCh38
rs375208203 Chr.21: 45422373-45422373 on Build GRCh38
rs367782128 Chr.21: 45422329-45422329 on Build GRCh38
rs769877098 Chr.21: 45422600-45422600 on Build GRCh38
rs746007237 Chr.21: 45422596-45422596 on Build GRCh38
rs976464774 Chr.21: 45422617-45422617 on Build GRCh38
rs369890417 Chr.21: 45422771-45422771 on Build GRCh38
rs770992222 Chr.21: 45422523-45422523 on Build GRCh38
rs1026852162 Chr.21: 45422687-45422687 on Build GRCh38
rs774432676 Chr.21: 45422523-45422524 on Build GRCh38
rs775819121 Chr.21: 45422529-45422529 on Build GRCh38
rs370134793 Chr.21: 45422380-45422380 on Build GRCh38
rs751164701 Chr.21: 45422383-45422383 on Build GRCh38
rs1002115632 Chr.21: 45422685-45422685 on Build GRCh38
rs79490253 Chr.21: 45422491-45422491 on Build GRCh38
rs777155233 Chr.21: 45422442-45422442 on Build GRCh38
rs775872220 Chr.21: 45422483-45422483 on Build GRCh38
rs78833614 Chr.21: 45422585-45422585 on Build GRCh38
rs267606165 Chr.21: 45422411-45422411 on Build GRCh38
rs992517240 Chr.21: 45422334-45422334 on Build GRCh38
rs531711320 Chr.21: 45422684-45422684 on Build GRCh38
rs953683397 Chr.21: 45422741-45422741 on Build GRCh38
rs959419217 Chr.21: 45422338-45422338 on Build GRCh38
rs952704914 Chr.21: 45422703-45422703 on Build GRCh38
rs117115507 Chr.21: 45422379-45422379 on Build GRCh38
rs141669114 Chr.21: 45422629-45422629 on Build GRCh38
rs554201032 Chr.21: 45422531-45422531 on Build GRCh38
rs377750483 Chr.21: 45422551-45422551 on Build GRCh38
rs750017535 Chr.21: 45422392-45422392 on Build GRCh38
rs1004053090 Chr.21: 45422707-45422707 on Build GRCh38
rs908295140 Chr.21: 45422592-45422592 on Build GRCh38
rs765840909 Chr.21: 45422499-45422499 on Build GRCh38
rs936860482 Chr.21: 45422642-45422642 on Build GRCh38
rs1051410124 Chr.21: 45422643-45422643 on Build GRCh38
rs536992230 Chr.21: 45422397-45422397 on Build GRCh38
rs548619911 Chr.21: 45422723-45422723 on Build GRCh38
rs369582157 Chr.21: 45422453-45422453 on Build GRCh38
rs755586122 Chr.21: 45422497-45422497 on Build GRCh38
rs199878290 Chr.21: 45422412-45422412 on Build GRCh38
rs79087200 Chr.21: 45422577-45422577 on Build GRCh38
rs551546336 Chr.21: 45422742-45422742 on Build GRCh38
rs892568251 Chr.21: 45422651-45422651 on Build GRCh38
rs765204627 Chr.21: 45422560-45422560 on Build GRCh38
rs952573812 Chr.21: 45422586-45422586 on Build GRCh38
rs776822575 Chr.21: 45422628-45422628 on Build GRCh38
rs760747330 Chr.21: 45422421-45422421 on Build GRCh38
rs532920733 Chr.21: 45422700-45422700 on Build GRCh38
rs753763624 Chr.21: 45422500-45422500 on Build GRCh38
rs373029104 Chr.21: 45422543-45422543 on Build GRCh38
rs780650706 Chr.21: 45422388-45422388 on Build GRCh38
rs754949025 Chr.21: 45422556-45422556 on Build GRCh38
rs982973425 Chr.21: 45422591-45422591 on Build GRCh38
rs990889735 Chr.21: 45422348-45422348 on Build GRCh38
rs8131523 Chr.21: 45422528-45422528 on Build GRCh38
rs1024959795 Chr.21: 45422680-45422680 on Build GRCh38
rs138042795 Chr.21: 45422625-45422625 on Build GRCh38
rs370964335 Chr.21: 45422587-45422587 on Build GRCh38
rs374133620 Chr.21: 45422594-45422594 on Build GRCh38

View Full Product Details