Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 18992808-18993316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGCR5
Entrez Gene ID
26220
Gene Name
DiGeorge syndrome critical region gene 5 (non-protein coding)
Gene Aliases
LINC00037, NCRNA00037
UniGene
Hs.646438
Gene Chromosome Location
Chr.22: 18970498-19031242 on Build GRCh38
Transcripts
NR_002733.2 NR_045121.1 NR_110533.1

SNP Detail

SNP ID Chromosome Location
rs956574176 Chr.22: 18993216-18993216 on Build GRCh38
rs917002009 Chr.22: 18992848-18992848 on Build GRCh38
rs1048609125 Chr.22: 18992918-18992918 on Build GRCh38
rs912497026 Chr.22: 18993252-18993252 on Build GRCh38
rs768334455 Chr.22: 18993294-18993294 on Build GRCh38
rs1000466507 Chr.22: 18993029-18993029 on Build GRCh38
rs920350528 Chr.22: 18993213-18993213 on Build GRCh38
rs1023287460 Chr.22: 18993138-18993138 on Build GRCh38
rs1041062700 Chr.22: 18992958-18992958 on Build GRCh38
rs368732758 Chr.22: 18993028-18993028 on Build GRCh38
rs540203986 Chr.22: 18993011-18993011 on Build GRCh38
rs756414721 Chr.22: 18992859-18992859 on Build GRCh38
rs74550913 Chr.22: 18992846-18992846 on Build GRCh38
rs968552670 Chr.22: 18993004-18993004 on Build GRCh38
rs1210637 Chr.22: 18992930-18992930 on Build GRCh38
rs3804048 Chr.22: 18993278-18993278 on Build GRCh38
rs765290499 Chr.22: 18993059-18993059 on Build GRCh38
rs971814194 Chr.22: 18993204-18993204 on Build GRCh38
rs1032497783 Chr.22: 18993196-18993196 on Build GRCh38
rs192741552 Chr.22: 18993267-18993267 on Build GRCh38
rs1004758250 Chr.22: 18993036-18993036 on Build GRCh38
rs918727142 Chr.22: 18992900-18992900 on Build GRCh38
rs143204516 Chr.22: 18993293-18993293 on Build GRCh38
rs912434459 Chr.22: 18992954-18992954 on Build GRCh38
rs558710659 Chr.22: 18992855-18992855 on Build GRCh38
rs530157889 Chr.22: 18992959-18992959 on Build GRCh38
rs745974622 Chr.22: 18993075-18993075 on Build GRCh38
rs566173541 Chr.22: 18992851-18992851 on Build GRCh38
rs187452357 Chr.22: 18993254-18993254 on Build GRCh38
rs552641449 Chr.22: 18993180-18993180 on Build GRCh38
rs536386975 Chr.22: 18993295-18993295 on Build GRCh38
rs560150723 Chr.22: 18993156-18993156 on Build GRCh38
rs958347995 Chr.22: 18992840-18992840 on Build GRCh38
rs1049060648 Chr.22: 18993262-18993262 on Build GRCh38
rs573158904 Chr.22: 18992836-18992836 on Build GRCh38
rs551623421 Chr.22: 18992982-18992982 on Build GRCh38
rs190219444 Chr.22: 18992922-18992922 on Build GRCh38
rs117092114 Chr.22: 18992907-18992907 on Build GRCh38
rs772174941 Chr.22: 18993099-18993099 on Build GRCh38
rs921361943 Chr.22: 18992975-18992975 on Build GRCh38
rs1048509127 Chr.22: 18992986-18992986 on Build GRCh38
rs1041289711 Chr.22: 18993287-18993287 on Build GRCh38
rs182849954 Chr.22: 18993168-18993168 on Build GRCh38
rs1010242474 Chr.22: 18992961-18992961 on Build GRCh38
rs975313635 Chr.22: 18993258-18993258 on Build GRCh38
rs988341037 Chr.22: 18993226-18993226 on Build GRCh38

View Full Product Details