Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 18993137-18993654 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGCR5
Entrez Gene ID
26220
Gene Name
DiGeorge syndrome critical region gene 5 (non-protein coding)
Gene Aliases
LINC00037, NCRNA00037
UniGene
Hs.646438
Gene Chromosome Location
Chr.22: 18970498-19031242 on Build GRCh38
Transcripts
NR_002733.2 NR_045121.1 NR_110533.1

SNP Detail

SNP ID Chromosome Location
rs956574176 Chr.22: 18993216-18993216 on Build GRCh38
rs574578768 Chr.22: 18993528-18993528 on Build GRCh38
rs1055893050 Chr.22: 18993564-18993564 on Build GRCh38
rs374051106 Chr.22: 18993589-18993589 on Build GRCh38
rs376710853 Chr.22: 18993542-18993542 on Build GRCh38
rs796112585 Chr.22: 18993457-18993457 on Build GRCh38
rs952695373 Chr.22: 18993534-18993534 on Build GRCh38
rs920350528 Chr.22: 18993213-18993213 on Build GRCh38
rs772808482 Chr.22: 18993573-18993573 on Build GRCh38
rs185437910 Chr.22: 18993337-18993337 on Build GRCh38
rs554767274 Chr.22: 18993499-18993499 on Build GRCh38
rs904744880 Chr.22: 18993602-18993602 on Build GRCh38
rs112441342 Chr.22: 18993299-18993299 on Build GRCh38
rs575500696 Chr.22: 18993440-18993440 on Build GRCh38
rs972166603 Chr.22: 18993410-18993410 on Build GRCh38
rs1014143026 Chr.22: 18993375-18993375 on Build GRCh38
rs192741552 Chr.22: 18993267-18993267 on Build GRCh38
rs566517932 Chr.22: 18993333-18993333 on Build GRCh38
rs904900350 Chr.22: 18993354-18993354 on Build GRCh38
rs546189834 Chr.22: 18993563-18993563 on Build GRCh38
rs948997360 Chr.22: 18993317-18993317 on Build GRCh38
rs1032094367 Chr.22: 18993399-18993399 on Build GRCh38
rs567924165 Chr.22: 18993425-18993425 on Build GRCh38
rs1042150922 Chr.22: 18993552-18993552 on Build GRCh38
rs9618441 Chr.22: 18993538-18993538 on Build GRCh38
rs1053832512 Chr.22: 18993630-18993630 on Build GRCh38
rs552641449 Chr.22: 18993180-18993180 on Build GRCh38
rs536386975 Chr.22: 18993295-18993295 on Build GRCh38
rs187452357 Chr.22: 18993254-18993254 on Build GRCh38
rs536552463 Chr.22: 18993625-18993625 on Build GRCh38
rs558529175 Chr.22: 18993378-18993378 on Build GRCh38
rs911355554 Chr.22: 18993447-18993447 on Build GRCh38
rs1014106998 Chr.22: 18993570-18993570 on Build GRCh38
rs975362895 Chr.22: 18993480-18993480 on Build GRCh38
rs1041289711 Chr.22: 18993287-18993287 on Build GRCh38
rs182849954 Chr.22: 18993168-18993168 on Build GRCh38
rs975313635 Chr.22: 18993258-18993258 on Build GRCh38
rs532045646 Chr.22: 18993621-18993621 on Build GRCh38
rs894323189 Chr.22: 18993357-18993357 on Build GRCh38
rs912497026 Chr.22: 18993252-18993252 on Build GRCh38
rs768334455 Chr.22: 18993294-18993294 on Build GRCh38
rs993174096 Chr.22: 18993590-18993590 on Build GRCh38
rs897758292 Chr.22: 18993583-18993583 on Build GRCh38
rs984507831 Chr.22: 18993539-18993539 on Build GRCh38
rs879493851 Chr.22: 18993338-18993338 on Build GRCh38
rs3804048 Chr.22: 18993278-18993278 on Build GRCh38
rs34367141 Chr.22: 18993362-18993363 on Build GRCh38
rs984297210 Chr.22: 18993441-18993441 on Build GRCh38
rs971814194 Chr.22: 18993204-18993204 on Build GRCh38
rs1032497783 Chr.22: 18993196-18993196 on Build GRCh38
rs188959741 Chr.22: 18993562-18993562 on Build GRCh38
rs563011966 Chr.22: 18993571-18993571 on Build GRCh38
rs143204516 Chr.22: 18993293-18993293 on Build GRCh38
rs961675118 Chr.22: 18993407-18993407 on Build GRCh38
rs940130105 Chr.22: 18993549-18993549 on Build GRCh38
rs537617925 Chr.22: 18993498-18993498 on Build GRCh38
rs1049060648 Chr.22: 18993262-18993262 on Build GRCh38
rs866545867 Chr.22: 18993341-18993341 on Build GRCh38
rs895974117 Chr.22: 18993314-18993314 on Build GRCh38
rs894292071 Chr.22: 18993568-18993568 on Build GRCh38
rs1024248610 Chr.22: 18993398-18993398 on Build GRCh38
rs560111295 Chr.22: 18993555-18993555 on Build GRCh38
rs988341037 Chr.22: 18993226-18993226 on Build GRCh38

View Full Product Details