Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 18994256-18994758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGCR5
Entrez Gene ID
26220
Gene Name
DiGeorge syndrome critical region gene 5 (non-protein coding)
Gene Aliases
LINC00037, NCRNA00037
UniGene
Hs.646438
Gene Chromosome Location
Chr.22: 18970498-19031242 on Build GRCh38
Transcripts
NR_002733.2 NR_045121.1 NR_110533.1

SNP Detail

SNP ID Chromosome Location
rs761734451 Chr.22: 18994568-18994568 on Build GRCh38
rs984867288 Chr.22: 18994458-18994458 on Build GRCh38
rs997646433 Chr.22: 18994582-18994582 on Build GRCh38
rs902020674 Chr.22: 18994570-18994570 on Build GRCh38
rs907477432 Chr.22: 18994403-18994403 on Build GRCh38
rs112648883 Chr.22: 18994592-18994592 on Build GRCh38
rs541606315 Chr.22: 18994317-18994317 on Build GRCh38
rs562557854 Chr.22: 18994713-18994713 on Build GRCh38
rs545932958 Chr.22: 18994424-18994424 on Build GRCh38
rs117500510 Chr.22: 18994667-18994667 on Build GRCh38
rs570052659 Chr.22: 18994583-18994583 on Build GRCh38
rs974569592 Chr.22: 18994486-18994486 on Build GRCh38
rs893193451 Chr.22: 18994345-18994345 on Build GRCh38
rs547165713 Chr.22: 18994485-18994485 on Build GRCh38
rs985216758 Chr.22: 18994292-18994292 on Build GRCh38
rs937282954 Chr.22: 18994678-18994678 on Build GRCh38
rs923218135 Chr.22: 18994310-18994310 on Build GRCh38
rs765645133 Chr.22: 18994593-18994593 on Build GRCh38
rs189119811 Chr.22: 18994643-18994643 on Build GRCh38
rs995959795 Chr.22: 18994711-18994711 on Build GRCh38
rs1057716 Chr.22: 18994578-18994578 on Build GRCh38
rs1042097152 Chr.22: 18994316-18994316 on Build GRCh38
rs1001455994 Chr.22: 18994325-18994325 on Build GRCh38
rs527614454 Chr.22: 18994461-18994461 on Build GRCh38
rs985287891 Chr.22: 18994277-18994277 on Build GRCh38
rs1003277122 Chr.22: 18994420-18994420 on Build GRCh38
rs114273319 Chr.22: 18994530-18994530 on Build GRCh38
rs962687157 Chr.22: 18994608-18994608 on Build GRCh38
rs1016282291 Chr.22: 18994685-18994685 on Build GRCh38
rs570673310 Chr.22: 18994496-18994496 on Build GRCh38
rs545023363 Chr.22: 18994668-18994668 on Build GRCh38
rs556578407 Chr.22: 18994554-18994554 on Build GRCh38
rs1023210433 Chr.22: 18994402-18994402 on Build GRCh38
rs556047311 Chr.22: 18994299-18994299 on Build GRCh38
rs1053950436 Chr.22: 18994338-18994338 on Build GRCh38
rs796436848 Chr.22: 18994618-18994618 on Build GRCh38
rs907377569 Chr.22: 18994655-18994655 on Build GRCh38
rs547786555 Chr.22: 18994441-18994441 on Build GRCh38
rs964499626 Chr.22: 18994475-18994475 on Build GRCh38
rs532090106 Chr.22: 18994386-18994386 on Build GRCh38
rs949877319 Chr.22: 18994489-18994489 on Build GRCh38
rs562336395 Chr.22: 18994425-18994425 on Build GRCh38
rs551830100 Chr.22: 18994522-18994522 on Build GRCh38
rs905766164 Chr.22: 18994519-18994519 on Build GRCh38
rs141041638 Chr.22: 18994298-18994298 on Build GRCh38
rs1026279621 Chr.22: 18994432-18994432 on Build GRCh38
rs1010359772 Chr.22: 18994547-18994547 on Build GRCh38
rs531296803 Chr.22: 18994428-18994428 on Build GRCh38
rs541776548 Chr.22: 18994665-18994665 on Build GRCh38
rs764693129 Chr.22: 18994497-18994497 on Build GRCh38
rs1003086590 Chr.22: 18994658-18994658 on Build GRCh38
rs750637521 Chr.22: 18994641-18994641 on Build GRCh38
rs144763197 Chr.22: 18994311-18994311 on Build GRCh38
rs1010725542 Chr.22: 18994370-18994370 on Build GRCh38
rs2913 Chr.22: 18994584-18994584 on Build GRCh38
rs765918910 Chr.22: 18994339-18994339 on Build GRCh38
rs1016623894 Chr.22: 18994596-18994596 on Build GRCh38
rs1020477600 Chr.22: 18994373-18994373 on Build GRCh38
rs966608885 Chr.22: 18994385-18994385 on Build GRCh38
rs923593476 Chr.22: 18994545-18994545 on Build GRCh38
rs1054491631 Chr.22: 18994529-18994529 on Build GRCh38
rs747138291 Chr.22: 18994712-18994712 on Build GRCh38
rs773924229 Chr.22: 18994445-18994445 on Build GRCh38
rs567986821 Chr.22: 18994444-18994444 on Build GRCh38
rs909006191 Chr.22: 18994297-18994297 on Build GRCh38
rs1020779906 Chr.22: 18994549-18994549 on Build GRCh38
rs564705645 Chr.22: 18994318-18994318 on Build GRCh38
rs372859429 Chr.22: 18994710-18994710 on Build GRCh38
rs766336440 Chr.22: 18994688-18994688 on Build GRCh38

View Full Product Details