Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 49777790-49778292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD1
Entrez Gene ID
23774
Gene Name
bromodomain containing 1
Gene Aliases
BRL, BRPF1, BRPF2
UniGene
Hs.127950
Gene Chromosome Location
Chr.22: 49773278-49827927 on Build GRCh38
Transcripts
NM_001304808.1 NM_001304809.1 XM_011530073.1 XM_017028714.1 XM_017028715.1 XM_017028716.1 XM_017028717.1 XM_017028718.1 XM_017028719.1 XM_017028720.1 XM_017028721.1 XM_017028722.1 XM_017028723.1

SNP Detail

SNP ID Chromosome Location
rs551753134 Chr.22: 49778145-49778145 on Build GRCh38
rs757617725 Chr.22: 49777987-49777987 on Build GRCh38
rs932948296 Chr.22: 49777998-49777998 on Build GRCh38
rs981517637 Chr.22: 49777910-49777910 on Build GRCh38
rs751961258 Chr.22: 49777937-49777937 on Build GRCh38
rs906425622 Chr.22: 49778227-49778227 on Build GRCh38
rs894364336 Chr.22: 49778040-49778040 on Build GRCh38
rs940907939 Chr.22: 49778006-49778006 on Build GRCh38
rs779471824 Chr.22: 49777839-49777839 on Build GRCh38
rs144016821 Chr.22: 49778164-49778164 on Build GRCh38
rs750966240 Chr.22: 49778122-49778122 on Build GRCh38
rs376562200 Chr.22: 49777878-49777878 on Build GRCh38
rs754487735 Chr.22: 49777860-49777860 on Build GRCh38
rs914812299 Chr.22: 49777960-49777960 on Build GRCh38
rs574946765 Chr.22: 49778204-49778204 on Build GRCh38
rs541064666 Chr.22: 49778199-49778199 on Build GRCh38
rs376575684 Chr.22: 49777827-49777827 on Build GRCh38
rs769699850 Chr.22: 49777881-49777881 on Build GRCh38
rs993406638 Chr.22: 49778045-49778045 on Build GRCh38
rs751114240 Chr.22: 49777841-49777841 on Build GRCh38
rs957128912 Chr.22: 49778106-49778106 on Build GRCh38
rs897700543 Chr.22: 49777877-49777877 on Build GRCh38
rs373985299 Chr.22: 49778157-49778157 on Build GRCh38
rs7292710 Chr.22: 49778206-49778206 on Build GRCh38
rs892855777 Chr.22: 49778246-49778246 on Build GRCh38
rs150684342 Chr.22: 49778214-49778214 on Build GRCh38
rs959142198 Chr.22: 49777942-49777942 on Build GRCh38
rs941859405 Chr.22: 49778030-49778030 on Build GRCh38
rs897647244 Chr.22: 49778041-49778041 on Build GRCh38
rs73892866 Chr.22: 49777879-49777879 on Build GRCh38
rs1024568262 Chr.22: 49777872-49777872 on Build GRCh38
rs867726171 Chr.22: 49778180-49778180 on Build GRCh38
rs932226876 Chr.22: 49777939-49777939 on Build GRCh38
rs766965168 Chr.22: 49778036-49778036 on Build GRCh38
rs749985579 Chr.22: 49778087-49778087 on Build GRCh38
rs967661905 Chr.22: 49777974-49777974 on Build GRCh38
rs577499113 Chr.22: 49778179-49778179 on Build GRCh38
rs761487399 Chr.22: 49777959-49777959 on Build GRCh38
rs977687157 Chr.22: 49778174-49778174 on Build GRCh38
rs115473526 Chr.22: 49777826-49777826 on Build GRCh38
rs975090512 Chr.22: 49777897-49777897 on Build GRCh38
rs7291371 Chr.22: 49778150-49778150 on Build GRCh38
rs899283554 Chr.22: 49778031-49778031 on Build GRCh38
rs1050184772 Chr.22: 49777999-49777999 on Build GRCh38
rs559702152 Chr.22: 49778200-49778200 on Build GRCh38
rs977656229 Chr.22: 49777979-49777979 on Build GRCh38
rs7291527 Chr.22: 49778258-49778258 on Build GRCh38
rs958779206 Chr.22: 49778152-49778152 on Build GRCh38
rs910204349 Chr.22: 49778195-49778195 on Build GRCh38
rs1014050462 Chr.22: 49777854-49777854 on Build GRCh38
rs1021822827 Chr.22: 49778158-49778158 on Build GRCh38
rs758335605 Chr.22: 49777838-49777838 on Build GRCh38
rs764312424 Chr.22: 49777928-49777928 on Build GRCh38
rs990301733 Chr.22: 49777953-49777953 on Build GRCh38
rs369225919 Chr.22: 49777863-49777863 on Build GRCh38
rs781705824 Chr.22: 49778019-49778019 on Build GRCh38
rs138967917 Chr.22: 49778231-49778231 on Build GRCh38
rs149452046 Chr.22: 49778266-49778266 on Build GRCh38
rs386822590 Chr.22: 49777879-49777880 on Build GRCh38
rs985773448 Chr.22: 49778190-49778190 on Build GRCh38
rs755578803 Chr.22: 49778127-49778127 on Build GRCh38
rs930738001 Chr.22: 49778191-49778191 on Build GRCh38
rs1046313250 Chr.22: 49778226-49778226 on Build GRCh38
rs907232474 Chr.22: 49778132-49778132 on Build GRCh38
rs1037577001 Chr.22: 49778037-49778037 on Build GRCh38
rs761604287 Chr.22: 49777813-49777813 on Build GRCh38
rs962509477 Chr.22: 49778173-49778173 on Build GRCh38
rs903116191 Chr.22: 49778220-49778220 on Build GRCh38
rs73892868 Chr.22: 49777880-49777880 on Build GRCh38
rs1054290174 Chr.22: 49778234-49778234 on Build GRCh38
rs1055725649 Chr.22: 49778039-49778039 on Build GRCh38
rs554699323 Chr.22: 49777903-49777903 on Build GRCh38
rs998730818 Chr.22: 49778047-49778047 on Build GRCh38

View Full Product Details