Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 2109958-2110467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTN4
Entrez Gene ID
152330
Gene Name
contactin 4
Gene Aliases
AXCAM, BIG-2
UniGene
Hs.298705
Gene Chromosome Location
Chr.3: 2098771-3059080 on Build GRCh38
Transcripts
NM_175607.2 XM_006713004.3 XM_011533425.2 XM_011533426.2 XM_011533427.2 XM_017005783.1 XM_017005784.1 XM_017005785.1 NR_046555.1

Gene Symbol
CNTN4-AS2
Entrez Gene ID
100873976
Gene Name
CNTN4 antisense RNA 2
Gene Aliases
-
UniGene
Hs.611138
Gene Chromosome Location
Chr.3: 2110409-2144241 on Build GRCh38
Transcripts
NM_175607.2 XM_006713004.3 XM_011533425.2 XM_011533426.2 XM_011533427.2 XM_017005783.1 XM_017005784.1 XM_017005785.1 NR_046555.1

SNP Detail

SNP ID Chromosome Location
rs900536729 Chr.3: 2110116-2110116 on Build GRCh38
rs751647486 Chr.3: 2110067-2110067 on Build GRCh38
rs755290193 Chr.3: 2110108-2110108 on Build GRCh38
rs979139327 Chr.3: 2110226-2110226 on Build GRCh38
rs1030671161 Chr.3: 2110421-2110421 on Build GRCh38
rs994918418 Chr.3: 2110124-2110124 on Build GRCh38
rs900480302 Chr.3: 2110325-2110325 on Build GRCh38
rs879655702 Chr.3: 2110083-2110083 on Build GRCh38
rs184742638 Chr.3: 2110129-2110129 on Build GRCh38
rs9849959 Chr.3: 2109993-2109993 on Build GRCh38
rs1004678681 Chr.3: 2110344-2110344 on Build GRCh38
rs142459446 Chr.3: 2110089-2110089 on Build GRCh38
rs556033665 Chr.3: 2110246-2110246 on Build GRCh38
rs886541177 Chr.3: 2110340-2110340 on Build GRCh38
rs575575608 Chr.3: 2110295-2110295 on Build GRCh38
rs758295206 Chr.3: 2110405-2110405 on Build GRCh38
rs934806119 Chr.3: 2110232-2110232 on Build GRCh38
rs7427468 Chr.3: 2110002-2110002 on Build GRCh38
rs908321890 Chr.3: 2110347-2110347 on Build GRCh38
rs763497888 Chr.3: 2109987-2109987 on Build GRCh38
rs956465443 Chr.3: 2110430-2110430 on Build GRCh38
rs901059443 Chr.3: 2110045-2110045 on Build GRCh38
rs1010991372 Chr.3: 2110099-2110099 on Build GRCh38
rs991037523 Chr.3: 2110279-2110279 on Build GRCh38
rs2727902 Chr.3: 2110373-2110373 on Build GRCh38
rs767824293 Chr.3: 2110177-2110177 on Build GRCh38
rs1049143569 Chr.3: 2110339-2110339 on Build GRCh38
rs778502063 Chr.3: 2110367-2110367 on Build GRCh38
rs375454716 Chr.3: 2110305-2110305 on Build GRCh38
rs1039044478 Chr.3: 2110109-2110109 on Build GRCh38
rs944970892 Chr.3: 2110291-2110291 on Build GRCh38
rs569331341 Chr.3: 2110357-2110357 on Build GRCh38
rs990857306 Chr.3: 2110198-2110198 on Build GRCh38
rs753630333 Chr.3: 2110228-2110228 on Build GRCh38
rs983820577 Chr.3: 2110330-2110330 on Build GRCh38
rs968143806 Chr.3: 2110206-2110206 on Build GRCh38
rs189423027 Chr.3: 2110195-2110195 on Build GRCh38
rs1027778846 Chr.3: 2110297-2110297 on Build GRCh38
rs141898978 Chr.3: 2110335-2110335 on Build GRCh38
rs1040947204 Chr.3: 2110009-2110009 on Build GRCh38
rs148132107 Chr.3: 2110260-2110260 on Build GRCh38
rs766590545 Chr.3: 2110061-2110061 on Build GRCh38
rs552786241 Chr.3: 2110173-2110173 on Build GRCh38
rs950515681 Chr.3: 2110162-2110162 on Build GRCh38
rs1009276513 Chr.3: 2110086-2110086 on Build GRCh38
rs540467097 Chr.3: 2110407-2110407 on Build GRCh38
rs1034911263 Chr.3: 2110182-2110182 on Build GRCh38
rs916239132 Chr.3: 2110205-2110205 on Build GRCh38
rs368912368 Chr.3: 2110318-2110318 on Build GRCh38
rs1024002836 Chr.3: 2110372-2110372 on Build GRCh38
rs757523132 Chr.3: 2110334-2110334 on Build GRCh38
rs994865982 Chr.3: 2110328-2110328 on Build GRCh38
rs896343012 Chr.3: 2110361-2110361 on Build GRCh38

View Full Product Details