Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 37969305-37969811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTDSPL
Entrez Gene ID
10217
Gene Name
CTD small phosphatase like
Gene Aliases
C3orf8, HYA22, PSR1, RBSP3, SCP3
UniGene
Hs.475963
Gene Chromosome Location
Chr.3: 37862152-37984469 on Build GRCh38
Transcripts
NM_001008392.1 NM_005808.2 XM_017005519.1 XM_017005520.1 NR_029499.1

Gene Symbol
MIR26A1
Entrez Gene ID
407015
Gene Name
microRNA 26a-1
Gene Aliases
MIR26A, MIRN26A1, mir-26a-1
UniGene
Gene Chromosome Location
Chr.3: 37969404-37969480 on Build GRCh38
Transcripts
NM_001008392.1 NM_005808.2 XM_017005519.1 XM_017005520.1 NR_029499.1

SNP Detail

SNP ID Chromosome Location
rs767882361 Chr.3: 37969501-37969501 on Build GRCh38
rs570447983 Chr.3: 37969514-37969514 on Build GRCh38
rs952740244 Chr.3: 37969474-37969474 on Build GRCh38
rs941335599 Chr.3: 37969351-37969351 on Build GRCh38
rs989078733 Chr.3: 37969621-37969621 on Build GRCh38
rs776350969 Chr.3: 37969700-37969700 on Build GRCh38
rs750391225 Chr.3: 37969443-37969443 on Build GRCh38
rs894700984 Chr.3: 37969718-37969718 on Build GRCh38
rs535957883 Chr.3: 37969403-37969403 on Build GRCh38
rs373823013 Chr.3: 37969493-37969493 on Build GRCh38
rs953984627 Chr.3: 37969571-37969571 on Build GRCh38
rs769257509 Chr.3: 37969362-37969362 on Build GRCh38
rs368569783 Chr.3: 37969535-37969535 on Build GRCh38
rs41276521 Chr.3: 37969336-37969336 on Build GRCh38
rs1036991964 Chr.3: 37969705-37969705 on Build GRCh38
rs760255264 Chr.3: 37969633-37969633 on Build GRCh38
rs924765915 Chr.3: 37969667-37969667 on Build GRCh38
rs759373204 Chr.3: 37969750-37969750 on Build GRCh38
rs766768042 Chr.3: 37969396-37969396 on Build GRCh38
rs760914126 Chr.3: 37969511-37969511 on Build GRCh38
rs757341704 Chr.3: 37969513-37969513 on Build GRCh38
rs34538620 Chr.3: 37969530-37969530 on Build GRCh38
rs764409426 Chr.3: 37969512-37969512 on Build GRCh38
rs552372244 Chr.3: 37969758-37969758 on Build GRCh38
rs558857317 Chr.3: 37969500-37969500 on Build GRCh38
rs756293180 Chr.3: 37969431-37969431 on Build GRCh38
rs532767507 Chr.3: 37969730-37969730 on Build GRCh38
rs367606275 Chr.3: 37969475-37969475 on Build GRCh38
rs767679948 Chr.3: 37969411-37969411 on Build GRCh38
rs770727497 Chr.3: 37969664-37969664 on Build GRCh38
rs370719000 Chr.3: 37969412-37969412 on Build GRCh38
rs182070256 Chr.3: 37969473-37969473 on Build GRCh38
rs1015704868 Chr.3: 37969782-37969782 on Build GRCh38
rs369580857 Chr.3: 37969751-37969751 on Build GRCh38
rs748789618 Chr.3: 37969490-37969490 on Build GRCh38
rs1013181729 Chr.3: 37969748-37969748 on Build GRCh38
rs747746725 Chr.3: 37969480-37969480 on Build GRCh38
rs531870847 Chr.3: 37969768-37969768 on Build GRCh38
rs774897357 Chr.3: 37969497-37969497 on Build GRCh38
rs141354299 Chr.3: 37969766-37969766 on Build GRCh38
rs763389442 Chr.3: 37969389-37969389 on Build GRCh38
rs1057403728 Chr.3: 37969444-37969444 on Build GRCh38
rs775869601 Chr.3: 37969510-37969510 on Build GRCh38
rs1031383136 Chr.3: 37969478-37969478 on Build GRCh38
rs965570548 Chr.3: 37969785-37969785 on Build GRCh38
rs190898565 Chr.3: 37969447-37969447 on Build GRCh38
rs114514890 Chr.3: 37969639-37969639 on Build GRCh38
rs370553161 Chr.3: 37969481-37969481 on Build GRCh38
rs773726836 Chr.3: 37969382-37969382 on Build GRCh38
rs537998449 Chr.3: 37969781-37969781 on Build GRCh38
rs1037021583 Chr.3: 37969378-37969378 on Build GRCh38
rs771522629 Chr.3: 37969472-37969472 on Build GRCh38
rs756871872 Chr.3: 37969572-37969572 on Build GRCh38
rs1003533569 Chr.3: 37969347-37969347 on Build GRCh38
rs544552612 Chr.3: 37969559-37969559 on Build GRCh38
rs759930626 Chr.3: 37969404-37969404 on Build GRCh38
rs79726899 Chr.3: 37969634-37969634 on Build GRCh38
rs752661807 Chr.3: 37969786-37969786 on Build GRCh38
rs375068821 Chr.3: 37969622-37969622 on Build GRCh38
rs370187111 Chr.3: 37969491-37969491 on Build GRCh38
rs764213240 Chr.3: 37969441-37969441 on Build GRCh38
rs560643120 Chr.3: 37969674-37969674 on Build GRCh38
rs998007882 Chr.3: 37969789-37969789 on Build GRCh38
rs938885844 Chr.3: 37969691-37969691 on Build GRCh38

View Full Product Details