Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49549510-49550019 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSN-AS2
Entrez Gene ID
100132677
Gene Name
BSN antisense RNA 2 (head to head)
Gene Aliases
-
UniGene
Hs.435651
Gene Chromosome Location
Chr.3: 49549306-49554366 on Build GRCh38
Transcripts
NR_038866.1

SNP Detail

SNP ID Chromosome Location
rs59579780 Chr.3: 49549778-49549778 on Build GRCh38
rs566279791 Chr.3: 49549974-49549974 on Build GRCh38
rs765308239 Chr.3: 49549743-49549743 on Build GRCh38
rs949282641 Chr.3: 49549555-49549555 on Build GRCh38
rs999403813 Chr.3: 49549670-49549670 on Build GRCh38
rs761013240 Chr.3: 49549567-49549567 on Build GRCh38
rs548013550 Chr.3: 49549593-49549593 on Build GRCh38
rs983582988 Chr.3: 49549598-49549598 on Build GRCh38
rs916621175 Chr.3: 49549820-49549820 on Build GRCh38
rs1017233229 Chr.3: 49549717-49549717 on Build GRCh38
rs935486933 Chr.3: 49549673-49549673 on Build GRCh38
rs900170233 Chr.3: 49549742-49549742 on Build GRCh38
rs939426173 Chr.3: 49549614-49549614 on Build GRCh38
rs954481302 Chr.3: 49549761-49549761 on Build GRCh38
rs1027479538 Chr.3: 49549588-49549588 on Build GRCh38
rs1046221670 Chr.3: 49549595-49549595 on Build GRCh38
rs968020055 Chr.3: 49549877-49549877 on Build GRCh38
rs928052036 Chr.3: 49549613-49549613 on Build GRCh38
rs950633574 Chr.3: 49549749-49549749 on Build GRCh38
rs1029181323 Chr.3: 49549752-49549752 on Build GRCh38
rs572871097 Chr.3: 49549807-49549807 on Build GRCh38
rs950603944 Chr.3: 49549597-49549597 on Build GRCh38
rs972998209 Chr.3: 49549802-49549802 on Build GRCh38
rs563168314 Chr.3: 49549631-49549631 on Build GRCh38
rs531650257 Chr.3: 49549677-49549677 on Build GRCh38
rs550018017 Chr.3: 49549745-49549745 on Build GRCh38
rs200704753 Chr.3: 49549641-49549641 on Build GRCh38
rs924203502 Chr.3: 49549916-49549916 on Build GRCh38
rs1008564456 Chr.3: 49549706-49549706 on Build GRCh38
rs973260922 Chr.3: 49549568-49549568 on Build GRCh38
rs193207104 Chr.3: 49549960-49549960 on Build GRCh38
rs931326654 Chr.3: 49549839-49549839 on Build GRCh38
rs1043995653 Chr.3: 49549655-49549655 on Build GRCh38
rs935576164 Chr.3: 49549966-49549966 on Build GRCh38
rs893488253 Chr.3: 49549719-49549719 on Build GRCh38
rs902702145 Chr.3: 49549669-49549669 on Build GRCh38
rs375773602 Chr.3: 49549621-49549621 on Build GRCh38
rs920162978 Chr.3: 49549822-49549822 on Build GRCh38
rs77701561 Chr.3: 49549552-49549552 on Build GRCh38
rs1027967518 Chr.3: 49549748-49549748 on Build GRCh38
rs751359611 Chr.3: 49549975-49549975 on Build GRCh38
rs890221742 Chr.3: 49549678-49549678 on Build GRCh38
rs772428289 Chr.3: 49549538-49549538 on Build GRCh38
rs1019788633 Chr.3: 49549776-49549776 on Build GRCh38
rs916549288 Chr.3: 49549629-49549629 on Build GRCh38
rs367712376 Chr.3: 49549546-49549547 on Build GRCh38
rs916685791 Chr.3: 49549938-49549938 on Build GRCh38
rs764366261 Chr.3: 49549580-49549580 on Build GRCh38
rs761809389 Chr.3: 49549712-49549712 on Build GRCh38
rs990892981 Chr.3: 49549815-49549815 on Build GRCh38
rs751480730 Chr.3: 49549967-49549967 on Build GRCh38
rs571607774 Chr.3: 49549769-49549769 on Build GRCh38
rs979707656 Chr.3: 49549901-49549901 on Build GRCh38

View Full Product Details