Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52062574-52063084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00696
Entrez Gene ID
100128378
Gene Name
long intergenic non-protein coding RNA 696
Gene Aliases
C3orf74
UniGene
Hs.640106
Gene Chromosome Location
Chr.3: 52062094-52065112 on Build GRCh38
Transcripts
NR_027331.1

SNP Detail

SNP ID Chromosome Location
rs571889687 Chr.3: 52062988-52062988 on Build GRCh38
rs183320701 Chr.3: 52062639-52062639 on Build GRCh38
rs556244802 Chr.3: 52062666-52062666 on Build GRCh38
rs993384267 Chr.3: 52062881-52062881 on Build GRCh38
rs770426808 Chr.3: 52062636-52062636 on Build GRCh38
rs748116048 Chr.3: 52063021-52063021 on Build GRCh38
rs919310256 Chr.3: 52062826-52062826 on Build GRCh38
rs1046826648 Chr.3: 52062620-52062620 on Build GRCh38
rs763395401 Chr.3: 52062820-52062820 on Build GRCh38
rs779028027 Chr.3: 52063020-52063020 on Build GRCh38
rs577545285 Chr.3: 52062725-52062725 on Build GRCh38
rs4687608 Chr.3: 52063044-52063044 on Build GRCh38
rs905141996 Chr.3: 52062623-52062623 on Build GRCh38
rs374637748 Chr.3: 52062951-52062951 on Build GRCh38
rs954680303 Chr.3: 52062908-52062908 on Build GRCh38
rs781152208 Chr.3: 52062606-52062606 on Build GRCh38
rs879544569 Chr.3: 52062769-52062769 on Build GRCh38
rs893646879 Chr.3: 52062912-52062912 on Build GRCh38
rs536776585 Chr.3: 52062815-52062815 on Build GRCh38
rs911210966 Chr.3: 52062622-52062622 on Build GRCh38
rs546213680 Chr.3: 52062754-52062754 on Build GRCh38
rs997903003 Chr.3: 52062744-52062744 on Build GRCh38
rs529392614 Chr.3: 52062915-52062915 on Build GRCh38
rs375778513 Chr.3: 52062724-52062724 on Build GRCh38
rs773298494 Chr.3: 52063030-52063030 on Build GRCh38
rs1020625287 Chr.3: 52062671-52062671 on Build GRCh38
rs972150673 Chr.3: 52062825-52062825 on Build GRCh38
rs909399173 Chr.3: 52062786-52062786 on Build GRCh38
rs1053600401 Chr.3: 52062853-52062853 on Build GRCh38
rs1053117797 Chr.3: 52062742-52062742 on Build GRCh38
rs926597823 Chr.3: 52062847-52062847 on Build GRCh38
rs900956356 Chr.3: 52062948-52062948 on Build GRCh38
rs191454932 Chr.3: 52062625-52062625 on Build GRCh38
rs771263222 Chr.3: 52063045-52063045 on Build GRCh38
rs550638812 Chr.3: 52062987-52062987 on Build GRCh38
rs963510518 Chr.3: 52062801-52062801 on Build GRCh38
rs774015444 Chr.3: 52062842-52062842 on Build GRCh38
rs919264933 Chr.3: 52062605-52062605 on Build GRCh38
rs370083405 Chr.3: 52062785-52062785 on Build GRCh38
rs759133853 Chr.3: 52062950-52062950 on Build GRCh38
rs777019208 Chr.3: 52063050-52063050 on Build GRCh38
rs1016491845 Chr.3: 52062989-52062989 on Build GRCh38
rs1044780672 Chr.3: 52062809-52062809 on Build GRCh38
rs80052444 Chr.3: 52063024-52063024 on Build GRCh38
rs937948670 Chr.3: 52062852-52062852 on Build GRCh38
rs1030572163 Chr.3: 52062889-52062889 on Build GRCh38
rs963986947 Chr.3: 52062978-52062978 on Build GRCh38
rs949660672 Chr.3: 52062755-52062755 on Build GRCh38
rs561935873 Chr.3: 52062897-52062897 on Build GRCh38
rs75145905 Chr.3: 52062807-52062807 on Build GRCh38
rs1042610392 Chr.3: 52062919-52062919 on Build GRCh38
rs773921149 Chr.3: 52062667-52062667 on Build GRCh38
rs547235616 Chr.3: 52062828-52062828 on Build GRCh38
rs1007872883 Chr.3: 52062924-52062924 on Build GRCh38
rs760247103 Chr.3: 52063046-52063046 on Build GRCh38
rs935476475 Chr.3: 52062740-52062740 on Build GRCh38
rs140976500 Chr.3: 52062760-52062760 on Build GRCh38

View Full Product Details