Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52062741-52063275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00696
Entrez Gene ID
100128378
Gene Name
long intergenic non-protein coding RNA 696
Gene Aliases
C3orf74
UniGene
Hs.640106
Gene Chromosome Location
Chr.3: 52062094-52065112 on Build GRCh38
Transcripts
NR_027331.1

SNP Detail

SNP ID Chromosome Location
rs1042254289 Chr.3: 52063248-52063248 on Build GRCh38
rs765134573 Chr.3: 52063199-52063199 on Build GRCh38
rs370136831 Chr.3: 52063129-52063129 on Build GRCh38
rs571889687 Chr.3: 52062988-52062988 on Build GRCh38
rs778645165 Chr.3: 52063131-52063131 on Build GRCh38
rs993384267 Chr.3: 52062881-52062881 on Build GRCh38
rs777601664 Chr.3: 52063232-52063232 on Build GRCh38
rs751588371 Chr.3: 52063234-52063234 on Build GRCh38
rs760456685 Chr.3: 52063098-52063098 on Build GRCh38
rs373351265 Chr.3: 52063179-52063179 on Build GRCh38
rs781412678 Chr.3: 52063157-52063157 on Build GRCh38
rs759924005 Chr.3: 52063071-52063071 on Build GRCh38
rs748116048 Chr.3: 52063021-52063021 on Build GRCh38
rs766249983 Chr.3: 52063115-52063115 on Build GRCh38
rs919310256 Chr.3: 52062826-52062826 on Build GRCh38
rs759397299 Chr.3: 52063198-52063198 on Build GRCh38
rs763395401 Chr.3: 52062820-52062820 on Build GRCh38
rs779028027 Chr.3: 52063020-52063020 on Build GRCh38
rs772822985 Chr.3: 52063174-52063174 on Build GRCh38
rs752743363 Chr.3: 52063135-52063135 on Build GRCh38
rs4687608 Chr.3: 52063044-52063044 on Build GRCh38
rs989357851 Chr.3: 52063089-52063089 on Build GRCh38
rs374637748 Chr.3: 52062951-52062951 on Build GRCh38
rs915195808 Chr.3: 52063137-52063137 on Build GRCh38
rs770164738 Chr.3: 52063067-52063075 on Build GRCh38
rs954680303 Chr.3: 52062908-52062908 on Build GRCh38
rs758390910 Chr.3: 52063139-52063139 on Build GRCh38
rs879544569 Chr.3: 52062769-52062769 on Build GRCh38
rs201995264 Chr.3: 52063190-52063190 on Build GRCh38
rs893646879 Chr.3: 52062912-52062912 on Build GRCh38
rs775857107 Chr.3: 52063164-52063164 on Build GRCh38
rs536776585 Chr.3: 52062815-52062815 on Build GRCh38
rs747167351 Chr.3: 52063144-52063144 on Build GRCh38
rs534040139 Chr.3: 52063118-52063118 on Build GRCh38
rs529392614 Chr.3: 52062915-52062915 on Build GRCh38
rs750031049 Chr.3: 52063087-52063087 on Build GRCh38
rs369524903 Chr.3: 52063083-52063083 on Build GRCh38
rs773298494 Chr.3: 52063030-52063030 on Build GRCh38
rs115973139 Chr.3: 52063085-52063085 on Build GRCh38
rs972150673 Chr.3: 52062825-52062825 on Build GRCh38
rs909399173 Chr.3: 52062786-52062786 on Build GRCh38
rs1053600401 Chr.3: 52062853-52062853 on Build GRCh38
rs773357576 Chr.3: 52063249-52063264 on Build GRCh38
rs926597823 Chr.3: 52062847-52062847 on Build GRCh38
rs900956356 Chr.3: 52062948-52062948 on Build GRCh38
rs770149295 Chr.3: 52063158-52063158 on Build GRCh38
rs771263222 Chr.3: 52063045-52063045 on Build GRCh38
rs537970114 Chr.3: 52063242-52063242 on Build GRCh38
rs550638812 Chr.3: 52062987-52062987 on Build GRCh38
rs777956857 Chr.3: 52063140-52063140 on Build GRCh38
rs567468169 Chr.3: 52063240-52063240 on Build GRCh38
rs963510518 Chr.3: 52062801-52062801 on Build GRCh38
rs774015444 Chr.3: 52062842-52062842 on Build GRCh38
rs370083405 Chr.3: 52062785-52062785 on Build GRCh38
rs759133853 Chr.3: 52062950-52062950 on Build GRCh38
rs753543055 Chr.3: 52063185-52063185 on Build GRCh38
rs777019208 Chr.3: 52063050-52063050 on Build GRCh38
rs1016491845 Chr.3: 52062989-52062989 on Build GRCh38
rs1044780672 Chr.3: 52062809-52062809 on Build GRCh38
rs771465717 Chr.3: 52063173-52063173 on Build GRCh38
rs80052444 Chr.3: 52063024-52063024 on Build GRCh38
rs937948670 Chr.3: 52062852-52062852 on Build GRCh38
rs963986947 Chr.3: 52062978-52062978 on Build GRCh38
rs1030572163 Chr.3: 52062889-52062889 on Build GRCh38
rs1037369673 Chr.3: 52063177-52063177 on Build GRCh38
rs767123303 Chr.3: 52063086-52063086 on Build GRCh38
rs561935873 Chr.3: 52062897-52062897 on Build GRCh38
rs75145905 Chr.3: 52062807-52062807 on Build GRCh38
rs771099713 Chr.3: 52063153-52063153 on Build GRCh38
rs549242086 Chr.3: 52063134-52063134 on Build GRCh38
rs372951139 Chr.3: 52063084-52063084 on Build GRCh38
rs1042610392 Chr.3: 52062919-52062919 on Build GRCh38
rs547235616 Chr.3: 52062828-52062828 on Build GRCh38
rs1007872883 Chr.3: 52062924-52062924 on Build GRCh38
rs763573887 Chr.3: 52063165-52063165 on Build GRCh38
rs752550733 Chr.3: 52063211-52063211 on Build GRCh38
rs758401513 Chr.3: 52063216-52063216 on Build GRCh38
rs201261921 Chr.3: 52063180-52063180 on Build GRCh38
rs760247103 Chr.3: 52063046-52063046 on Build GRCh38

View Full Product Details