Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 55658625-55659157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERC2
Entrez Gene ID
26059
Gene Name
ELKS/RAB6-interacting/CAST family member 2
Gene Aliases
CAST, CAST1, ELKSL, SPBC110, Spc110
UniGene
Hs.476389
Gene Chromosome Location
Chr.3: 55508311-56468473 on Build GRCh38
Transcripts
NM_015576.2 NR_132749.1 XM_017006138.1 XM_017006139.1 XM_017006140.1 XM_017006141.1 XM_017006142.1 XM_017006143.1 XM_017006144.1 XM_017006145.1 XM_017006146.1 XM_017006147.1 XM_017006148.1 XM_017006149.1 XM_017006150.1 XM_017006151.1 XM_017006152.1 XM_017006153.1 XM_017006154.1 XM_017006155.1 XM_017006156.1 XM_017006157.1 XM_017006158.1 XM_017006159.1 NR_024615.1

Gene Symbol
ERC2-IT1
Entrez Gene ID
711
Gene Name
ERC2 intronic transcript 1
Gene Aliases
C1orf1, C3orf51, Po42
UniGene
Hs.667161
Gene Chromosome Location
Chr.3: 55657206-55659469 on Build GRCh38
Transcripts
NM_015576.2 NR_132749.1 XM_017006138.1 XM_017006139.1 XM_017006140.1 XM_017006141.1 XM_017006142.1 XM_017006143.1 XM_017006144.1 XM_017006145.1 XM_017006146.1 XM_017006147.1 XM_017006148.1 XM_017006149.1 XM_017006150.1 XM_017006151.1 XM_017006152.1 XM_017006153.1 XM_017006154.1 XM_017006155.1 XM_017006156.1 XM_017006157.1 XM_017006158.1 XM_017006159.1 NR_024615.1

SNP Detail

SNP ID Chromosome Location
rs572293483 Chr.3: 55658998-55658998 on Build GRCh38
rs1004900818 Chr.3: 55659114-55659114 on Build GRCh38
rs540607482 Chr.3: 55659122-55659122 on Build GRCh38
rs749057332 Chr.3: 55658811-55658811 on Build GRCh38
rs933881399 Chr.3: 55659002-55659002 on Build GRCh38
rs140552096 Chr.3: 55658800-55658800 on Build GRCh38
rs1012885531 Chr.3: 55659062-55659062 on Build GRCh38
rs745876544 Chr.3: 55659042-55659042 on Build GRCh38
rs975327635 Chr.3: 55658960-55658960 on Build GRCh38
rs915108381 Chr.3: 55658661-55658661 on Build GRCh38
rs756334334 Chr.3: 55658793-55658793 on Build GRCh38
rs139967100 Chr.3: 55659012-55659019 on Build GRCh38
rs576261663 Chr.3: 55658845-55658845 on Build GRCh38
rs778694231 Chr.3: 55658971-55658971 on Build GRCh38
rs1047525821 Chr.3: 55659019-55659019 on Build GRCh38
rs1047172772 Chr.3: 55658698-55658698 on Build GRCh38
rs2231387 Chr.3: 55658832-55658832 on Build GRCh38
rs1030641182 Chr.3: 55658870-55658870 on Build GRCh38
rs981783855 Chr.3: 55658869-55658869 on Build GRCh38
rs989212536 Chr.3: 55658907-55658907 on Build GRCh38
rs1038450865 Chr.3: 55659100-55659100 on Build GRCh38
rs1016201250 Chr.3: 55658846-55658846 on Build GRCh38
rs1042589160 Chr.3: 55658674-55658674 on Build GRCh38
rs750017447 Chr.3: 55658856-55658856 on Build GRCh38
rs1002943913 Chr.3: 55658701-55658701 on Build GRCh38
rs1017523782 Chr.3: 55658724-55658724 on Build GRCh38
rs541487776 Chr.3: 55658706-55658706 on Build GRCh38
rs2231386 Chr.3: 55659090-55659090 on Build GRCh38
rs956661682 Chr.3: 55658881-55658881 on Build GRCh38
rs147615103 Chr.3: 55658765-55658765 on Build GRCh38
rs939187850 Chr.3: 55659016-55659016 on Build GRCh38
rs567346872 Chr.3: 55658752-55658752 on Build GRCh38
rs115748214 Chr.3: 55658934-55658934 on Build GRCh38
rs534113078 Chr.3: 55658948-55658948 on Build GRCh38
rs761049254 Chr.3: 55659011-55659012 on Build GRCh38
rs371257385 Chr.3: 55659013-55659020 on Build GRCh38
rs779759079 Chr.3: 55659097-55659097 on Build GRCh38
rs945240862 Chr.3: 55658663-55658663 on Build GRCh38
rs758126556 Chr.3: 55659057-55659057 on Build GRCh38
rs933977614 Chr.3: 55658683-55658683 on Build GRCh38
rs1027247196 Chr.3: 55658747-55658747 on Build GRCh38
rs368714751 Chr.3: 55658895-55658895 on Build GRCh38
rs894669881 Chr.3: 55659119-55659119 on Build GRCh38
rs868336505 Chr.3: 55658995-55658995 on Build GRCh38
rs894531432 Chr.3: 55659051-55659051 on Build GRCh38
rs888437066 Chr.3: 55659106-55659106 on Build GRCh38
rs752960993 Chr.3: 55658726-55658726 on Build GRCh38
rs1005611409 Chr.3: 55658716-55658716 on Build GRCh38

View Full Product Details