Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 139123870-139124378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPESC1
Entrez Gene ID
60467
Gene Name
blepharophimosis, epicanthus inversus and ptosis, candidate 1 (non-protein coding)
Gene Aliases
NCRNA00187
UniGene
Hs.512673
Gene Chromosome Location
Chr.3: 139104185-139125167 on Build GRCh38
Transcripts
NR_026783.3

SNP Detail

SNP ID Chromosome Location
rs902883530 Chr.3: 139124127-139124127 on Build GRCh38
rs1001935782 Chr.3: 139123960-139123960 on Build GRCh38
rs1040286176 Chr.3: 139124044-139124044 on Build GRCh38
rs561685088 Chr.3: 139124126-139124126 on Build GRCh38
rs375922066 Chr.3: 139124109-139124109 on Build GRCh38
rs1054025528 Chr.3: 139124146-139124146 on Build GRCh38
rs1012801075 Chr.3: 139124166-139124166 on Build GRCh38
rs905013590 Chr.3: 139124203-139124203 on Build GRCh38
rs968301630 Chr.3: 139123987-139123987 on Build GRCh38
rs984127186 Chr.3: 139124288-139124288 on Build GRCh38
rs896204486 Chr.3: 139124213-139124213 on Build GRCh38
rs75970478 Chr.3: 139124255-139124255 on Build GRCh38
rs1034712392 Chr.3: 139123964-139123964 on Build GRCh38
rs923311449 Chr.3: 139124071-139124071 on Build GRCh38
rs910703932 Chr.3: 139124042-139124042 on Build GRCh38
rs909852368 Chr.3: 139124309-139124309 on Build GRCh38
rs528882343 Chr.3: 139123942-139123942 on Build GRCh38
rs999830511 Chr.3: 139124144-139124144 on Build GRCh38
rs904961022 Chr.3: 139123957-139123957 on Build GRCh38
rs933955715 Chr.3: 139123906-139123906 on Build GRCh38
rs369852083 Chr.3: 139124198-139124198 on Build GRCh38
rs111944814 Chr.3: 139124254-139124254 on Build GRCh38
rs745920607 Chr.3: 139124056-139124056 on Build GRCh38
rs566097997 Chr.3: 139124282-139124282 on Build GRCh38
rs114662327 Chr.3: 139124237-139124237 on Build GRCh38
rs531662943 Chr.3: 139124050-139124050 on Build GRCh38
rs1021634538 Chr.3: 139123982-139123982 on Build GRCh38
rs933772749 Chr.3: 139124058-139124058 on Build GRCh38
rs187987309 Chr.3: 139123968-139123968 on Build GRCh38
rs552738761 Chr.3: 139124259-139124259 on Build GRCh38
rs1000090148 Chr.3: 139123939-139123939 on Build GRCh38
rs929298099 Chr.3: 139124122-139124122 on Build GRCh38
rs1047417561 Chr.3: 139123909-139123909 on Build GRCh38
rs2688682 Chr.3: 139123967-139123967 on Build GRCh38
rs116719496 Chr.3: 139124212-139124212 on Build GRCh38
rs901840291 Chr.3: 139123903-139123903 on Build GRCh38
rs183259619 Chr.3: 139123938-139123938 on Build GRCh38
rs778846848 Chr.3: 139124208-139124208 on Build GRCh38
rs544857472 Chr.3: 139124329-139124329 on Build GRCh38
rs769828168 Chr.3: 139124145-139124145 on Build GRCh38
rs1016738345 Chr.3: 139123948-139123948 on Build GRCh38
rs1040379051 Chr.3: 139123897-139123897 on Build GRCh38
rs910156457 Chr.3: 139124000-139124000 on Build GRCh38
rs980029721 Chr.3: 139123988-139123988 on Build GRCh38
rs1038232936 Chr.3: 139124167-139124167 on Build GRCh38
rs942928016 Chr.3: 139124002-139124002 on Build GRCh38
rs1001993726 Chr.3: 139124207-139124207 on Build GRCh38
rs941263349 Chr.3: 139124160-139124160 on Build GRCh38
rs151025533 Chr.3: 139124104-139124104 on Build GRCh38
rs1047688372 Chr.3: 139124129-139124129 on Build GRCh38
rs376190765 Chr.3: 139123943-139123943 on Build GRCh38

View Full Product Details