Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186784695-186785208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4A2
Entrez Gene ID
1974
Gene Name
eukaryotic translation initiation factor 4A2
Gene Aliases
BM-010, DDX2B, EIF4A, EIF4F, eIF-4A-II, eIF4A-II
UniGene
Hs.518475
Gene Chromosome Location
Chr.3: 186783572-186789897 on Build GRCh38
Transcripts
NM_001967.3 NR_002587.1

Gene Symbol
SNORD2
Entrez Gene ID
619567
Gene Name
small nucleolar RNA, C/D box 2
Gene Aliases
R39B, SNR39B
UniGene
Gene Chromosome Location
Chr.3: 186784796-186784865 on Build GRCh38
Transcripts
NM_001967.3 NR_002587.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4849882 c.292G>T p.E98* Chr.3: 186785045-186785045 on Build GRCh38
6255481 c.217G>T p.V73L Chr.3: 186784970-186784970 on Build GRCh38
6956802 c.313C>A p.L105I Chr.3: 186785066-186785066 on Build GRCh38
5398820 c.270C>T p.S90S Chr.3: 186785023-186785023 on Build GRCh38
7094073 c.253G>T p.A85S Chr.3: 186785006-186785006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773707826 Chr.3: 186785120-186785120 on Build GRCh38
rs750580382 Chr.3: 186784814-186784814 on Build GRCh38
rs375543403 Chr.3: 186784897-186784897 on Build GRCh38
rs764969586 Chr.3: 186784773-186784773 on Build GRCh38
rs775303809 Chr.3: 186784863-186784866 on Build GRCh38
rs762844796 Chr.3: 186784777-186784777 on Build GRCh38
rs777728886 Chr.3: 186784745-186784746 on Build GRCh38
rs367643607 Chr.3: 186784731-186784731 on Build GRCh38
rs753306564 Chr.3: 186785062-186785062 on Build GRCh38
rs753913348 Chr.3: 186784795-186784795 on Build GRCh38
rs997372981 Chr.3: 186785071-186785071 on Build GRCh38
rs777494202 Chr.3: 186785109-186785109 on Build GRCh38
rs766158427 Chr.3: 186784783-186784783 on Build GRCh38
rs771820404 Chr.3: 186784846-186784847 on Build GRCh38
rs768097577 Chr.3: 186784757-186784757 on Build GRCh38
rs778635126 Chr.3: 186785121-186785121 on Build GRCh38
rs775555447 Chr.3: 186785009-186785009 on Build GRCh38
rs201418586 Chr.3: 186784735-186784735 on Build GRCh38
rs11538617 Chr.3: 186785083-186785083 on Build GRCh38
rs547575338 Chr.3: 186784852-186784852 on Build GRCh38
rs192246586 Chr.3: 186784904-186784904 on Build GRCh38
rs916056628 Chr.3: 186784771-186784771 on Build GRCh38
rs1009087477 Chr.3: 186784853-186784853 on Build GRCh38
rs773335544 Chr.3: 186784995-186784995 on Build GRCh38
rs377425549 Chr.3: 186784813-186784813 on Build GRCh38
rs775869166 Chr.3: 186784741-186784741 on Build GRCh38
rs369813206 Chr.3: 186785108-186785108 on Build GRCh38
rs552816121 Chr.3: 186784740-186784740 on Build GRCh38
rs748819825 Chr.3: 186785118-186785118 on Build GRCh38
rs762227302 Chr.3: 186784949-186784951 on Build GRCh38
rs773029428 Chr.3: 186784845-186784845 on Build GRCh38
rs377269772 Chr.3: 186784926-186784926 on Build GRCh38
rs775441076 Chr.3: 186784952-186784952 on Build GRCh38
rs898121540 Chr.3: 186784900-186784900 on Build GRCh38
rs565320884 Chr.3: 186784918-186784918 on Build GRCh38
rs781765741 Chr.3: 186784905-186784905 on Build GRCh38
rs771792470 Chr.3: 186784929-186784929 on Build GRCh38
rs770682987 Chr.3: 186784860-186784860 on Build GRCh38
rs145694989 Chr.3: 186785027-186785027 on Build GRCh38
rs756667726 Chr.3: 186785119-186785119 on Build GRCh38
rs774164716 Chr.3: 186784863-186784863 on Build GRCh38
rs1020882553 Chr.3: 186785105-186785105 on Build GRCh38
rs369392748 Chr.3: 186784801-186784801 on Build GRCh38
rs780906671 Chr.3: 186784832-186784832 on Build GRCh38
rs751936510 Chr.3: 186784903-186784903 on Build GRCh38
rs749561100 Chr.3: 186784729-186784729 on Build GRCh38
rs759495727 Chr.3: 186784791-186784791 on Build GRCh38
rs750690230 Chr.3: 186784882-186784882 on Build GRCh38
rs36060455 Chr.3: 186784894-186784895 on Build GRCh38
rs11538611 Chr.3: 186784980-186784980 on Build GRCh38
rs745474441 Chr.3: 186785144-186785144 on Build GRCh38
rs748935479 Chr.3: 186784915-186784915 on Build GRCh38
rs757440601 Chr.3: 186784730-186784730 on Build GRCh38
rs747506625 Chr.3: 186784826-186784826 on Build GRCh38
rs541382107 Chr.3: 186784797-186784797 on Build GRCh38
rs1040598096 Chr.3: 186785183-186785183 on Build GRCh38
rs893849663 Chr.3: 186784817-186784817 on Build GRCh38
rs916465363 Chr.3: 186784725-186784725 on Build GRCh38
rs377637826 Chr.3: 186784763-186784763 on Build GRCh38
rs952404904 Chr.3: 186784889-186784889 on Build GRCh38
rs765521699 Chr.3: 186784952-186784952 on Build GRCh38
rs747497696 Chr.3: 186784749-186784749 on Build GRCh38
rs3816112 Chr.3: 186784876-186784876 on Build GRCh38
rs926024001 Chr.3: 186784754-186784755 on Build GRCh38
rs767433110 Chr.3: 186784871-186784871 on Build GRCh38
rs780441673 Chr.3: 186784825-186784825 on Build GRCh38
rs11538614 Chr.3: 186785007-186785007 on Build GRCh38
rs773986749 Chr.3: 186784812-186784814 on Build GRCh38
rs745439958 Chr.3: 186784838-186784840 on Build GRCh38
rs77070252 Chr.3: 186784738-186784738 on Build GRCh38
rs749190693 Chr.3: 186784763-186784764 on Build GRCh38
rs556676353 Chr.3: 186784815-186784815 on Build GRCh38
rs770418129 Chr.3: 186784794-186784795 on Build GRCh38
rs771880241 Chr.3: 186785152-186785152 on Build GRCh38
rs769198532 Chr.3: 186784830-186784830 on Build GRCh38
rs1011303416 Chr.3: 186784839-186784839 on Build GRCh38
rs17850936 Chr.3: 186785017-186785017 on Build GRCh38
rs946749236 Chr.3: 186785065-186785065 on Build GRCh38
rs75553325 Chr.3: 186784908-186784908 on Build GRCh38
rs199724027 Chr.3: 186784954-186784954 on Build GRCh38
rs755425078 Chr.3: 186785044-186785044 on Build GRCh38
rs747674776 Chr.3: 186784840-186784840 on Build GRCh38
rs767943275 Chr.3: 186785053-186785053 on Build GRCh38
rs761621888 Chr.3: 186784765-186784765 on Build GRCh38
rs369787126 Chr.3: 186784726-186784726 on Build GRCh38
rs201339295 Chr.3: 186785028-186785028 on Build GRCh38
rs889379838 Chr.3: 186785098-186785098 on Build GRCh38
rs11538616 Chr.3: 186785032-186785032 on Build GRCh38
rs751476534 Chr.3: 186784785-186784785 on Build GRCh38
rs765264400 Chr.3: 186784880-186784880 on Build GRCh38
rs764120426 Chr.3: 186784792-186784792 on Build GRCh38
rs1050836155 Chr.3: 186785078-186785078 on Build GRCh38
rs993912320 Chr.3: 186784873-186784873 on Build GRCh38
rs751663411 Chr.3: 186785035-186785035 on Build GRCh38
rs772941758 Chr.3: 186784776-186784776 on Build GRCh38
rs1052512682 Chr.3: 186784807-186784807 on Build GRCh38
rs141851340 Chr.3: 186784920-186784920 on Build GRCh38
rs1050726749 Chr.3: 186784850-186784850 on Build GRCh38
rs777400639 Chr.3: 186784910-186784910 on Build GRCh38
rs538734682 Chr.3: 186784877-186784877 on Build GRCh38
rs758612890 Chr.3: 186784886-186784886 on Build GRCh38

View Full Product Details