Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 52712642-52713144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DANCR
Entrez Gene ID
57291
Gene Name
differentiation antagonizing non-protein coding RNA
Gene Aliases
AGU2, ANCR, KIAA0114, SNHG13, lncRNA-ANCR
UniGene
Hs.744077
Gene Chromosome Location
Chr.4: 52712454-52714138 on Build GRCh38
Transcripts
NR_024031.1 NR_039651.1

Gene Symbol
MIR4449
Entrez Gene ID
100616436
Gene Name
microRNA 4449
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.4: 52712682-52712747 on Build GRCh38
Transcripts
NR_024031.1 NR_039651.1

SNP Detail

SNP ID Chromosome Location
rs890675903 Chr.4: 52713106-52713106 on Build GRCh38
rs950559163 Chr.4: 52712938-52712938 on Build GRCh38
rs555690377 Chr.4: 52712720-52712720 on Build GRCh38
rs1009070948 Chr.4: 52713113-52713113 on Build GRCh38
rs1011684951 Chr.4: 52712810-52712810 on Build GRCh38
rs916484193 Chr.4: 52712997-52712997 on Build GRCh38
rs765036366 Chr.4: 52712787-52712787 on Build GRCh38
rs1053394744 Chr.4: 52713088-52713088 on Build GRCh38
rs371689570 Chr.4: 52712802-52712802 on Build GRCh38
rs919099695 Chr.4: 52712687-52712687 on Build GRCh38
rs960375789 Chr.4: 52712908-52712908 on Build GRCh38
rs564256291 Chr.4: 52713108-52713108 on Build GRCh38
rs962435451 Chr.4: 52712955-52712955 on Build GRCh38
rs914883531 Chr.4: 52713091-52713091 on Build GRCh38
rs894066595 Chr.4: 52712838-52712838 on Build GRCh38
rs527783562 Chr.4: 52713032-52713032 on Build GRCh38
rs193157776 Chr.4: 52712985-52712985 on Build GRCh38
rs561775096 Chr.4: 52713011-52713011 on Build GRCh38
rs1001753509 Chr.4: 52712849-52712849 on Build GRCh38
rs749729168 Chr.4: 52712927-52712928 on Build GRCh38
rs909116369 Chr.4: 52712727-52712727 on Build GRCh38
rs1039444530 Chr.4: 52712755-52712755 on Build GRCh38
rs1029133205 Chr.4: 52712880-52712881 on Build GRCh38
rs375718168 Chr.4: 52712688-52712688 on Build GRCh38
rs1053186052 Chr.4: 52712792-52712792 on Build GRCh38
rs868273944 Chr.4: 52713047-52713047 on Build GRCh38
rs947593790 Chr.4: 52713096-52713096 on Build GRCh38
rs1033772654 Chr.4: 52712861-52712861 on Build GRCh38
rs970279113 Chr.4: 52712821-52712821 on Build GRCh38
rs943284903 Chr.4: 52712984-52712984 on Build GRCh38
rs990712837 Chr.4: 52712996-52712996 on Build GRCh38
rs992338308 Chr.4: 52712913-52712913 on Build GRCh38
rs533694195 Chr.4: 52712767-52712767 on Build GRCh38
rs931177900 Chr.4: 52712691-52712691 on Build GRCh38
rs535996615 Chr.4: 52712705-52712705 on Build GRCh38
rs576950711 Chr.4: 52712857-52712857 on Build GRCh38
rs1053862489 Chr.4: 52712830-52712830 on Build GRCh38
rs891987684 Chr.4: 52712798-52712798 on Build GRCh38
rs556471264 Chr.4: 52712942-52712942 on Build GRCh38
rs973430412 Chr.4: 52712959-52712959 on Build GRCh38
rs1053639672 Chr.4: 52713048-52713048 on Build GRCh38
rs545831935 Chr.4: 52712936-52712936 on Build GRCh38
rs949176991 Chr.4: 52712730-52712730 on Build GRCh38
rs909168749 Chr.4: 52712945-52712945 on Build GRCh38
rs541968313 Chr.4: 52713000-52713000 on Build GRCh38
rs1043207014 Chr.4: 52713097-52713097 on Build GRCh38
rs943385267 Chr.4: 52712753-52712753 on Build GRCh38
rs533127804 Chr.4: 52713118-52713118 on Build GRCh38
rs148992476 Chr.4: 52713084-52713084 on Build GRCh38
rs935048001 Chr.4: 52712795-52712795 on Build GRCh38
rs991890253 Chr.4: 52712665-52712665 on Build GRCh38
rs1021864801 Chr.4: 52712814-52712814 on Build GRCh38
rs576241524 Chr.4: 52713028-52713028 on Build GRCh38
rs949277880 Chr.4: 52712998-52712998 on Build GRCh38
rs568136712 Chr.4: 52712759-52712759 on Build GRCh38

View Full Product Details