Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99204227-99204775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADH6
Entrez Gene ID
130
Gene Name
alcohol dehydrogenase 6 (class V)
Gene Aliases
ADH-5
UniGene
Hs.586161
Gene Chromosome Location
Chr.4: 99202638-99219246 on Build GRCh38
Transcripts
NM_000672.3 NM_001102470.1 NR_132990.1 NR_037884.1

Gene Symbol
LOC100507053
Entrez Gene ID
100507053
Gene Name
uncharacterized LOC100507053
Gene Aliases
-
UniGene
Hs.635084
Gene Chromosome Location
Chr.4: 99088857-99301356 on Build GRCh38
Transcripts
NM_000672.3 NM_001102470.1 NR_132990.1 NR_037884.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3598522 c.1107C>T p.I369I Chr.4: 99204240-99204240 on Build GRCh38
3598521 c.1117C>T p.L373L Chr.4: 99204230-99204230 on Build GRCh38
1049381 c.1109G>A p.R370H Chr.4: 99204238-99204238 on Build GRCh38
4120923 c.1108C>T p.R370C Chr.4: 99204239-99204239 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs952867400 Chr.4: 99204496-99204496 on Build GRCh38
rs1050936219 Chr.4: 99204547-99204547 on Build GRCh38
rs28720163 Chr.4: 99204666-99204666 on Build GRCh38
rs1019846422 Chr.4: 99204320-99204320 on Build GRCh38
rs1031701925 Chr.4: 99204655-99204655 on Build GRCh38
rs927902031 Chr.4: 99204656-99204657 on Build GRCh38
rs563664971 Chr.4: 99204450-99204450 on Build GRCh38
rs1036955680 Chr.4: 99204483-99204483 on Build GRCh38
rs562184543 Chr.4: 99204586-99204586 on Build GRCh38
rs576771286 Chr.4: 99204657-99204657 on Build GRCh38
rs748989669 Chr.4: 99204264-99204264 on Build GRCh38
rs554378657 Chr.4: 99204634-99204634 on Build GRCh38
rs545286612 Chr.4: 99204615-99204615 on Build GRCh38
rs568258837 Chr.4: 99204607-99204607 on Build GRCh38
rs189708916 Chr.4: 99204530-99204530 on Build GRCh38
rs1025723958 Chr.4: 99204392-99204393 on Build GRCh38
rs750681199 Chr.4: 99204252-99204254 on Build GRCh38
rs903866299 Chr.4: 99204647-99204647 on Build GRCh38
rs899824610 Chr.4: 99204484-99204484 on Build GRCh38
rs933852889 Chr.4: 99204506-99204506 on Build GRCh38
rs982014269 Chr.4: 99204636-99204636 on Build GRCh38
rs927597716 Chr.4: 99204252-99204253 on Build GRCh38
rs979823558 Chr.4: 99204445-99204445 on Build GRCh38
rs147839189 Chr.4: 99204371-99204371 on Build GRCh38
rs115864341 Chr.4: 99204743-99204743 on Build GRCh38
rs530796408 Chr.4: 99204498-99204498 on Build GRCh38
rs999479236 Chr.4: 99204654-99204654 on Build GRCh38
rs529189289 Chr.4: 99204644-99204644 on Build GRCh38
rs1050201146 Chr.4: 99204713-99204713 on Build GRCh38
rs1007071027 Chr.4: 99204292-99204292 on Build GRCh38
rs761254585 Chr.4: 99204289-99204289 on Build GRCh38
rs184868258 Chr.4: 99204526-99204526 on Build GRCh38
rs935014933 Chr.4: 99204254-99204254 on Build GRCh38
rs917013043 Chr.4: 99204409-99204409 on Build GRCh38
rs758702316 Chr.4: 99204254-99204254 on Build GRCh38
rs897068655 Chr.4: 99204505-99204505 on Build GRCh38
rs992329337 Chr.4: 99204341-99204341 on Build GRCh38
rs996825943 Chr.4: 99204553-99204553 on Build GRCh38
rs545539768 Chr.4: 99204428-99204428 on Build GRCh38
rs142327148 Chr.4: 99204262-99204262 on Build GRCh38
rs148900043 Chr.4: 99204703-99204703 on Build GRCh38
rs146361551 Chr.4: 99204263-99204263 on Build GRCh38
rs764922196 Chr.4: 99204694-99204694 on Build GRCh38
rs12507078 Chr.4: 99204711-99204711 on Build GRCh38
rs979858056 Chr.4: 99204746-99204746 on Build GRCh38
rs768253305 Chr.4: 99204268-99204268 on Build GRCh38
rs745792482 Chr.4: 99204260-99204260 on Build GRCh38
rs909563057 Chr.4: 99204449-99204449 on Build GRCh38
rs773851508 Chr.4: 99204279-99204279 on Build GRCh38
rs753373920 Chr.4: 99204391-99204391 on Build GRCh38
rs1052493617 Chr.4: 99204673-99204673 on Build GRCh38
rs1006941672 Chr.4: 99204595-99204595 on Build GRCh38
rs889893165 Chr.4: 99204584-99204584 on Build GRCh38

View Full Product Details