Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 152536830-152537327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKFZP434I0714
Entrez Gene ID
54553
Gene Name
uncharacterized protein DKFZP434I0714
Gene Aliases
-
UniGene
Hs.142307
Gene Chromosome Location
Chr.4: 152536264-152539263 on Build GRCh38
Transcripts
NR_033797.2

SNP Detail

SNP ID Chromosome Location
rs572282312 Chr.4: 152537249-152537249 on Build GRCh38
rs1027979363 Chr.4: 152536937-152536937 on Build GRCh38
rs972481734 Chr.4: 152537290-152537290 on Build GRCh38
rs531393603 Chr.4: 152536931-152536931 on Build GRCh38
rs1043860400 Chr.4: 152536945-152536945 on Build GRCh38
rs545757642 Chr.4: 152536867-152536867 on Build GRCh38
rs957605661 Chr.4: 152536896-152536896 on Build GRCh38
rs896672395 Chr.4: 152537029-152537029 on Build GRCh38
rs1052155715 Chr.4: 152537230-152537230 on Build GRCh38
rs781182227 Chr.4: 152536978-152536978 on Build GRCh38
rs984847444 Chr.4: 152537120-152537120 on Build GRCh38
rs543977515 Chr.4: 152537038-152537038 on Build GRCh38
rs961418006 Chr.4: 152537270-152537270 on Build GRCh38
rs564340486 Chr.4: 152536868-152536868 on Build GRCh38
rs903643082 Chr.4: 152536948-152536948 on Build GRCh38
rs745348850 Chr.4: 152537213-152537213 on Build GRCh38
rs896620273 Chr.4: 152537248-152537248 on Build GRCh38
rs559468232 Chr.4: 152537139-152537139 on Build GRCh38
rs1046247273 Chr.4: 152536882-152536882 on Build GRCh38
rs1023893419 Chr.4: 152537118-152537118 on Build GRCh38
rs6817854 Chr.4: 152537152-152537152 on Build GRCh38
rs1045721734 Chr.4: 152537273-152537273 on Build GRCh38
rs953857437 Chr.4: 152536942-152536942 on Build GRCh38
rs972971516 Chr.4: 152537040-152537040 on Build GRCh38
rs753134654 Chr.4: 152536968-152536968 on Build GRCh38
rs529421522 Chr.4: 152537119-152537119 on Build GRCh38
rs537202340 Chr.4: 152537163-152537163 on Build GRCh38
rs894506790 Chr.4: 152536875-152536875 on Build GRCh38
rs986541598 Chr.4: 152536963-152536963 on Build GRCh38
rs935053476 Chr.4: 152537224-152537224 on Build GRCh38
rs562115806 Chr.4: 152537101-152537101 on Build GRCh38
rs770592425 Chr.4: 152537181-152537181 on Build GRCh38
rs1013769081 Chr.4: 152537097-152537097 on Build GRCh38
rs776853513 Chr.4: 152536854-152536855 on Build GRCh38
rs562305468 Chr.4: 152537026-152537026 on Build GRCh38
rs73863137 Chr.4: 152537146-152537146 on Build GRCh38
rs961201570 Chr.4: 152537023-152537023 on Build GRCh38
rs150698886 Chr.4: 152537128-152537128 on Build GRCh38
rs1013739821 Chr.4: 152537271-152537271 on Build GRCh38
rs962149194 Chr.4: 152537138-152537138 on Build GRCh38
rs1011857903 Chr.4: 152536901-152536901 on Build GRCh38
rs1043424587 Chr.4: 152537211-152537211 on Build GRCh38
rs750511893 Chr.4: 152536940-152536940 on Build GRCh38
rs1028596164 Chr.4: 152537203-152537203 on Build GRCh38
rs778583404 Chr.4: 152537198-152537198 on Build GRCh38
rs375840609 Chr.4: 152537160-152537160 on Build GRCh38
rs200096244 Chr.4: 152537060-152537063 on Build GRCh38
rs4696323 Chr.4: 152536889-152536889 on Build GRCh38
rs999433575 Chr.4: 152536887-152536887 on Build GRCh38
rs1051024702 Chr.4: 152536878-152536878 on Build GRCh38
rs560054726 Chr.4: 152537167-152537167 on Build GRCh38
rs907345972 Chr.4: 152536884-152536884 on Build GRCh38
rs1008314080 Chr.4: 152537236-152537236 on Build GRCh38

View Full Product Details