Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 152537590-152538096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKFZP434I0714
Entrez Gene ID
54553
Gene Name
uncharacterized protein DKFZP434I0714
Gene Aliases
-
UniGene
Hs.142307
Gene Chromosome Location
Chr.4: 152536264-152539263 on Build GRCh38
Transcripts
NR_033797.2

SNP Detail

SNP ID Chromosome Location
rs897793544 Chr.4: 152537859-152537859 on Build GRCh38
rs149885595 Chr.4: 152537852-152537852 on Build GRCh38
rs947484723 Chr.4: 152537983-152537983 on Build GRCh38
rs896793772 Chr.4: 152538057-152538057 on Build GRCh38
rs917703690 Chr.4: 152537726-152537726 on Build GRCh38
rs987790090 Chr.4: 152538020-152538020 on Build GRCh38
rs763109378 Chr.4: 152537869-152537869 on Build GRCh38
rs988852546 Chr.4: 152537914-152537914 on Build GRCh38
rs896850200 Chr.4: 152537691-152537691 on Build GRCh38
rs949135374 Chr.4: 152538038-152538038 on Build GRCh38
rs1008466870 Chr.4: 152537903-152537903 on Build GRCh38
rs756036450 Chr.4: 152537949-152537949 on Build GRCh38
rs753607404 Chr.4: 152538026-152538026 on Build GRCh38
rs78527666 Chr.4: 152537737-152537737 on Build GRCh38
rs541524410 Chr.4: 152537935-152537935 on Build GRCh38
rs993797505 Chr.4: 152538063-152538065 on Build GRCh38
rs765528161 Chr.4: 152537647-152537647 on Build GRCh38
rs1023504015 Chr.4: 152537818-152537818 on Build GRCh38
rs376110922 Chr.4: 152538072-152538075 on Build GRCh38
rs956370068 Chr.4: 152538013-152538013 on Build GRCh38
rs925212263 Chr.4: 152538012-152538012 on Build GRCh38
rs766378640 Chr.4: 152537900-152537900 on Build GRCh38
rs890025268 Chr.4: 152537622-152537622 on Build GRCh38
rs1008516918 Chr.4: 152537626-152537626 on Build GRCh38
rs1019682677 Chr.4: 152537632-152537632 on Build GRCh38
rs1012043329 Chr.4: 152537753-152537753 on Build GRCh38
rs935002113 Chr.4: 152537668-152537668 on Build GRCh38
rs769558826 Chr.4: 152537998-152537998 on Build GRCh38
rs964239977 Chr.4: 152537978-152537978 on Build GRCh38
rs1038359890 Chr.4: 152537858-152537858 on Build GRCh38
rs971031831 Chr.4: 152537822-152537822 on Build GRCh38
rs1052101267 Chr.4: 152537681-152537681 on Build GRCh38
rs570695929 Chr.4: 152537990-152537990 on Build GRCh38
rs1026709556 Chr.4: 152537734-152537734 on Build GRCh38
rs185327490 Chr.4: 152537937-152537937 on Build GRCh38
rs982080010 Chr.4: 152537841-152537841 on Build GRCh38
rs1030419521 Chr.4: 152537895-152537895 on Build GRCh38
rs905371862 Chr.4: 152537759-152537759 on Build GRCh38
rs949564340 Chr.4: 152537729-152537729 on Build GRCh38
rs562177093 Chr.4: 152537845-152537845 on Build GRCh38

View Full Product Details