Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 152537974-152538484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKFZP434I0714
Entrez Gene ID
54553
Gene Name
uncharacterized protein DKFZP434I0714
Gene Aliases
-
UniGene
Hs.142307
Gene Chromosome Location
Chr.4: 152536264-152539263 on Build GRCh38
Transcripts
NR_033797.2

SNP Detail

SNP ID Chromosome Location
rs1041654007 Chr.4: 152538380-152538380 on Build GRCh38
rs918847956 Chr.4: 152538386-152538386 on Build GRCh38
rs914613350 Chr.4: 152538349-152538349 on Build GRCh38
rs896793772 Chr.4: 152538057-152538057 on Build GRCh38
rs942199126 Chr.4: 152538134-152538134 on Build GRCh38
rs987790090 Chr.4: 152538020-152538020 on Build GRCh38
rs533172698 Chr.4: 152538142-152538142 on Build GRCh38
rs988801632 Chr.4: 152538323-152538323 on Build GRCh38
rs985593348 Chr.4: 152538361-152538361 on Build GRCh38
rs1051796942 Chr.4: 152538259-152538259 on Build GRCh38
rs75092985 Chr.4: 152538304-152538304 on Build GRCh38
rs147165309 Chr.4: 152538456-152538456 on Build GRCh38
rs949135374 Chr.4: 152538038-152538038 on Build GRCh38
rs753607404 Chr.4: 152538026-152538026 on Build GRCh38
rs1351902 Chr.4: 152538373-152538373 on Build GRCh38
rs970599813 Chr.4: 152538221-152538221 on Build GRCh38
rs890468982 Chr.4: 152538303-152538303 on Build GRCh38
rs993797505 Chr.4: 152538063-152538065 on Build GRCh38
rs779461668 Chr.4: 152538416-152538416 on Build GRCh38
rs898120386 Chr.4: 152538234-152538234 on Build GRCh38
rs376110922 Chr.4: 152538072-152538075 on Build GRCh38
rs956370068 Chr.4: 152538013-152538013 on Build GRCh38
rs925212263 Chr.4: 152538012-152538012 on Build GRCh38
rs1032201439 Chr.4: 152538375-152538375 on Build GRCh38
rs888189816 Chr.4: 152538090-152538090 on Build GRCh38
rs1011992224 Chr.4: 152538103-152538103 on Build GRCh38
rs1048047971 Chr.4: 152538448-152538448 on Build GRCh38
rs757081547 Chr.4: 152538149-152538149 on Build GRCh38
rs911353368 Chr.4: 152538367-152538367 on Build GRCh38
rs929276141 Chr.4: 152538245-152538245 on Build GRCh38
rs769558826 Chr.4: 152537998-152537998 on Build GRCh38
rs956650168 Chr.4: 152538387-152538387 on Build GRCh38
rs926769290 Chr.4: 152538128-152538128 on Build GRCh38
rs988031187 Chr.4: 152538441-152538441 on Build GRCh38
rs552055164 Chr.4: 152538191-152538191 on Build GRCh38
rs531026900 Chr.4: 152538352-152538352 on Build GRCh38
rs759566091 Chr.4: 152538449-152538450 on Build GRCh38
rs1023875166 Chr.4: 152538156-152538156 on Build GRCh38
rs1003846089 Chr.4: 152538250-152538250 on Build GRCh38
rs1024573561 Chr.4: 152538452-152538452 on Build GRCh38
rs980945949 Chr.4: 152538124-152538124 on Build GRCh38
rs1018108386 Chr.4: 152538311-152538311 on Build GRCh38
rs867749554 Chr.4: 152538286-152538286 on Build GRCh38

View Full Product Details