Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 6312463-6312957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ33360
Entrez Gene ID
401172
Gene Name
FLJ33360 protein
Gene Aliases
-
UniGene
Hs.376760
Gene Chromosome Location
Chr.5: 6310441-6337292 on Build GRCh38
Transcripts
NR_028351.1

SNP Detail

SNP ID Chromosome Location
rs758658504 Chr.5: 6312732-6312737 on Build GRCh38
rs879642461 Chr.5: 6312934-6312934 on Build GRCh38
rs758190588 Chr.5: 6312737-6312737 on Build GRCh38
rs575425477 Chr.5: 6312563-6312563 on Build GRCh38
rs746941884 Chr.5: 6312750-6312750 on Build GRCh38
rs759907997 Chr.5: 6312590-6312590 on Build GRCh38
rs539721145 Chr.5: 6312917-6312917 on Build GRCh38
rs764838139 Chr.5: 6312731-6312732 on Build GRCh38
rs757567049 Chr.5: 6312516-6312516 on Build GRCh38
rs530972297 Chr.5: 6312880-6312880 on Build GRCh38
rs543645533 Chr.5: 6312600-6312600 on Build GRCh38
rs761179894 Chr.5: 6312608-6312608 on Build GRCh38
rs997107061 Chr.5: 6312526-6312526 on Build GRCh38
rs111894906 Chr.5: 6312663-6312789 on Build GRCh38
rs762783262 Chr.5: 6312873-6312873 on Build GRCh38
rs772097415 Chr.5: 6312621-6312623 on Build GRCh38
rs62330484 Chr.5: 6312770-6312770 on Build GRCh38
rs1003095308 Chr.5: 6312624-6312624 on Build GRCh38
rs914805521 Chr.5: 6312743-6312743 on Build GRCh38
rs546108105 Chr.5: 6312660-6312660 on Build GRCh38
rs188461013 Chr.5: 6312658-6312658 on Build GRCh38
rs750978414 Chr.5: 6312757-6312761 on Build GRCh38
rs139268040 Chr.5: 6312741-6312742 on Build GRCh38
rs201071355 Chr.5: 6312583-6312583 on Build GRCh38
rs62330483 Chr.5: 6312764-6312764 on Build GRCh38
rs796802906 Chr.5: 6312644-6312644 on Build GRCh38
rs951977304 Chr.5: 6312635-6312635 on Build GRCh38
rs754032040 Chr.5: 6312649-6312649 on Build GRCh38
rs778449580 Chr.5: 6312761-6312770 on Build GRCh38
rs1055119256 Chr.5: 6312719-6312719 on Build GRCh38
rs776141064 Chr.5: 6312737-6312738 on Build GRCh38
rs376858383 Chr.5: 6312500-6312500 on Build GRCh38
rs111416070 Chr.5: 6312732-6312739 on Build GRCh38
rs574681209 Chr.5: 6312545-6312545 on Build GRCh38
rs372499721 Chr.5: 6312616-6312616 on Build GRCh38
rs62330482 Chr.5: 6312762-6312762 on Build GRCh38
rs142405388 Chr.5: 6312732-6312741 on Build GRCh38
rs924269479 Chr.5: 6312670-6312670 on Build GRCh38
rs1048211271 Chr.5: 6312539-6312539 on Build GRCh38
rs757739993 Chr.5: 6312765-6312768 on Build GRCh38
rs760459076 Chr.5: 6312732-6312743 on Build GRCh38
rs2168655 Chr.5: 6312766-6312766 on Build GRCh38
rs771792980 Chr.5: 6312534-6312534 on Build GRCh38
rs532327107 Chr.5: 6312653-6312653 on Build GRCh38
rs992098215 Chr.5: 6312676-6312676 on Build GRCh38
rs776243249 Chr.5: 6312487-6312487 on Build GRCh38
rs62330481 Chr.5: 6312760-6312760 on Build GRCh38
rs766644599 Chr.5: 6312489-6312489 on Build GRCh38
rs981111566 Chr.5: 6312669-6312669 on Build GRCh38
rs756201670 Chr.5: 6312755-6312768 on Build GRCh38
rs993407765 Chr.5: 6312626-6312626 on Build GRCh38
rs79006354 Chr.5: 6312834-6312834 on Build GRCh38
rs1013196100 Chr.5: 6312920-6312920 on Build GRCh38
rs12655089 Chr.5: 6312915-6312915 on Build GRCh38
rs745780750 Chr.5: 6312757-6312768 on Build GRCh38
rs1036712105 Chr.5: 6312910-6312910 on Build GRCh38
rs62330480 Chr.5: 6312758-6312758 on Build GRCh38
rs768020710 Chr.5: 6312597-6312597 on Build GRCh38
rs779177946 Chr.5: 6312766-6312767 on Build GRCh38
rs970333083 Chr.5: 6312594-6312594 on Build GRCh38
rs749633024 Chr.5: 6312764-6312765 on Build GRCh38
rs745619238 Chr.5: 6312520-6312520 on Build GRCh38
rs191593224 Chr.5: 6312882-6312882 on Build GRCh38
rs776214281 Chr.5: 6312588-6312588 on Build GRCh38
rs13162189 Chr.5: 6312768-6312768 on Build GRCh38
rs1008314975 Chr.5: 6312814-6312814 on Build GRCh38
rs34537542 Chr.5: 6312787-6312787 on Build GRCh38
rs1027540950 Chr.5: 6312628-6312628 on Build GRCh38
rs996986836 Chr.5: 6312518-6312518 on Build GRCh38
rs746787754 Chr.5: 6312883-6312884 on Build GRCh38
rs761355938 Chr.5: 6312555-6312555 on Build GRCh38
rs764629300 Chr.5: 6312755-6312759 on Build GRCh38
rs563540555 Chr.5: 6312627-6312627 on Build GRCh38
rs762365695 Chr.5: 6312599-6312599 on Build GRCh38
rs747111268 Chr.5: 6312767-6312770 on Build GRCh38
rs776186184 Chr.5: 6312550-6312550 on Build GRCh38
rs144125299 Chr.5: 6312483-6312484 on Build GRCh38
rs778051386 Chr.5: 6312506-6312506 on Build GRCh38
rs369873565 Chr.5: 6312541-6312541 on Build GRCh38
rs897167040 Chr.5: 6312833-6312833 on Build GRCh38
rs749490931 Chr.5: 6312515-6312515 on Build GRCh38
rs751085844 Chr.5: 6312759-6312768 on Build GRCh38
rs768122503 Chr.5: 6312546-6312546 on Build GRCh38
rs995482758 Chr.5: 6312848-6312848 on Build GRCh38
rs1015586313 Chr.5: 6312557-6312557 on Build GRCh38
rs778561166 Chr.5: 6312519-6312519 on Build GRCh38
rs750091837 Chr.5: 6312732-6312735 on Build GRCh38
rs559072583 Chr.5: 6312769-6312769 on Build GRCh38
rs977468343 Chr.5: 6312728-6312728 on Build GRCh38
rs867768308 Chr.5: 6312587-6312587 on Build GRCh38
rs932576961 Chr.5: 6312823-6312823 on Build GRCh38
rs200121547 Chr.5: 6312582-6312582 on Build GRCh38
rs756632806 Chr.5: 6312761-6312768 on Build GRCh38
rs757912698 Chr.5: 6312735-6312736 on Build GRCh38
rs1003187197 Chr.5: 6312928-6312928 on Build GRCh38
rs397996712 Chr.5: 6312767-6312768 on Build GRCh38
rs756217808 Chr.5: 6312505-6312505 on Build GRCh38
rs752015983 Chr.5: 6312496-6312496 on Build GRCh38
rs547979834 Chr.5: 6312829-6312829 on Build GRCh38
rs530942697 Chr.5: 6312719-6312720 on Build GRCh38
rs757299434 Chr.5: 6312652-6312652 on Build GRCh38
rs779863385 Chr.5: 6312739-6312739 on Build GRCh38
rs753290217 Chr.5: 6312504-6312504 on Build GRCh38
rs1041047817 Chr.5: 6312759-6312770 on Build GRCh38
rs1050287705 Chr.5: 6312522-6312522 on Build GRCh38
rs761489737 Chr.5: 6312488-6312488 on Build GRCh38
rs934291904 Chr.5: 6312704-6312704 on Build GRCh38
rs775387746 Chr.5: 6312779-6312779 on Build GRCh38
rs765113148 Chr.5: 6312665-6312665 on Build GRCh38

View Full Product Details