Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 81237418-81237925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKMT2
Entrez Gene ID
1160
Gene Name
creatine kinase, mitochondrial 2
Gene Aliases
SMTCK
UniGene
Hs.80691
Gene Chromosome Location
Chr.5: 81233320-81266398 on Build GRCh38
Transcripts
NM_001099735.1 NM_001099736.1 NM_001825.2 NR_034121.1 NR_034122.1 NR_034123.1

Gene Symbol
CKMT2-AS1
Entrez Gene ID
100131067
Gene Name
CKMT2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.655855
Gene Chromosome Location
Chr.5: 81237565-81301569 on Build GRCh38
Transcripts
NM_001099735.1 NM_001099736.1 NM_001825.2 NR_034121.1 NR_034122.1 NR_034123.1

SNP Detail

SNP ID Chromosome Location
rs977393980 Chr.5: 81237826-81237826 on Build GRCh38
rs116353325 Chr.5: 81237445-81237445 on Build GRCh38
rs971225217 Chr.5: 81237603-81237603 on Build GRCh38
rs796850999 Chr.5: 81237520-81237520 on Build GRCh38
rs77804833 Chr.5: 81237677-81237677 on Build GRCh38
rs1042319897 Chr.5: 81237757-81237757 on Build GRCh38
rs557565833 Chr.5: 81237642-81237642 on Build GRCh38
rs538305080 Chr.5: 81237850-81237850 on Build GRCh38
rs11299817 Chr.5: 81237515-81237515 on Build GRCh38
rs147017254 Chr.5: 81237673-81237673 on Build GRCh38
rs932338776 Chr.5: 81237733-81237733 on Build GRCh38
rs922231370 Chr.5: 81237567-81237567 on Build GRCh38
rs557101304 Chr.5: 81237789-81237789 on Build GRCh38
rs4704707 Chr.5: 81237540-81237540 on Build GRCh38
rs186821033 Chr.5: 81237667-81237667 on Build GRCh38
rs1020523320 Chr.5: 81237480-81237480 on Build GRCh38
rs189693128 Chr.5: 81237722-81237722 on Build GRCh38
rs548200963 Chr.5: 81237898-81237898 on Build GRCh38
rs543597525 Chr.5: 81237770-81237772 on Build GRCh38
rs1033753732 Chr.5: 81237774-81237774 on Build GRCh38
rs890900744 Chr.5: 81237746-81237746 on Build GRCh38
rs1021517189 Chr.5: 81237802-81237802 on Build GRCh38
rs906513254 Chr.5: 81237757-81237758 on Build GRCh38
rs148059284 Chr.5: 81237720-81237720 on Build GRCh38
rs1024379726 Chr.5: 81237856-81237856 on Build GRCh38
rs569289741 Chr.5: 81237768-81237768 on Build GRCh38
rs574013865 Chr.5: 81237853-81237853 on Build GRCh38
rs1025359179 Chr.5: 81237450-81237450 on Build GRCh38
rs917337702 Chr.5: 81237573-81237573 on Build GRCh38
rs141317527 Chr.5: 81237558-81237558 on Build GRCh38
rs377734078 Chr.5: 81237818-81237818 on Build GRCh38
rs932433657 Chr.5: 81237588-81237588 on Build GRCh38
rs118018665 Chr.5: 81237482-81237482 on Build GRCh38
rs958670024 Chr.5: 81237803-81237803 on Build GRCh38
rs766178721 Chr.5: 81237801-81237801 on Build GRCh38
rs897075231 Chr.5: 81237750-81237750 on Build GRCh38
rs1009050281 Chr.5: 81237753-81237753 on Build GRCh38
rs909645602 Chr.5: 81237880-81237880 on Build GRCh38
rs762796824 Chr.5: 81237788-81237788 on Build GRCh38
rs1002274909 Chr.5: 81237478-81237478 on Build GRCh38
rs1009493863 Chr.5: 81237460-81237460 on Build GRCh38
rs753373993 Chr.5: 81237645-81237645 on Build GRCh38
rs903430389 Chr.5: 81237771-81237771 on Build GRCh38
rs920890625 Chr.5: 81237717-81237717 on Build GRCh38
rs561632326 Chr.5: 81237544-81237544 on Build GRCh38
rs922316901 Chr.5: 81237848-81237848 on Build GRCh38
rs1050838742 Chr.5: 81237744-81237744 on Build GRCh38
rs576065944 Chr.5: 81237487-81237487 on Build GRCh38
rs983048799 Chr.5: 81237629-81237629 on Build GRCh38
rs532643377 Chr.5: 81237674-81237674 on Build GRCh38
rs1011077101 Chr.5: 81237798-81237798 on Build GRCh38
rs958188020 Chr.5: 81237503-81237503 on Build GRCh38
rs1000196565 Chr.5: 81237511-81237511 on Build GRCh38

View Full Product Details