Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 81238517-81239062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKMT2
Entrez Gene ID
1160
Gene Name
creatine kinase, mitochondrial 2
Gene Aliases
SMTCK
UniGene
Hs.80691
Gene Chromosome Location
Chr.5: 81233320-81266398 on Build GRCh38
Transcripts
NM_001099735.1 NM_001099736.1 NM_001825.2 NR_034121.1 NR_034122.1 NR_034123.1

Gene Symbol
CKMT2-AS1
Entrez Gene ID
100131067
Gene Name
CKMT2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.655855
Gene Chromosome Location
Chr.5: 81237565-81301569 on Build GRCh38
Transcripts
NM_001099735.1 NM_001099736.1 NM_001825.2 NR_034121.1 NR_034122.1 NR_034123.1

SNP Detail

SNP ID Chromosome Location
rs1049774846 Chr.5: 81238806-81238806 on Build GRCh38
rs372561954 Chr.5: 81238970-81238970 on Build GRCh38
rs920194303 Chr.5: 81239028-81239028 on Build GRCh38
rs898759861 Chr.5: 81238665-81238665 on Build GRCh38
rs958412065 Chr.5: 81238770-81238770 on Build GRCh38
rs1040924023 Chr.5: 81238834-81238834 on Build GRCh38
rs777366834 Chr.5: 81238627-81238627 on Build GRCh38
rs906848796 Chr.5: 81238718-81238718 on Build GRCh38
rs1008461773 Chr.5: 81238827-81238827 on Build GRCh38
rs115420437 Chr.5: 81238636-81238636 on Build GRCh38
rs547401548 Chr.5: 81238628-81238628 on Build GRCh38
rs938336524 Chr.5: 81239034-81239034 on Build GRCh38
rs1037251810 Chr.5: 81238772-81238772 on Build GRCh38
rs565799422 Chr.5: 81238817-81238817 on Build GRCh38
rs748534722 Chr.5: 81239018-81239018 on Build GRCh38
rs940317349 Chr.5: 81238765-81238765 on Build GRCh38
rs970617258 Chr.5: 81238735-81238735 on Build GRCh38
rs928876853 Chr.5: 81238761-81238761 on Build GRCh38
rs6888427 Chr.5: 81238616-81238616 on Build GRCh38
rs35823599 Chr.5: 81238773-81238774 on Build GRCh38
rs577523038 Chr.5: 81238980-81238980 on Build GRCh38
rs923962851 Chr.5: 81238833-81238833 on Build GRCh38
rs774583508 Chr.5: 81239017-81239017 on Build GRCh38
rs942761593 Chr.5: 81238931-81238931 on Build GRCh38
rs780477346 Chr.5: 81238822-81238822 on Build GRCh38
rs1032264748 Chr.5: 81238895-81238895 on Build GRCh38
rs982008507 Chr.5: 81238945-81238945 on Build GRCh38
rs181451642 Chr.5: 81238576-81238576 on Build GRCh38
rs189849048 Chr.5: 81238872-81238872 on Build GRCh38
rs1011947921 Chr.5: 81238912-81238912 on Build GRCh38
rs981656513 Chr.5: 81238745-81238745 on Build GRCh38
rs1047306981 Chr.5: 81239021-81239021 on Build GRCh38
rs966677899 Chr.5: 81238613-81238613 on Build GRCh38
rs1021436078 Chr.5: 81238804-81238804 on Build GRCh38
rs999410016 Chr.5: 81238889-81238889 on Build GRCh38
rs920093225 Chr.5: 81238995-81238995 on Build GRCh38
rs1047197269 Chr.5: 81238671-81238671 on Build GRCh38
rs931329477 Chr.5: 81238803-81238803 on Build GRCh38
rs185039053 Chr.5: 81238759-81238759 on Build GRCh38
rs117042618 Chr.5: 81238933-81238933 on Build GRCh38
rs1055429623 Chr.5: 81239038-81239038 on Build GRCh38
rs770095573 Chr.5: 81238782-81238782 on Build GRCh38
rs1032046421 Chr.5: 81238620-81238620 on Build GRCh38
rs562948083 Chr.5: 81238565-81238565 on Build GRCh38
rs916346421 Chr.5: 81238706-81238706 on Build GRCh38
rs772661610 Chr.5: 81239026-81239026 on Build GRCh38
rs1035827605 Chr.5: 81238978-81238978 on Build GRCh38
rs1038459653 Chr.5: 81238650-81238650 on Build GRCh38
rs200040414 Chr.5: 81238668-81238668 on Build GRCh38
rs76661037 Chr.5: 81238597-81238597 on Build GRCh38
rs930244414 Chr.5: 81239008-81239008 on Build GRCh38
rs535676063 Chr.5: 81238781-81238781 on Build GRCh38
rs1019938035 Chr.5: 81238606-81238606 on Build GRCh38
rs1002662736 Chr.5: 81238744-81238744 on Build GRCh38
rs559048423 Chr.5: 81238774-81238775 on Build GRCh38
rs942877729 Chr.5: 81238644-81238644 on Build GRCh38
rs1049801165 Chr.5: 81238575-81238575 on Build GRCh38
rs533086931 Chr.5: 81238700-81238700 on Build GRCh38
rs749350072 Chr.5: 81238901-81238901 on Build GRCh38
rs977355006 Chr.5: 81238832-81238832 on Build GRCh38
rs555587836 Chr.5: 81238795-81238795 on Build GRCh38
rs994401347 Chr.5: 81238666-81238666 on Build GRCh38
rs550090964 Chr.5: 81238742-81238742 on Build GRCh38
rs148583851 Chr.5: 81239033-81239033 on Build GRCh38
rs527764671 Chr.5: 81238624-81238624 on Build GRCh38
rs548998536 Chr.5: 81238923-81238923 on Build GRCh38

View Full Product Details