Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 81238943-81239443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKMT2
Entrez Gene ID
1160
Gene Name
creatine kinase, mitochondrial 2
Gene Aliases
SMTCK
UniGene
Hs.80691
Gene Chromosome Location
Chr.5: 81233320-81266398 on Build GRCh38
Transcripts
NM_001099735.1 NM_001099736.1 NM_001825.2 NR_034121.1 NR_034122.1 NR_034123.1

Gene Symbol
CKMT2-AS1
Entrez Gene ID
100131067
Gene Name
CKMT2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.655855
Gene Chromosome Location
Chr.5: 81237565-81301569 on Build GRCh38
Transcripts
NM_001099735.1 NM_001099736.1 NM_001825.2 NR_034121.1 NR_034122.1 NR_034123.1

SNP Detail

SNP ID Chromosome Location
rs944039983 Chr.5: 81239289-81239289 on Build GRCh38
rs527801488 Chr.5: 81239181-81239181 on Build GRCh38
rs556860441 Chr.5: 81239137-81239137 on Build GRCh38
rs372561954 Chr.5: 81238970-81238970 on Build GRCh38
rs780198216 Chr.5: 81239138-81239138 on Build GRCh38
rs920194303 Chr.5: 81239028-81239028 on Build GRCh38
rs1011915259 Chr.5: 81239173-81239173 on Build GRCh38
rs758229049 Chr.5: 81239397-81239397 on Build GRCh38
rs982990621 Chr.5: 81239401-81239401 on Build GRCh38
rs1053658487 Chr.5: 81239400-81239400 on Build GRCh38
rs964011372 Chr.5: 81239391-81239394 on Build GRCh38
rs928875483 Chr.5: 81239239-81239239 on Build GRCh38
rs541079387 Chr.5: 81239199-81239199 on Build GRCh38
rs142950211 Chr.5: 81239210-81239210 on Build GRCh38
rs938336524 Chr.5: 81239034-81239034 on Build GRCh38
rs748534722 Chr.5: 81239018-81239018 on Build GRCh38
rs1007416156 Chr.5: 81239378-81239378 on Build GRCh38
rs542053196 Chr.5: 81239144-81239144 on Build GRCh38
rs1042811732 Chr.5: 81239310-81239310 on Build GRCh38
rs577523038 Chr.5: 81238980-81238980 on Build GRCh38
rs764670814 Chr.5: 81239332-81239332 on Build GRCh38
rs774583508 Chr.5: 81239017-81239017 on Build GRCh38
rs868830989 Chr.5: 81239089-81239089 on Build GRCh38
rs1018200117 Chr.5: 81239171-81239171 on Build GRCh38
rs998699974 Chr.5: 81239354-81239354 on Build GRCh38
rs1047306981 Chr.5: 81239021-81239021 on Build GRCh38
rs148874583 Chr.5: 81239395-81239396 on Build GRCh38
rs920093225 Chr.5: 81238995-81238995 on Build GRCh38
rs1053690062 Chr.5: 81239161-81239161 on Build GRCh38
rs1041535045 Chr.5: 81239100-81239100 on Build GRCh38
rs1055429623 Chr.5: 81239038-81239038 on Build GRCh38
rs114859511 Chr.5: 81239143-81239143 on Build GRCh38
rs183155785 Chr.5: 81239275-81239275 on Build GRCh38
rs772661610 Chr.5: 81239026-81239026 on Build GRCh38
rs1035827605 Chr.5: 81238978-81238978 on Build GRCh38
rs930244414 Chr.5: 81239008-81239008 on Build GRCh38
rs1027177726 Chr.5: 81239220-81239220 on Build GRCh38
rs564415461 Chr.5: 81239205-81239205 on Build GRCh38
rs906197398 Chr.5: 81239180-81239180 on Build GRCh38
rs549885212 Chr.5: 81239253-81239253 on Build GRCh38
rs911192368 Chr.5: 81239285-81239285 on Build GRCh38
rs985859290 Chr.5: 81239284-81239284 on Build GRCh38
rs1003253374 Chr.5: 81239189-81239189 on Build GRCh38
rs961494339 Chr.5: 81239209-81239209 on Build GRCh38
rs759032731 Chr.5: 81239322-81239322 on Build GRCh38
rs561911980 Chr.5: 81239176-81239176 on Build GRCh38
rs893751211 Chr.5: 81239172-81239172 on Build GRCh38
rs944171682 Chr.5: 81239077-81239077 on Build GRCh38
rs10942945 Chr.5: 81239368-81239368 on Build GRCh38
rs894247923 Chr.5: 81239047-81239047 on Build GRCh38
rs902999894 Chr.5: 81239099-81239099 on Build GRCh38
rs893718760 Chr.5: 81239416-81239416 on Build GRCh38
rs985503565 Chr.5: 81239071-81239071 on Build GRCh38
rs879428646 Chr.5: 81239382-81239382 on Build GRCh38
rs911294743 Chr.5: 81239076-81239076 on Build GRCh38
rs1030254952 Chr.5: 81239122-81239122 on Build GRCh38
rs1030096737 Chr.5: 81239356-81239356 on Build GRCh38
rs148583851 Chr.5: 81239033-81239033 on Build GRCh38
rs367679974 Chr.5: 81239179-81239179 on Build GRCh38

View Full Product Details