Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 81239311-81239790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKMT2
Entrez Gene ID
1160
Gene Name
creatine kinase, mitochondrial 2
Gene Aliases
SMTCK
UniGene
Hs.80691
Gene Chromosome Location
Chr.5: 81233320-81266398 on Build GRCh38
Transcripts
NM_001099735.1 NM_001099736.1 NM_001825.2 NR_034121.1 NR_034122.1 NR_034123.1

Gene Symbol
CKMT2-AS1
Entrez Gene ID
100131067
Gene Name
CKMT2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.655855
Gene Chromosome Location
Chr.5: 81237565-81301569 on Build GRCh38
Transcripts
NM_001099735.1 NM_001099736.1 NM_001825.2 NR_034121.1 NR_034122.1 NR_034123.1

SNP Detail

SNP ID Chromosome Location
rs956809147 Chr.5: 81239566-81239566 on Build GRCh38
rs758229049 Chr.5: 81239397-81239397 on Build GRCh38
rs916175654 Chr.5: 81239680-81239680 on Build GRCh38
rs982990621 Chr.5: 81239401-81239401 on Build GRCh38
rs1053658487 Chr.5: 81239400-81239400 on Build GRCh38
rs991724452 Chr.5: 81239452-81239452 on Build GRCh38
rs964011372 Chr.5: 81239391-81239394 on Build GRCh38
rs899043124 Chr.5: 81239607-81239607 on Build GRCh38
rs965378408 Chr.5: 81239511-81239511 on Build GRCh38
rs111709434 Chr.5: 81239729-81239729 on Build GRCh38
rs551493005 Chr.5: 81239529-81239529 on Build GRCh38
rs960194861 Chr.5: 81239655-81239655 on Build GRCh38
rs906173348 Chr.5: 81239451-81239451 on Build GRCh38
rs951509618 Chr.5: 81239619-81239619 on Build GRCh38
rs1007416156 Chr.5: 81239378-81239378 on Build GRCh38
rs947066651 Chr.5: 81239470-81239470 on Build GRCh38
rs947898255 Chr.5: 81239628-81239628 on Build GRCh38
rs537076041 Chr.5: 81239546-81239546 on Build GRCh38
rs960307626 Chr.5: 81239436-81239436 on Build GRCh38
rs998699974 Chr.5: 81239354-81239354 on Build GRCh38
rs991776804 Chr.5: 81239663-81239663 on Build GRCh38
rs565834502 Chr.5: 81239486-81239486 on Build GRCh38
rs148874583 Chr.5: 81239395-81239396 on Build GRCh38
rs549223818 Chr.5: 81239452-81239452 on Build GRCh38
rs1044957910 Chr.5: 81239681-81239681 on Build GRCh38
rs1038920142 Chr.5: 81239584-81239584 on Build GRCh38
rs186136473 Chr.5: 81239614-81239614 on Build GRCh38
rs915036910 Chr.5: 81239617-81239617 on Build GRCh38
rs897604990 Chr.5: 81239491-81239491 on Build GRCh38
rs372721197 Chr.5: 81239625-81239625 on Build GRCh38
rs1007469950 Chr.5: 81239579-81239579 on Build GRCh38
rs1045388397 Chr.5: 81239445-81239445 on Build GRCh38
rs534719141 Chr.5: 81239497-81239497 on Build GRCh38
rs1035886533 Chr.5: 81239647-81239647 on Build GRCh38
rs939014928 Chr.5: 81239753-81239753 on Build GRCh38
rs546190511 Chr.5: 81239754-81239754 on Build GRCh38
rs571249559 Chr.5: 81239540-81239540 on Build GRCh38
rs1007104157 Chr.5: 81239506-81239506 on Build GRCh38
rs978461439 Chr.5: 81239730-81239730 on Build GRCh38
rs947916870 Chr.5: 81239444-81239444 on Build GRCh38
rs10942945 Chr.5: 81239368-81239368 on Build GRCh38
rs1036028501 Chr.5: 81239768-81239768 on Build GRCh38
rs893718760 Chr.5: 81239416-81239416 on Build GRCh38
rs1051573832 Chr.5: 81239561-81239563 on Build GRCh38
rs924257440 Chr.5: 81239747-81239747 on Build GRCh38
rs1036102052 Chr.5: 81239459-81239459 on Build GRCh38
rs879428646 Chr.5: 81239382-81239382 on Build GRCh38
rs1018961113 Chr.5: 81239507-81239507 on Build GRCh38
rs952891399 Chr.5: 81239503-81239503 on Build GRCh38
rs1030096737 Chr.5: 81239356-81239356 on Build GRCh38
rs994294637 Chr.5: 81239496-81239496 on Build GRCh38

View Full Product Details