Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 81250592-81251098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKMT2
Entrez Gene ID
1160
Gene Name
creatine kinase, mitochondrial 2
Gene Aliases
SMTCK
UniGene
Hs.80691
Gene Chromosome Location
Chr.5: 81233320-81266398 on Build GRCh38
Transcripts
NM_001099735.1 NM_001099736.1 NM_001825.2 NR_034121.1 NR_034122.1 NR_034123.1

Gene Symbol
CKMT2-AS1
Entrez Gene ID
100131067
Gene Name
CKMT2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.655855
Gene Chromosome Location
Chr.5: 81237565-81301569 on Build GRCh38
Transcripts
NM_001099735.1 NM_001099736.1 NM_001825.2 NR_034121.1 NR_034122.1 NR_034123.1

SNP Detail

SNP ID Chromosome Location
rs946423331 Chr.5: 81250773-81250773 on Build GRCh38
rs998334280 Chr.5: 81250842-81250842 on Build GRCh38
rs953553967 Chr.5: 81250751-81250751 on Build GRCh38
rs373408385 Chr.5: 81250940-81250941 on Build GRCh38
rs973782666 Chr.5: 81250650-81250650 on Build GRCh38
rs368459891 Chr.5: 81250877-81250877 on Build GRCh38
rs1004326672 Chr.5: 81250657-81250657 on Build GRCh38
rs959546930 Chr.5: 81251033-81251033 on Build GRCh38
rs142418532 Chr.5: 81250941-81250952 on Build GRCh38
rs1027794441 Chr.5: 81250742-81250742 on Build GRCh38
rs3756636 Chr.5: 81250680-81250680 on Build GRCh38
rs1055312384 Chr.5: 81250735-81250735 on Build GRCh38
rs112820833 Chr.5: 81251020-81251020 on Build GRCh38
rs907150870 Chr.5: 81250978-81250978 on Build GRCh38
rs1015345992 Chr.5: 81250671-81250671 on Build GRCh38
rs919367237 Chr.5: 81250651-81250651 on Build GRCh38
rs867009146 Chr.5: 81250940-81250940 on Build GRCh38
rs1054483884 Chr.5: 81250876-81250876 on Build GRCh38
rs1045650184 Chr.5: 81250942-81250942 on Build GRCh38
rs906851892 Chr.5: 81250649-81250649 on Build GRCh38
rs376996280 Chr.5: 81250834-81250834 on Build GRCh38
rs560649390 Chr.5: 81250676-81250676 on Build GRCh38
rs1028098769 Chr.5: 81251005-81251005 on Build GRCh38
rs200518871 Chr.5: 81250982-81250982 on Build GRCh38
rs540455278 Chr.5: 81250635-81250635 on Build GRCh38
rs775929110 Chr.5: 81250793-81250793 on Build GRCh38
rs139841645 Chr.5: 81250906-81250906 on Build GRCh38
rs398050514 Chr.5: 81250942-81250943 on Build GRCh38
rs878968908 Chr.5: 81250980-81250980 on Build GRCh38
rs144278096 Chr.5: 81251070-81251070 on Build GRCh38
rs966063395 Chr.5: 81250795-81250795 on Build GRCh38
rs982337166 Chr.5: 81250674-81250674 on Build GRCh38
rs950842334 Chr.5: 81250664-81250664 on Build GRCh38
rs1026719008 Chr.5: 81250992-81250992 on Build GRCh38
rs772556939 Chr.5: 81250787-81250787 on Build GRCh38
rs71962414 Chr.5: 81250983-81250984 on Build GRCh38
rs995239990 Chr.5: 81251004-81251004 on Build GRCh38
rs547242331 Chr.5: 81251013-81251013 on Build GRCh38
rs962923733 Chr.5: 81250979-81250980 on Build GRCh38
rs1042431311 Chr.5: 81250776-81250776 on Build GRCh38
rs112680973 Chr.5: 81251032-81251032 on Build GRCh38
rs150013847 Chr.5: 81251050-81251050 on Build GRCh38
rs995271025 Chr.5: 81250681-81250681 on Build GRCh38
rs756411805 Chr.5: 81250873-81250873 on Build GRCh38
rs531585922 Chr.5: 81250853-81250853 on Build GRCh38
rs147458522 Chr.5: 81250869-81250869 on Build GRCh38
rs191648854 Chr.5: 81250679-81250679 on Build GRCh38
rs539493361 Chr.5: 81251024-81251024 on Build GRCh38
rs139063325 Chr.5: 81250941-81250942 on Build GRCh38
rs924617366 Chr.5: 81250856-81250856 on Build GRCh38
rs1019119199 Chr.5: 81251031-81251031 on Build GRCh38
rs538597454 Chr.5: 81250746-81250747 on Build GRCh38
rs570585694 Chr.5: 81251017-81251017 on Build GRCh38
rs67943397 Chr.5: 81250978-81250979 on Build GRCh38
rs562500932 Chr.5: 81250841-81250841 on Build GRCh38
rs534030588 Chr.5: 81250988-81250988 on Build GRCh38
rs1050952403 Chr.5: 81250851-81250851 on Build GRCh38
rs986379667 Chr.5: 81250765-81250765 on Build GRCh38
rs977791326 Chr.5: 81250855-81250855 on Build GRCh38
rs1047546673 Chr.5: 81251007-81251007 on Build GRCh38
rs950870546 Chr.5: 81250992-81250993 on Build GRCh38
rs994871188 Chr.5: 81250986-81250986 on Build GRCh38
rs898174363 Chr.5: 81250993-81250993 on Build GRCh38
rs71876955 Chr.5: 81250982-81250983 on Build GRCh38
rs977720325 Chr.5: 81251046-81251046 on Build GRCh38

View Full Product Details