Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 87397368-87397877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC01590
Entrez Gene ID
63914
Gene Name
long intergenic non-protein coding RNA 1590
Gene Aliases
C6orf164, dJ102H19.4
UniGene
Hs.645177
Gene Chromosome Location
Chr.6: 87397124-87399741 on Build GRCh38
Transcripts
NR_026784.1

SNP Detail

SNP ID Chromosome Location
rs549422234 Chr.6: 87397407-87397407 on Build GRCh38
rs764976641 Chr.6: 87397423-87397423 on Build GRCh38
rs111919267 Chr.6: 87397576-87397576 on Build GRCh38
rs985134318 Chr.6: 87397787-87397788 on Build GRCh38
rs993682933 Chr.6: 87397406-87397406 on Build GRCh38
rs192523945 Chr.6: 87397474-87397474 on Build GRCh38
rs572234006 Chr.6: 87397563-87397563 on Build GRCh38
rs77954359 Chr.6: 87397759-87397759 on Build GRCh38
rs13217556 Chr.6: 87397559-87397559 on Build GRCh38
rs977924105 Chr.6: 87397454-87397454 on Build GRCh38
rs545601352 Chr.6: 87397703-87397703 on Build GRCh38
rs537278997 Chr.6: 87397851-87397851 on Build GRCh38
rs887250653 Chr.6: 87397718-87397718 on Build GRCh38
rs988670737 Chr.6: 87397582-87397582 on Build GRCh38
rs910490623 Chr.6: 87397615-87397615 on Build GRCh38
rs969335648 Chr.6: 87397536-87397536 on Build GRCh38
rs942987704 Chr.6: 87397651-87397651 on Build GRCh38
rs1025190713 Chr.6: 87397532-87397532 on Build GRCh38
rs985414919 Chr.6: 87397399-87397399 on Build GRCh38
rs752322091 Chr.6: 87397797-87397797 on Build GRCh38
rs9344708 Chr.6: 87397610-87397610 on Build GRCh38
rs565417638 Chr.6: 87397763-87397763 on Build GRCh38
rs923775037 Chr.6: 87397513-87397513 on Build GRCh38
rs1037357934 Chr.6: 87397661-87397661 on Build GRCh38
rs1047971994 Chr.6: 87397715-87397715 on Build GRCh38
rs967977989 Chr.6: 87397831-87397831 on Build GRCh38
rs556648754 Chr.6: 87397654-87397654 on Build GRCh38
rs998370242 Chr.6: 87397736-87397736 on Build GRCh38
rs988208226 Chr.6: 87397827-87397827 on Build GRCh38
rs1032301841 Chr.6: 87397778-87397778 on Build GRCh38
rs543268562 Chr.6: 87397767-87397767 on Build GRCh38
rs780830367 Chr.6: 87397425-87397425 on Build GRCh38
rs1002431336 Chr.6: 87397777-87397777 on Build GRCh38
rs140262649 Chr.6: 87397587-87397587 on Build GRCh38
rs1014188227 Chr.6: 87397732-87397732 on Build GRCh38
rs571111353 Chr.6: 87397411-87397411 on Build GRCh38
rs9362428 Chr.6: 87397697-87397697 on Build GRCh38
rs925313844 Chr.6: 87397569-87397569 on Build GRCh38
rs574048805 Chr.6: 87397598-87397598 on Build GRCh38

View Full Product Details