Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 138696183-138696677 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ46906
Entrez Gene ID
441172
Gene Name
uncharacterized LOC441172
Gene Aliases
-
UniGene
Hs.92290
Gene Chromosome Location
Chr.6: 138691668-138697288 on Build GRCh38
Transcripts
NR_033896.1

SNP Detail

SNP ID Chromosome Location
rs949082218 Chr.6: 138696488-138696488 on Build GRCh38
rs757956085 Chr.6: 138696236-138696236 on Build GRCh38
rs975894556 Chr.6: 138696344-138696344 on Build GRCh38
rs780143589 Chr.6: 138696345-138696345 on Build GRCh38
rs988040836 Chr.6: 138696238-138696238 on Build GRCh38
rs375264357 Chr.6: 138696449-138696449 on Build GRCh38
rs4348337 Chr.6: 138696643-138696643 on Build GRCh38
rs1047978514 Chr.6: 138696257-138696257 on Build GRCh38
rs986784120 Chr.6: 138696291-138696291 on Build GRCh38
rs369173944 Chr.6: 138696603-138696603 on Build GRCh38
rs779509033 Chr.6: 138696303-138696303 on Build GRCh38
rs191004501 Chr.6: 138696559-138696559 on Build GRCh38
rs552664049 Chr.6: 138696393-138696393 on Build GRCh38
rs887236094 Chr.6: 138696302-138696303 on Build GRCh38
rs578213074 Chr.6: 138696625-138696625 on Build GRCh38
rs774025065 Chr.6: 138696652-138696652 on Build GRCh38
rs779513471 Chr.6: 138696580-138696580 on Build GRCh38
rs887329465 Chr.6: 138696483-138696483 on Build GRCh38
rs778011823 Chr.6: 138696508-138696508 on Build GRCh38
rs966515796 Chr.6: 138696557-138696557 on Build GRCh38
rs9484199 Chr.6: 138696520-138696520 on Build GRCh38
rs139280868 Chr.6: 138696253-138696253 on Build GRCh38
rs78314231 Chr.6: 138696343-138696343 on Build GRCh38
rs746352169 Chr.6: 138696584-138696584 on Build GRCh38
rs931451087 Chr.6: 138696412-138696412 on Build GRCh38
rs989704756 Chr.6: 138696513-138696513 on Build GRCh38
rs1047589068 Chr.6: 138696654-138696654 on Build GRCh38
rs937691781 Chr.6: 138696620-138696620 on Build GRCh38
rs536797173 Chr.6: 138696244-138696244 on Build GRCh38
rs186420561 Chr.6: 138696270-138696270 on Build GRCh38
rs1004694831 Chr.6: 138696310-138696310 on Build GRCh38
rs1019651479 Chr.6: 138696327-138696327 on Build GRCh38
rs893657038 Chr.6: 138696617-138696617 on Build GRCh38
rs772314986 Chr.6: 138696619-138696619 on Build GRCh38

View Full Product Details