Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 138696964-138697482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ46906
Entrez Gene ID
441172
Gene Name
uncharacterized LOC441172
Gene Aliases
-
UniGene
Hs.92290
Gene Chromosome Location
Chr.6: 138691668-138697288 on Build GRCh38
Transcripts
NR_033896.1

SNP Detail

SNP ID Chromosome Location
rs1012344344 Chr.6: 138697392-138697392 on Build GRCh38
rs907788891 Chr.6: 138697190-138697190 on Build GRCh38
rs1027825960 Chr.6: 138697404-138697404 on Build GRCh38
rs975980791 Chr.6: 138697453-138697453 on Build GRCh38
rs1038963673 Chr.6: 138697215-138697215 on Build GRCh38
rs965951763 Chr.6: 138697214-138697215 on Build GRCh38
rs868686661 Chr.6: 138696987-138696987 on Build GRCh38
rs902809903 Chr.6: 138697309-138697309 on Build GRCh38
rs970931419 Chr.6: 138697366-138697366 on Build GRCh38
rs921751907 Chr.6: 138697219-138697219 on Build GRCh38
rs964096610 Chr.6: 138697002-138697002 on Build GRCh38
rs1035146255 Chr.6: 138697130-138697130 on Build GRCh38
rs996854651 Chr.6: 138697223-138697223 on Build GRCh38
rs893781650 Chr.6: 138697107-138697107 on Build GRCh38
rs1011936601 Chr.6: 138697166-138697166 on Build GRCh38
rs1016069335 Chr.6: 138697326-138697326 on Build GRCh38
rs1027079699 Chr.6: 138697057-138697057 on Build GRCh38
rs867122170 Chr.6: 138697074-138697074 on Build GRCh38
rs908746393 Chr.6: 138697113-138697113 on Build GRCh38
rs146543743 Chr.6: 138697122-138697122 on Build GRCh38
rs558500348 Chr.6: 138697234-138697234 on Build GRCh38
rs546604545 Chr.6: 138697141-138697141 on Build GRCh38
rs982864316 Chr.6: 138697112-138697112 on Build GRCh38
rs952736722 Chr.6: 138697089-138697089 on Build GRCh38
rs915299358 Chr.6: 138697243-138697243 on Build GRCh38
rs1027029072 Chr.6: 138697244-138697244 on Build GRCh38
rs959654297 Chr.6: 138697412-138697412 on Build GRCh38
rs1040194325 Chr.6: 138697041-138697041 on Build GRCh38
rs752521283 Chr.6: 138697220-138697220 on Build GRCh38
rs372825963 Chr.6: 138697213-138697213 on Build GRCh38
rs1056586751 Chr.6: 138697337-138697337 on Build GRCh38
rs529719732 Chr.6: 138697137-138697137 on Build GRCh38
rs557239465 Chr.6: 138697419-138697419 on Build GRCh38
rs914936043 Chr.6: 138697421-138697421 on Build GRCh38
rs74772752 Chr.6: 138697170-138697170 on Build GRCh38
rs3813334 Chr.6: 138697045-138697045 on Build GRCh38
rs926838141 Chr.6: 138697143-138697143 on Build GRCh38
rs965982406 Chr.6: 138697441-138697441 on Build GRCh38
rs937697659 Chr.6: 138697145-138697145 on Build GRCh38
rs890319181 Chr.6: 138697205-138697205 on Build GRCh38
rs1033935818 Chr.6: 138697370-138697370 on Build GRCh38
rs535383328 Chr.6: 138697230-138697230 on Build GRCh38
rs1053343288 Chr.6: 138697149-138697149 on Build GRCh38
rs902730116 Chr.6: 138697121-138697121 on Build GRCh38
rs1043044899 Chr.6: 138697119-138697119 on Build GRCh38
rs780854758 Chr.6: 138697199-138697199 on Build GRCh38
rs1014587034 Chr.6: 138697433-138697433 on Build GRCh38
rs566343804 Chr.6: 138697167-138697167 on Build GRCh38
rs1129180 Chr.6: 138696995-138696995 on Build GRCh38
rs530626192 Chr.6: 138697026-138697026 on Build GRCh38
rs951815980 Chr.6: 138697169-138697169 on Build GRCh38
rs537664034 Chr.6: 138697277-138697277 on Build GRCh38
rs945163269 Chr.6: 138697197-138697197 on Build GRCh38
rs1004995323 Chr.6: 138697426-138697426 on Build GRCh38
rs1050290323 Chr.6: 138697227-138697227 on Build GRCh38
rs754589714 Chr.6: 138697163-138697163 on Build GRCh38
rs3088031 Chr.6: 138696989-138696989 on Build GRCh38
rs76321834 Chr.6: 138697255-138697255 on Build GRCh38
rs546489136 Chr.6: 138697082-138697082 on Build GRCh38
rs982651715 Chr.6: 138697368-138697368 on Build GRCh38

View Full Product Details