Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 130083224-130083759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASAP1
Entrez Gene ID
50807
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 1
Gene Aliases
AMAP1, CENTB4, DDEF1, PAG2, PAP, ZG14P
UniGene
Hs.655552
Gene Chromosome Location
Chr.8: 130052104-130443848 on Build GRCh38
Transcripts
NM_001247996.1 NM_018482.3 XM_005250925.1 XM_006716563.3 XM_006716564.1 XM_006716565.3 XM_006716566.1 XM_011517052.2 XM_011517053.1 XM_017013467.1 XM_017013468.1 XM_017013469.1 NR_045385.1

Gene Symbol
ASAP1-IT2
Entrez Gene ID
100507117
Gene Name
ASAP1 intronic transcript 2
Gene Aliases
-
UniGene
Hs.106015
Gene Chromosome Location
Chr.8: 130082738-130084768 on Build GRCh38
Transcripts
NM_001247996.1 NM_018482.3 XM_005250925.1 XM_006716563.3 XM_006716564.1 XM_006716565.3 XM_006716566.1 XM_011517052.2 XM_011517053.1 XM_017013467.1 XM_017013468.1 XM_017013469.1 NR_045385.1

SNP Detail

SNP ID Chromosome Location
rs979640673 Chr.8: 130083719-130083719 on Build GRCh38
rs1037500188 Chr.8: 130083627-130083627 on Build GRCh38
rs961682298 Chr.8: 130083296-130083296 on Build GRCh38
rs1017010777 Chr.8: 130083267-130083267 on Build GRCh38
rs962778369 Chr.8: 130083280-130083280 on Build GRCh38
rs118096112 Chr.8: 130083552-130083552 on Build GRCh38
rs541946781 Chr.8: 130083684-130083684 on Build GRCh38
rs752940491 Chr.8: 130083270-130083271 on Build GRCh38
rs964281793 Chr.8: 130083404-130083404 on Build GRCh38
rs905102212 Chr.8: 130083594-130083594 on Build GRCh38
rs905050411 Chr.8: 130083257-130083257 on Build GRCh38
rs988684500 Chr.8: 130083475-130083475 on Build GRCh38
rs979586520 Chr.8: 130083441-130083441 on Build GRCh38
rs935070610 Chr.8: 130083480-130083480 on Build GRCh38
rs375178856 Chr.8: 130083440-130083440 on Build GRCh38
rs147235003 Chr.8: 130083648-130083648 on Build GRCh38
rs994173864 Chr.8: 130083284-130083284 on Build GRCh38
rs1008640449 Chr.8: 130083702-130083702 on Build GRCh38
rs1025619695 Chr.8: 130083301-130083301 on Build GRCh38
rs1056817874 Chr.8: 130083566-130083566 on Build GRCh38
rs936995476 Chr.8: 130083565-130083565 on Build GRCh38
rs535953329 Chr.8: 130083470-130083470 on Build GRCh38
rs577129857 Chr.8: 130083584-130083584 on Build GRCh38
rs1018560304 Chr.8: 130083706-130083706 on Build GRCh38
rs964651356 Chr.8: 130083713-130083713 on Build GRCh38
rs143923068 Chr.8: 130083559-130083559 on Build GRCh38
rs955528077 Chr.8: 130083369-130083369 on Build GRCh38
rs116849444 Chr.8: 130083604-130083604 on Build GRCh38
rs1018923299 Chr.8: 130083399-130083399 on Build GRCh38
rs548302335 Chr.8: 130083277-130083277 on Build GRCh38
rs558779148 Chr.8: 130083564-130083564 on Build GRCh38
rs1026138429 Chr.8: 130083698-130083698 on Build GRCh38
rs754396972 Chr.8: 130083346-130083346 on Build GRCh38
rs1006465690 Chr.8: 130083259-130083259 on Build GRCh38
rs917602735 Chr.8: 130083524-130083524 on Build GRCh38
rs539476984 Chr.8: 130083699-130083699 on Build GRCh38
rs764258864 Chr.8: 130083301-130083301 on Build GRCh38
rs780913812 Chr.8: 130083640-130083640 on Build GRCh38
rs912682029 Chr.8: 130083454-130083454 on Build GRCh38
rs987865376 Chr.8: 130083498-130083498 on Build GRCh38
rs186388808 Chr.8: 130083383-130083383 on Build GRCh38
rs188397129 Chr.8: 130083639-130083639 on Build GRCh38

View Full Product Details