Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35860022-35860531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00950
Entrez Gene ID
92973
Gene Name
long intergenic non-protein coding RNA 950
Gene Aliases
FP588
UniGene
Hs.12509
Gene Chromosome Location
Chr.9: 35860274-35865518 on Build GRCh38
Transcripts
NR_024006.1 XM_011517916.2

Gene Symbol
TMEM8B
Entrez Gene ID
51754
Gene Name
transmembrane protein 8B
Gene Aliases
C9orf127, NAG-5, NAG5, NGX6
UniGene
Hs.493808
Gene Chromosome Location
Chr.9: 35825817-35865515 on Build GRCh38
Transcripts
NR_024006.1 XM_011517916.2

SNP Detail

SNP ID Chromosome Location
rs922380057 Chr.9: 35860088-35860088 on Build GRCh38
rs879154794 Chr.9: 35860361-35860361 on Build GRCh38
rs984270065 Chr.9: 35860333-35860333 on Build GRCh38
rs964320844 Chr.9: 35860342-35860342 on Build GRCh38
rs1034360653 Chr.9: 35860271-35860271 on Build GRCh38
rs1035124913 Chr.9: 35860379-35860379 on Build GRCh38
rs933629008 Chr.9: 35860413-35860413 on Build GRCh38
rs750820711 Chr.9: 35860288-35860288 on Build GRCh38
rs942366863 Chr.9: 35860080-35860080 on Build GRCh38
rs766318680 Chr.9: 35860409-35860409 on Build GRCh38
rs187377481 Chr.9: 35860141-35860141 on Build GRCh38
rs919203576 Chr.9: 35860169-35860169 on Build GRCh38
rs758010820 Chr.9: 35860293-35860293 on Build GRCh38
rs538291872 Chr.9: 35860207-35860207 on Build GRCh38
rs905113116 Chr.9: 35860214-35860214 on Build GRCh38
rs150311486 Chr.9: 35860163-35860163 on Build GRCh38
rs910317142 Chr.9: 35860469-35860469 on Build GRCh38
rs749717725 Chr.9: 35860138-35860138 on Build GRCh38
rs199720806 Chr.9: 35860287-35860287 on Build GRCh38
rs1021970129 Chr.9: 35860401-35860401 on Build GRCh38
rs544875599 Chr.9: 35860054-35860054 on Build GRCh38
rs117967052 Chr.9: 35860286-35860286 on Build GRCh38
rs768894540 Chr.9: 35860158-35860158 on Build GRCh38
rs1043655359 Chr.9: 35860162-35860162 on Build GRCh38
rs772237941 Chr.9: 35860310-35860310 on Build GRCh38
rs929178141 Chr.9: 35860204-35860204 on Build GRCh38
rs533450467 Chr.9: 35860109-35860109 on Build GRCh38
rs1017236407 Chr.9: 35860339-35860339 on Build GRCh38
rs4879932 Chr.9: 35860344-35860344 on Build GRCh38
rs528826314 Chr.9: 35860444-35860444 on Build GRCh38
rs774670522 Chr.9: 35860161-35860161 on Build GRCh38
rs370419934 Chr.9: 35860328-35860328 on Build GRCh38
rs565117626 Chr.9: 35860108-35860108 on Build GRCh38
rs140192815 Chr.9: 35860048-35860048 on Build GRCh38
rs975104664 Chr.9: 35860082-35860082 on Build GRCh38
rs189546934 Chr.9: 35860494-35860494 on Build GRCh38
rs910121744 Chr.9: 35860058-35860058 on Build GRCh38
rs776538256 Chr.9: 35860428-35860428 on Build GRCh38
rs10814287 Chr.9: 35860486-35860486 on Build GRCh38
rs1001973475 Chr.9: 35860221-35860221 on Build GRCh38
rs968153734 Chr.9: 35860442-35860442 on Build GRCh38
rs144952601 Chr.9: 35860448-35860448 on Build GRCh38
rs967827743 Chr.9: 35860134-35860134 on Build GRCh38
rs546964355 Chr.9: 35860236-35860236 on Build GRCh38
rs566194671 Chr.9: 35860381-35860381 on Build GRCh38
rs543963022 Chr.9: 35860126-35860126 on Build GRCh38
rs1051783867 Chr.9: 35860481-35860481 on Build GRCh38
rs745657481 Chr.9: 35860397-35860400 on Build GRCh38
rs1003698324 Chr.9: 35860373-35860373 on Build GRCh38

View Full Product Details