Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35863756-35864267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00950
Entrez Gene ID
92973
Gene Name
long intergenic non-protein coding RNA 950
Gene Aliases
FP588
UniGene
Hs.12509
Gene Chromosome Location
Chr.9: 35860274-35865518 on Build GRCh38
Transcripts
NR_024006.1 XM_011517916.2

Gene Symbol
TMEM8B
Entrez Gene ID
51754
Gene Name
transmembrane protein 8B
Gene Aliases
C9orf127, NAG-5, NAG5, NGX6
UniGene
Hs.493808
Gene Chromosome Location
Chr.9: 35825817-35865515 on Build GRCh38
Transcripts
NR_024006.1 XM_011517916.2

SNP Detail

SNP ID Chromosome Location
rs965768506 Chr.9: 35863881-35863881 on Build GRCh38
rs1049394516 Chr.9: 35864124-35864124 on Build GRCh38
rs568390354 Chr.9: 35864120-35864120 on Build GRCh38
rs935811496 Chr.9: 35863940-35863940 on Build GRCh38
rs4879933 Chr.9: 35863931-35863931 on Build GRCh38
rs980397319 Chr.9: 35863932-35863932 on Build GRCh38
rs995119045 Chr.9: 35863813-35863813 on Build GRCh38
rs1031372391 Chr.9: 35864108-35864108 on Build GRCh38
rs190974760 Chr.9: 35863790-35863790 on Build GRCh38
rs927969445 Chr.9: 35864233-35864233 on Build GRCh38
rs1027939489 Chr.9: 35864059-35864059 on Build GRCh38
rs930728649 Chr.9: 35863853-35863853 on Build GRCh38
rs561566357 Chr.9: 35863817-35863817 on Build GRCh38
rs919828808 Chr.9: 35863980-35863980 on Build GRCh38
rs180720642 Chr.9: 35864002-35864002 on Build GRCh38
rs758804068 Chr.9: 35863787-35863787 on Build GRCh38
rs911681367 Chr.9: 35863880-35863880 on Build GRCh38
rs74379925 Chr.9: 35864104-35864104 on Build GRCh38
rs534219154 Chr.9: 35864148-35864148 on Build GRCh38
rs948653830 Chr.9: 35864185-35864185 on Build GRCh38
rs889362805 Chr.9: 35864075-35864075 on Build GRCh38
rs1033100849 Chr.9: 35863907-35863907 on Build GRCh38
rs981353596 Chr.9: 35864232-35864232 on Build GRCh38
rs955094477 Chr.9: 35863941-35863941 on Build GRCh38
rs1007770220 Chr.9: 35864085-35864085 on Build GRCh38
rs948120075 Chr.9: 35863976-35863976 on Build GRCh38
rs915708607 Chr.9: 35864181-35864181 on Build GRCh38
rs549569422 Chr.9: 35864024-35864024 on Build GRCh38
rs939924548 Chr.9: 35863816-35863816 on Build GRCh38
rs547263405 Chr.9: 35863882-35863882 on Build GRCh38
rs890898437 Chr.9: 35864125-35864125 on Build GRCh38
rs894547624 Chr.9: 35863867-35863867 on Build GRCh38
rs1018571880 Chr.9: 35864090-35864090 on Build GRCh38
rs916010940 Chr.9: 35864047-35864048 on Build GRCh38
rs953102528 Chr.9: 35863860-35863860 on Build GRCh38
rs141933774 Chr.9: 35864063-35864063 on Build GRCh38
rs920566027 Chr.9: 35863824-35863824 on Build GRCh38
rs1026934766 Chr.9: 35864205-35864205 on Build GRCh38
rs542644419 Chr.9: 35864150-35864150 on Build GRCh38
rs756213909 Chr.9: 35864086-35864086 on Build GRCh38
rs952071951 Chr.9: 35863989-35863989 on Build GRCh38
rs1007938931 Chr.9: 35864134-35864134 on Build GRCh38
rs541804394 Chr.9: 35863788-35863788 on Build GRCh38
rs892838375 Chr.9: 35863930-35863930 on Build GRCh38
rs986612070 Chr.9: 35863804-35863804 on Build GRCh38
rs781418052 Chr.9: 35863945-35863945 on Build GRCh38
rs1015849189 Chr.9: 35864174-35864174 on Build GRCh38
rs995401191 Chr.9: 35864047-35864047 on Build GRCh38
rs764421095 Chr.9: 35863805-35863805 on Build GRCh38
rs1003643507 Chr.9: 35864033-35864033 on Build GRCh38
rs961591272 Chr.9: 35863967-35863967 on Build GRCh38
rs934817190 Chr.9: 35864091-35864091 on Build GRCh38
rs118007914 Chr.9: 35864045-35864045 on Build GRCh38
rs372070319 Chr.9: 35864046-35864046 on Build GRCh38
rs915808751 Chr.9: 35863956-35863956 on Build GRCh38
rs572478340 Chr.9: 35864106-35864106 on Build GRCh38
rs778259080 Chr.9: 35863826-35863826 on Build GRCh38
rs778792825 Chr.9: 35864101-35864101 on Build GRCh38
rs527564060 Chr.9: 35863872-35863872 on Build GRCh38
rs990389257 Chr.9: 35864149-35864149 on Build GRCh38
rs1055839232 Chr.9: 35863865-35863865 on Build GRCh38
rs977105576 Chr.9: 35863918-35863918 on Build GRCh38
rs769001350 Chr.9: 35864158-35864158 on Build GRCh38
rs959254506 Chr.9: 35864222-35864222 on Build GRCh38
rs1045878481 Chr.9: 35864066-35864066 on Build GRCh38
rs1045477165 Chr.9: 35864008-35864008 on Build GRCh38

View Full Product Details