Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 38622109-38622595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM201A
Entrez Gene ID
158228
Gene Name
family with sequence similarity 201 member A
Gene Aliases
C9orf122
UniGene
Hs.632652
Gene Chromosome Location
Chr.9: 38621088-38623284 on Build GRCh38
Transcripts
NR_027294.1

SNP Detail

SNP ID Chromosome Location
rs574600251 Chr.9: 38622566-38622566 on Build GRCh38
rs769710433 Chr.9: 38622538-38622538 on Build GRCh38
rs1016926363 Chr.9: 38622340-38622340 on Build GRCh38
rs905202883 Chr.9: 38622351-38622351 on Build GRCh38
rs901367945 Chr.9: 38622294-38622294 on Build GRCh38
rs925630095 Chr.9: 38622567-38622567 on Build GRCh38
rs945705231 Chr.9: 38622221-38622221 on Build GRCh38
rs34333377 Chr.9: 38622178-38622179 on Build GRCh38
rs776923882 Chr.9: 38622319-38622319 on Build GRCh38
rs72713749 Chr.9: 38622260-38622260 on Build GRCh38
rs1020460634 Chr.9: 38622425-38622425 on Build GRCh38
rs1001431459 Chr.9: 38622352-38622352 on Build GRCh38
rs992653891 Chr.9: 38622397-38622397 on Build GRCh38
rs927168747 Chr.9: 38622144-38622144 on Build GRCh38
rs1047607600 Chr.9: 38622309-38622309 on Build GRCh38
rs570812358 Chr.9: 38622199-38622199 on Build GRCh38
rs535524514 Chr.9: 38622174-38622174 on Build GRCh38
rs190549057 Chr.9: 38622298-38622298 on Build GRCh38
rs912953358 Chr.9: 38622213-38622213 on Build GRCh38
rs960159665 Chr.9: 38622372-38622372 on Build GRCh38
rs557156695 Chr.9: 38622208-38622208 on Build GRCh38
rs540134835 Chr.9: 38622280-38622280 on Build GRCh38
rs7046879 Chr.9: 38622371-38622371 on Build GRCh38
rs995364412 Chr.9: 38622487-38622487 on Build GRCh38
rs559374650 Chr.9: 38622467-38622467 on Build GRCh38
rs978504688 Chr.9: 38622530-38622530 on Build GRCh38
rs768115841 Chr.9: 38622132-38622132 on Build GRCh38
rs774616700 Chr.9: 38622563-38622563 on Build GRCh38
rs573479604 Chr.9: 38622324-38622324 on Build GRCh38
rs959882839 Chr.9: 38622148-38622148 on Build GRCh38
rs925602032 Chr.9: 38622274-38622274 on Build GRCh38
rs773023012 Chr.9: 38622311-38622311 on Build GRCh38
rs776175405 Chr.9: 38622187-38622187 on Build GRCh38
rs879400623 Chr.9: 38622463-38622463 on Build GRCh38
rs967187584 Chr.9: 38622496-38622496 on Build GRCh38
rs1039957542 Chr.9: 38622283-38622283 on Build GRCh38

View Full Product Details