Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 69820546-69821043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf135
Entrez Gene ID
138255
Gene Name
chromosome 9 open reading frame 135
Gene Aliases
-
UniGene
Hs.444459
Gene Chromosome Location
Chr.9: 69820804-69906237 on Build GRCh38
Transcripts
NM_001010940.2 NM_001308084.1 NM_001308085.1 NM_001308086.1 NR_131756.1 NR_131757.1 XM_011518229.2 XM_011518230.2 XM_011518231.2 XM_011518234.2 NR_038833.1

Gene Symbol
C9orf135-AS1
Entrez Gene ID
494558
Gene Name
C9orf135 antisense RNA 1 (head to head)
Gene Aliases
-
UniGene
Hs.130722
Gene Chromosome Location
Chr.9: 69819405-69820739 on Build GRCh38
Transcripts
NM_001010940.2 NM_001308084.1 NM_001308085.1 NM_001308086.1 NR_131756.1 NR_131757.1 XM_011518229.2 XM_011518230.2 XM_011518231.2 XM_011518234.2 NR_038833.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
80949 c.120G>A p.K40K Chr.9: 69820999-69820999 on Build GRCh38
3848700 c.86G>A p.R29H Chr.9: 69820965-69820965 on Build GRCh38
3731050 c.37G>A p.D13N Chr.9: 69820916-69820916 on Build GRCh38
1109743 c.106C>T p.R36C Chr.9: 69820985-69820985 on Build GRCh38
3907746 c.150C>T p.S50S Chr.9: 69821029-69821029 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760463078 Chr.9: 69820913-69820913 on Build GRCh38
rs777881224 Chr.9: 69820670-69820670 on Build GRCh38
rs962498709 Chr.9: 69820761-69820761 on Build GRCh38
rs966721163 Chr.9: 69820661-69820661 on Build GRCh38
rs768524639 Chr.9: 69820985-69820985 on Build GRCh38
rs745462491 Chr.9: 69820970-69820970 on Build GRCh38
rs909667821 Chr.9: 69820738-69820738 on Build GRCh38
rs751642816 Chr.9: 69821015-69821015 on Build GRCh38
rs149641101 Chr.9: 69820968-69820968 on Build GRCh38
rs954068543 Chr.9: 69820717-69820717 on Build GRCh38
rs972508447 Chr.9: 69820770-69820770 on Build GRCh38
rs779093954 Chr.9: 69820841-69820841 on Build GRCh38
rs1024984067 Chr.9: 69820583-69820583 on Build GRCh38
rs748566007 Chr.9: 69820961-69820961 on Build GRCh38
rs1033327761 Chr.9: 69820889-69820889 on Build GRCh38
rs981780013 Chr.9: 69820608-69820608 on Build GRCh38
rs998124034 Chr.9: 69820677-69820677 on Build GRCh38
rs1054557043 Chr.9: 69820780-69820780 on Build GRCh38
rs755057937 Chr.9: 69821020-69821020 on Build GRCh38
rs143163232 Chr.9: 69820910-69820910 on Build GRCh38
rs918334838 Chr.9: 69820796-69820796 on Build GRCh38
rs1047519140 Chr.9: 69820667-69820667 on Build GRCh38
rs757591893 Chr.9: 69820764-69820764 on Build GRCh38
rs753690197 Chr.9: 69820924-69820924 on Build GRCh38
rs1010822059 Chr.9: 69820798-69820798 on Build GRCh38
rs552797337 Chr.9: 69820612-69820612 on Build GRCh38
rs774119117 Chr.9: 69820882-69820882 on Build GRCh38
rs747552571 Chr.9: 69820954-69820954 on Build GRCh38
rs755499542 Chr.9: 69820959-69820959 on Build GRCh38
rs750600894 Chr.9: 69820928-69820928 on Build GRCh38
rs1029609421 Chr.9: 69820686-69820686 on Build GRCh38
rs900486201 Chr.9: 69820735-69820735 on Build GRCh38
rs781711958 Chr.9: 69820960-69820960 on Build GRCh38
rs778571754 Chr.9: 69820950-69820951 on Build GRCh38
rs758439214 Chr.9: 69820846-69820846 on Build GRCh38
rs535463064 Chr.9: 69820803-69820803 on Build GRCh38
rs138120161 Chr.9: 69820867-69820867 on Build GRCh38
rs1010914523 Chr.9: 69820646-69820646 on Build GRCh38
rs766889158 Chr.9: 69820902-69820902 on Build GRCh38
rs1020986883 Chr.9: 69820653-69820653 on Build GRCh38
rs764088063 Chr.9: 69820916-69820916 on Build GRCh38
rs776565645 Chr.9: 69820814-69820816 on Build GRCh38
rs944599031 Chr.9: 69820712-69820712 on Build GRCh38
rs911837863 Chr.9: 69820676-69820676 on Build GRCh38
rs61146180 Chr.9: 69820876-69820876 on Build GRCh38
rs760218675 Chr.9: 69820979-69820979 on Build GRCh38
rs959141792 Chr.9: 69820636-69820636 on Build GRCh38
rs779936251 Chr.9: 69820937-69820937 on Build GRCh38
rs370333172 Chr.9: 69820915-69820915 on Build GRCh38
rs571018466 Chr.9: 69820614-69820614 on Build GRCh38
rs202155059 Chr.9: 69820842-69820842 on Build GRCh38
rs867663881 Chr.9: 69820942-69820942 on Build GRCh38
rs755324521 Chr.9: 69820884-69820885 on Build GRCh38
rs780639954 Chr.9: 69820856-69820856 on Build GRCh38
rs758492391 Chr.9: 69820929-69820929 on Build GRCh38
rs373790286 Chr.9: 69821004-69821004 on Build GRCh38
rs759150916 Chr.9: 69820883-69820883 on Build GRCh38
rs547283686 Chr.9: 69820725-69820725 on Build GRCh38
rs757022096 Chr.9: 69820925-69820925 on Build GRCh38
rs907481694 Chr.9: 69820844-69820844 on Build GRCh38
rs1055679737 Chr.9: 69820624-69820624 on Build GRCh38
rs150197009 Chr.9: 69820972-69820972 on Build GRCh38
rs868544514 Chr.9: 69820880-69820880 on Build GRCh38
rs182952358 Chr.9: 69820640-69820640 on Build GRCh38
rs1010374075 Chr.9: 69820580-69820580 on Build GRCh38
rs1024473089 Chr.9: 69820838-69820838 on Build GRCh38
rs1002391397 Chr.9: 69820601-69820601 on Build GRCh38
rs7047711 Chr.9: 69820576-69820576 on Build GRCh38
rs985328496 Chr.9: 69820734-69820734 on Build GRCh38
rs17439752 Chr.9: 69820797-69820797 on Build GRCh38
rs557351518 Chr.9: 69820809-69820809 on Build GRCh38
rs769075775 Chr.9: 69820875-69820875 on Build GRCh38
rs757541388 Chr.9: 69820840-69820840 on Build GRCh38
rs770759896 Chr.9: 69820877-69820877 on Build GRCh38
rs761626492 Chr.9: 69820990-69820990 on Build GRCh38
rs766616336 Chr.9: 69821012-69821012 on Build GRCh38
rs750655220 Chr.9: 69820997-69820997 on Build GRCh38
rs138857531 Chr.9: 69820978-69820978 on Build GRCh38
rs148273758 Chr.9: 69820926-69820926 on Build GRCh38

View Full Product Details