Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 94809972-94810286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf3
Entrez Gene ID
84909
Gene Name
chromosome 9 open reading frame 3
Gene Aliases
AOPEP, AP-O, APO, C90RF3, ONPEP
UniGene
Hs.434253
Gene Chromosome Location
Chr.9: 94726604-95148264 on Build GRCh38
Transcripts
NM_001193329.1 NM_001193331.2 NM_032823.5 XM_005252284.2 XM_006717306.1 XM_011519119.1 XM_011519120.2 XM_011519121.2 XM_011519122.2 XM_011519123.2 XM_011519127.1 XM_011519129.1 XM_011519130.1 XM_011519132.1 XM_017015229.1 XM_017015230.1 XM_017015231.1 XM_017015232.1 XM_017015233.1 XM_017015234.1 XM_017015235.1 XM_017015236.1 XM_017015237.1 NR_031755.1

Gene Symbol
MIR2278
Entrez Gene ID
100313780
Gene Name
microRNA 2278
Gene Aliases
mir-2278
UniGene
Gene Chromosome Location
Chr.9: 94809962-94810057 on Build GRCh38
Transcripts
NM_001193329.1 NM_001193331.2 NM_032823.5 XM_005252284.2 XM_006717306.1 XM_011519119.1 XM_011519120.2 XM_011519121.2 XM_011519122.2 XM_011519123.2 XM_011519127.1 XM_011519129.1 XM_011519130.1 XM_011519132.1 XM_017015229.1 XM_017015230.1 XM_017015231.1 XM_017015232.1 XM_017015233.1 XM_017015234.1 XM_017015235.1 XM_017015236.1 XM_017015237.1 NR_031755.1

SNP Detail

SNP ID Chromosome Location
rs944457753 Chr.9: 94810052-94810052 on Build GRCh38
rs878909253 Chr.9: 94810067-94810069 on Build GRCh38
rs947001045 Chr.9: 94810026-94810026 on Build GRCh38
rs993753563 Chr.9: 94810141-94810141 on Build GRCh38
rs752254545 Chr.9: 94810065-94810065 on Build GRCh38
rs1056750500 Chr.9: 94810105-94810105 on Build GRCh38
rs1035060950 Chr.9: 94810159-94810159 on Build GRCh38
rs780204954 Chr.9: 94810097-94810097 on Build GRCh38
rs938124353 Chr.9: 94809994-94809994 on Build GRCh38
rs537538626 Chr.9: 94810008-94810008 on Build GRCh38
rs959458995 Chr.9: 94810160-94810160 on Build GRCh38
rs758057469 Chr.9: 94810053-94810053 on Build GRCh38
rs1004833201 Chr.9: 94810116-94810116 on Build GRCh38
rs1015993685 Chr.9: 94810119-94810119 on Build GRCh38
rs961411327 Chr.9: 94810122-94810122 on Build GRCh38
rs769682890 Chr.9: 94810151-94810151 on Build GRCh38
rs369995284 Chr.9: 94810011-94810011 on Build GRCh38
rs923601311 Chr.9: 94810251-94810251 on Build GRCh38
rs915438754 Chr.9: 94810203-94810203 on Build GRCh38
rs1041425708 Chr.9: 94810063-94810063 on Build GRCh38
rs118121535 Chr.9: 94810114-94810114 on Build GRCh38
rs894397079 Chr.9: 94810109-94810109 on Build GRCh38
rs945538627 Chr.9: 94810018-94810018 on Build GRCh38
rs991366131 Chr.9: 94810167-94810167 on Build GRCh38
rs760204021 Chr.9: 94810072-94810072 on Build GRCh38
rs564268100 Chr.9: 94810079-94810079 on Build GRCh38
rs1003623758 Chr.9: 94810135-94810135 on Build GRCh38
rs902209992 Chr.9: 94810037-94810037 on Build GRCh38
rs759195858 Chr.9: 94810038-94810038 on Build GRCh38
rs965323367 Chr.9: 94810206-94810206 on Build GRCh38
rs374286085 Chr.9: 94810091-94810091 on Build GRCh38
rs968685837 Chr.9: 94810215-94810215 on Build GRCh38
rs989709741 Chr.9: 94810230-94810230 on Build GRCh38
rs377608494 Chr.9: 94810092-94810092 on Build GRCh38
rs1042433153 Chr.9: 94810035-94810035 on Build GRCh38
rs377343613 Chr.9: 94809995-94809995 on Build GRCh38
rs766737375 Chr.9: 94810085-94810085 on Build GRCh38
rs1016600604 Chr.9: 94810131-94810131 on Build GRCh38
rs987450328 Chr.9: 94810180-94810180 on Build GRCh38
rs910595093 Chr.9: 94810183-94810183 on Build GRCh38
rs561787249 Chr.9: 94810242-94810242 on Build GRCh38
rs991003784 Chr.9: 94809996-94809996 on Build GRCh38
rs945537352 Chr.9: 94810248-94810248 on Build GRCh38
rs1002482049 Chr.9: 94810096-94810096 on Build GRCh38

View Full Product Details