Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 116543565-116544073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASTN2
Entrez Gene ID
23245
Gene Name
astrotactin 2
Gene Aliases
bA67K19.1
UniGene
Hs.601562
Gene Chromosome Location
Chr.9: 116425225-117415039 on Build GRCh38
Transcripts
NM_001184734.1 NM_014010.4 NM_198186.3 NM_198187.3 NM_198188.2 NR_033973.1

Gene Symbol
ASTN2-AS1
Entrez Gene ID
100128505
Gene Name
ASTN2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.675821
Gene Chromosome Location
Chr.9: 116504283-116562293 on Build GRCh38
Transcripts
NM_001184734.1 NM_014010.4 NM_198186.3 NM_198187.3 NM_198188.2 NR_033973.1

SNP Detail

SNP ID Chromosome Location
rs572429186 Chr.9: 116543928-116543928 on Build GRCh38
rs182166967 Chr.9: 116543906-116543906 on Build GRCh38
rs73657147 Chr.9: 116543837-116543837 on Build GRCh38
rs1005589911 Chr.9: 116543786-116543786 on Build GRCh38
rs41308928 Chr.9: 116543851-116543851 on Build GRCh38
rs766511187 Chr.9: 116543670-116543670 on Build GRCh38
rs899887907 Chr.9: 116543952-116543952 on Build GRCh38
rs1050719846 Chr.9: 116543677-116543677 on Build GRCh38
rs978917898 Chr.9: 116543700-116543700 on Build GRCh38
rs543263122 Chr.9: 116544002-116544002 on Build GRCh38
rs967042937 Chr.9: 116543665-116543665 on Build GRCh38
rs923207848 Chr.9: 116543662-116543662 on Build GRCh38
rs140868046 Chr.9: 116543961-116543961 on Build GRCh38
rs888625078 Chr.9: 116544042-116544042 on Build GRCh38
rs753133617 Chr.9: 116543785-116543785 on Build GRCh38
rs910350711 Chr.9: 116543680-116543681 on Build GRCh38
rs1008721595 Chr.9: 116544003-116544003 on Build GRCh38
rs61273754 Chr.9: 116543885-116543885 on Build GRCh38
rs933580059 Chr.9: 116543597-116543597 on Build GRCh38
rs10983228 Chr.9: 116543642-116543642 on Build GRCh38
rs924724192 Chr.9: 116543927-116543927 on Build GRCh38
rs577881663 Chr.9: 116543987-116543987 on Build GRCh38
rs1015986799 Chr.9: 116543842-116543842 on Build GRCh38
rs950291583 Chr.9: 116543722-116543722 on Build GRCh38
rs555202393 Chr.9: 116544007-116544007 on Build GRCh38
rs933221524 Chr.9: 116543672-116543672 on Build GRCh38
rs912888299 Chr.9: 116543659-116543659 on Build GRCh38
rs954575091 Chr.9: 116543838-116543838 on Build GRCh38
rs991626296 Chr.9: 116543886-116543886 on Build GRCh38
rs867550492 Chr.9: 116543922-116543922 on Build GRCh38
rs996389915 Chr.9: 116543798-116543798 on Build GRCh38
rs967438683 Chr.9: 116543853-116543853 on Build GRCh38
rs867231551 Chr.9: 116543811-116543811 on Build GRCh38
rs896113682 Chr.9: 116543791-116543791 on Build GRCh38
rs560185441 Chr.9: 116543711-116543711 on Build GRCh38
rs1047341786 Chr.9: 116543905-116543905 on Build GRCh38
rs545863249 Chr.9: 116543802-116543802 on Build GRCh38
rs1047373592 Chr.9: 116543753-116543753 on Build GRCh38
rs541269137 Chr.9: 116543944-116543944 on Build GRCh38
rs1008752744 Chr.9: 116543850-116543850 on Build GRCh38
rs1057044446 Chr.9: 116543735-116543735 on Build GRCh38
rs750443927 Chr.9: 116543629-116543629 on Build GRCh38
rs369944870 Chr.9: 116544019-116544019 on Build GRCh38
rs376742812 Chr.9: 116543707-116543707 on Build GRCh38
rs11792733 Chr.9: 116543701-116543701 on Build GRCh38
rs745586660 Chr.9: 116543934-116543934 on Build GRCh38
rs556239730 Chr.9: 116544033-116544033 on Build GRCh38
rs917407961 Chr.9: 116543889-116543889 on Build GRCh38
rs536776827 Chr.9: 116543880-116543880 on Build GRCh38
rs189764167 Chr.9: 116543605-116543605 on Build GRCh38
rs1051150049 Chr.9: 116543965-116543965 on Build GRCh38
rs151003998 Chr.9: 116543907-116543907 on Build GRCh38
rs887087988 Chr.9: 116543755-116543755 on Build GRCh38
rs57366433 Chr.9: 116543714-116543714 on Build GRCh38
rs987071101 Chr.9: 116543647-116543647 on Build GRCh38
rs961682902 Chr.9: 116543633-116543633 on Build GRCh38
rs903101348 Chr.9: 116544020-116544020 on Build GRCh38
rs1038827311 Chr.9: 116543794-116543794 on Build GRCh38
rs570038459 Chr.9: 116543884-116543884 on Build GRCh38
rs574150364 Chr.9: 116543797-116543797 on Build GRCh38
rs908874741 Chr.9: 116543613-116543613 on Build GRCh38
rs533856959 Chr.9: 116543900-116543900 on Build GRCh38
rs1023564333 Chr.9: 116543822-116543822 on Build GRCh38
rs528956462 Chr.9: 116543703-116543703 on Build GRCh38
rs947210305 Chr.9: 116543921-116543921 on Build GRCh38

View Full Product Details