Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 116547199-116547739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASTN2
Entrez Gene ID
23245
Gene Name
astrotactin 2
Gene Aliases
bA67K19.1
UniGene
Hs.601562
Gene Chromosome Location
Chr.9: 116425225-117415039 on Build GRCh38
Transcripts
NM_001184734.1 NM_014010.4 NM_198186.3 NM_198187.3 NM_198188.2 NR_033973.1

Gene Symbol
ASTN2-AS1
Entrez Gene ID
100128505
Gene Name
ASTN2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.675821
Gene Chromosome Location
Chr.9: 116504283-116562293 on Build GRCh38
Transcripts
NM_001184734.1 NM_014010.4 NM_198186.3 NM_198187.3 NM_198188.2 NR_033973.1

SNP Detail

SNP ID Chromosome Location
rs537484942 Chr.9: 116547343-116547343 on Build GRCh38
rs544178560 Chr.9: 116547282-116547282 on Build GRCh38
rs568105635 Chr.9: 116547551-116547551 on Build GRCh38
rs149140794 Chr.9: 116547415-116547415 on Build GRCh38
rs1056187984 Chr.9: 116547567-116547567 on Build GRCh38
rs773488392 Chr.9: 116547422-116547422 on Build GRCh38
rs180889342 Chr.9: 116547461-116547461 on Build GRCh38
rs976132136 Chr.9: 116547421-116547421 on Build GRCh38
rs982566379 Chr.9: 116547460-116547460 on Build GRCh38
rs1004333132 Chr.9: 116547345-116547345 on Build GRCh38
rs1010736352 Chr.9: 116547419-116547419 on Build GRCh38
rs4837613 Chr.9: 116547246-116547246 on Build GRCh38
rs964650999 Chr.9: 116547652-116547652 on Build GRCh38
rs1052718323 Chr.9: 116547477-116547477 on Build GRCh38
rs376455763 Chr.9: 116547285-116547285 on Build GRCh38
rs867567324 Chr.9: 116547257-116547257 on Build GRCh38
rs1017844629 Chr.9: 116547639-116547639 on Build GRCh38
rs533845371 Chr.9: 116547600-116547600 on Build GRCh38
rs966773850 Chr.9: 116547441-116547441 on Build GRCh38
rs561271466 Chr.9: 116547317-116547317 on Build GRCh38
rs762284002 Chr.9: 116547333-116547333 on Build GRCh38
rs933801562 Chr.9: 116547429-116547429 on Build GRCh38
rs573358595 Chr.9: 116547673-116547673 on Build GRCh38
rs1030394231 Chr.9: 116547686-116547686 on Build GRCh38
rs759184724 Chr.9: 116547601-116547601 on Build GRCh38
rs143158556 Chr.9: 116547531-116547531 on Build GRCh38
rs956334764 Chr.9: 116547706-116547706 on Build GRCh38
rs1041161956 Chr.9: 116547653-116547653 on Build GRCh38
rs928006591 Chr.9: 116547497-116547497 on Build GRCh38
rs964718852 Chr.9: 116547402-116547402 on Build GRCh38
rs1034801444 Chr.9: 116547251-116547251 on Build GRCh38
rs957932145 Chr.9: 116547417-116547417 on Build GRCh38
rs1043688522 Chr.9: 116547550-116547550 on Build GRCh38
rs1001877489 Chr.9: 116547556-116547556 on Build GRCh38
rs1040716126 Chr.9: 116547352-116547352 on Build GRCh38
rs866647918 Chr.9: 116547546-116547546 on Build GRCh38
rs930821112 Chr.9: 116547292-116547292 on Build GRCh38
rs532467562 Chr.9: 116547395-116547395 on Build GRCh38
rs1026543296 Chr.9: 116547298-116547298 on Build GRCh38
rs757703042 Chr.9: 116547379-116547379 on Build GRCh38
rs564059446 Chr.9: 116547358-116547358 on Build GRCh38
rs930873149 Chr.9: 116547604-116547604 on Build GRCh38
rs923360752 Chr.9: 116547427-116547427 on Build GRCh38
rs996786818 Chr.9: 116547367-116547367 on Build GRCh38
rs10817902 Chr.9: 116547330-116547330 on Build GRCh38
rs146769975 Chr.9: 116547638-116547638 on Build GRCh38
rs527515186 Chr.9: 116547231-116547231 on Build GRCh38
rs111518748 Chr.9: 116547453-116547453 on Build GRCh38
rs1026884395 Chr.9: 116547609-116547609 on Build GRCh38
rs566613489 Chr.9: 116547366-116547366 on Build GRCh38
rs920884998 Chr.9: 116547566-116547566 on Build GRCh38
rs913791156 Chr.9: 116547525-116547525 on Build GRCh38
rs896242428 Chr.9: 116547599-116547599 on Build GRCh38
rs537871774 Chr.9: 116547465-116547465 on Build GRCh38
rs952268655 Chr.9: 116547631-116547631 on Build GRCh38
rs767175489 Chr.9: 116547672-116547672 on Build GRCh38
rs10983231 Chr.9: 116547250-116547250 on Build GRCh38

View Full Product Details