Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 116550883-116551388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASTN2
Entrez Gene ID
23245
Gene Name
astrotactin 2
Gene Aliases
bA67K19.1
UniGene
Hs.601562
Gene Chromosome Location
Chr.9: 116425225-117415039 on Build GRCh38
Transcripts
NM_001184734.1 NM_014010.4 NM_198186.3 NM_198187.3 NM_198188.2 NR_033973.1

Gene Symbol
ASTN2-AS1
Entrez Gene ID
100128505
Gene Name
ASTN2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.675821
Gene Chromosome Location
Chr.9: 116504283-116562293 on Build GRCh38
Transcripts
NM_001184734.1 NM_014010.4 NM_198186.3 NM_198187.3 NM_198188.2 NR_033973.1

SNP Detail

SNP ID Chromosome Location
rs1035580931 Chr.9: 116551355-116551355 on Build GRCh38
rs931458639 Chr.9: 116551088-116551088 on Build GRCh38
rs1041734222 Chr.9: 116551162-116551162 on Build GRCh38
rs551948045 Chr.9: 116551351-116551351 on Build GRCh38
rs974443642 Chr.9: 116551230-116551230 on Build GRCh38
rs988878219 Chr.9: 116551256-116551256 on Build GRCh38
rs919681058 Chr.9: 116551044-116551044 on Build GRCh38
rs957081845 Chr.9: 116551256-116551256 on Build GRCh38
rs1022779230 Chr.9: 116551201-116551201 on Build GRCh38
rs972423439 Chr.9: 116551041-116551041 on Build GRCh38
rs377638072 Chr.9: 116551010-116551010 on Build GRCh38
rs949972032 Chr.9: 116551349-116551349 on Build GRCh38
rs1895062 Chr.9: 116551207-116551207 on Build GRCh38
rs1025919970 Chr.9: 116551150-116551150 on Build GRCh38
rs905953080 Chr.9: 116551047-116551047 on Build GRCh38
rs116866716 Chr.9: 116550916-116550916 on Build GRCh38
rs1033107420 Chr.9: 116551240-116551240 on Build GRCh38
rs957151183 Chr.9: 116550941-116550941 on Build GRCh38
rs552714843 Chr.9: 116551099-116551099 on Build GRCh38
rs1053844765 Chr.9: 116551178-116551178 on Build GRCh38
rs935319774 Chr.9: 116551173-116551173 on Build GRCh38
rs949951189 Chr.9: 116551018-116551018 on Build GRCh38
rs1035359649 Chr.9: 116550988-116550988 on Build GRCh38
rs1011431597 Chr.9: 116550945-116550945 on Build GRCh38
rs60343130 Chr.9: 116551166-116551166 on Build GRCh38
rs925584532 Chr.9: 116550955-116550955 on Build GRCh38
rs10124063 Chr.9: 116551096-116551096 on Build GRCh38
rs748348323 Chr.9: 116550949-116550949 on Build GRCh38
rs866678175 Chr.9: 116551172-116551172 on Build GRCh38
rs937368504 Chr.9: 116551059-116551059 on Build GRCh38
rs114162435 Chr.9: 116550946-116550946 on Build GRCh38
rs1053014786 Chr.9: 116551003-116551003 on Build GRCh38
rs749048893 Chr.9: 116551216-116551216 on Build GRCh38
rs571392036 Chr.9: 116550932-116550932 on Build GRCh38
rs898466494 Chr.9: 116551116-116551116 on Build GRCh38
rs748817403 Chr.9: 116551312-116551312 on Build GRCh38
rs911410827 Chr.9: 116551149-116551149 on Build GRCh38
rs566440528 Chr.9: 116551118-116551118 on Build GRCh38
rs913120058 Chr.9: 116551008-116551008 on Build GRCh38
rs12000922 Chr.9: 116550915-116550915 on Build GRCh38
rs559839604 Chr.9: 116551153-116551153 on Build GRCh38
rs143279863 Chr.9: 116551363-116551363 on Build GRCh38
rs961063698 Chr.9: 116551023-116551023 on Build GRCh38
rs59494108 Chr.9: 116550963-116550963 on Build GRCh38
rs373829449 Chr.9: 116551262-116551262 on Build GRCh38
rs1012774359 Chr.9: 116551199-116551199 on Build GRCh38
rs567491775 Chr.9: 116551001-116551001 on Build GRCh38
rs747722852 Chr.9: 116551196-116551196 on Build GRCh38

View Full Product Details