Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 126409094-126409591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MVB12B
Entrez Gene ID
89853
Gene Name
multivesicular body subunit 12B
Gene Aliases
C9orf28, FAM125B
UniGene
Hs.162659
Gene Chromosome Location
Chr.9: 126326844-126507041 on Build GRCh38
Transcripts
NM_033446.2 XM_005252297.1 XM_017015275.1 XM_017015276.1 XM_017015277.1 XM_017015278.1 XM_017015279.1 NR_045006.1

Gene Symbol
NRON
Entrez Gene ID
641373
Gene Name
non-protein coding RNA, repressor of NFAT
Gene Aliases
NCRNA00194
UniGene
Hs.742606
Gene Chromosome Location
Chr.9: 126407775-126410504 on Build GRCh38
Transcripts
NM_033446.2 XM_005252297.1 XM_017015275.1 XM_017015276.1 XM_017015277.1 XM_017015278.1 XM_017015279.1 NR_045006.1

SNP Detail

SNP ID Chromosome Location
rs578041408 Chr.9: 126409565-126409565 on Build GRCh38
rs13302579 Chr.9: 126409125-126409125 on Build GRCh38
rs771243850 Chr.9: 126409388-126409391 on Build GRCh38
rs374497199 Chr.9: 126409124-126409125 on Build GRCh38
rs918892700 Chr.9: 126409378-126409378 on Build GRCh38
rs924689201 Chr.9: 126409482-126409482 on Build GRCh38
rs35445972 Chr.9: 126409480-126409480 on Build GRCh38
rs963270915 Chr.9: 126409335-126409335 on Build GRCh38
rs754392584 Chr.9: 126409299-126409300 on Build GRCh38
rs71493516 Chr.9: 126409125-126409125 on Build GRCh38
rs1054853526 Chr.9: 126409484-126409484 on Build GRCh38
rs773031318 Chr.9: 126409300-126409311 on Build GRCh38
rs572075751 Chr.9: 126409569-126409569 on Build GRCh38
rs976487226 Chr.9: 126409117-126409117 on Build GRCh38
rs934767676 Chr.9: 126409483-126409483 on Build GRCh38
rs927903914 Chr.9: 126409331-126409331 on Build GRCh38
rs969182842 Chr.9: 126409317-126409317 on Build GRCh38
rs386738604 Chr.9: 126409123-126409125 on Build GRCh38
rs375989186 Chr.9: 126409480-126409480 on Build GRCh38
rs568226203 Chr.9: 126409115-126409116 on Build GRCh38
rs113788658 Chr.9: 126409481-126409481 on Build GRCh38
rs893162397 Chr.9: 126409509-126409509 on Build GRCh38
rs950090598 Chr.9: 126409152-126409152 on Build GRCh38
rs570488875 Chr.9: 126409118-126409118 on Build GRCh38
rs796453098 Chr.9: 126409481-126409481 on Build GRCh38
rs557875042 Chr.9: 126409546-126409546 on Build GRCh38
rs369006716 Chr.9: 126409116-126409116 on Build GRCh38
rs1016628673 Chr.9: 126409299-126409299 on Build GRCh38
rs60354636 Chr.9: 126409346-126409347 on Build GRCh38
rs1039629968 Chr.9: 126409517-126409517 on Build GRCh38
rs939026750 Chr.9: 126409122-126409122 on Build GRCh38
rs13302876 Chr.9: 126409124-126409124 on Build GRCh38
rs112884871 Chr.9: 126409165-126409165 on Build GRCh38
rs71929574 Chr.9: 126409300-126409303 on Build GRCh38
rs990171777 Chr.9: 126409242-126409242 on Build GRCh38
rs544546161 Chr.9: 126409352-126409352 on Build GRCh38
rs916104585 Chr.9: 126409150-126409150 on Build GRCh38
rs759277869 Chr.9: 126409404-126409404 on Build GRCh38
rs950206006 Chr.9: 126409426-126409426 on Build GRCh38
rs140022897 Chr.9: 126409116-126409117 on Build GRCh38
rs951171841 Chr.9: 126409287-126409287 on Build GRCh38
rs1045864442 Chr.9: 126409164-126409164 on Build GRCh38
rs187959662 Chr.9: 126409213-126409213 on Build GRCh38
rs372788258 Chr.9: 126409481-126409481 on Build GRCh38
rs199680815 Chr.9: 126409177-126409177 on Build GRCh38
rs780519631 Chr.9: 126409311-126409312 on Build GRCh38
rs1029368024 Chr.9: 126409265-126409265 on Build GRCh38
rs1019199872 Chr.9: 126409175-126409175 on Build GRCh38
rs556059391 Chr.9: 126409214-126409214 on Build GRCh38
rs768616843 Chr.9: 126409479-126409480 on Build GRCh38
rs893709466 Chr.9: 126409191-126409191 on Build GRCh38
rs1010855099 Chr.9: 126409208-126409208 on Build GRCh38
rs200784491 Chr.9: 126409483-126409484 on Build GRCh38
rs897579657 Chr.9: 126409123-126409124 on Build GRCh38
rs916036166 Chr.9: 126409413-126409413 on Build GRCh38
rs980691744 Chr.9: 126409325-126409326 on Build GRCh38
rs1054501203 Chr.9: 126409185-126409185 on Build GRCh38
rs372369134 Chr.9: 126409477-126409477 on Build GRCh38
rs1056355265 Chr.9: 126409133-126409133 on Build GRCh38
rs982404301 Chr.9: 126409296-126409296 on Build GRCh38
rs78419122 Chr.9: 126409124-126409125 on Build GRCh38
rs915048905 Chr.9: 126409117-126409118 on Build GRCh38
rs372716929 Chr.9: 126409480-126409481 on Build GRCh38
rs191257921 Chr.9: 126409226-126409226 on Build GRCh38
rs67395110 Chr.9: 126409343-126409346 on Build GRCh38
rs889322498 Chr.9: 126409481-126409482 on Build GRCh38
rs991774330 Chr.9: 126409340-126409340 on Build GRCh38
rs946594291 Chr.9: 126409513-126409513 on Build GRCh38
rs982955217 Chr.9: 126409120-126409120 on Build GRCh38
rs866712265 Chr.9: 126409561-126409561 on Build GRCh38
rs1048829356 Chr.9: 126409433-126409433 on Build GRCh38
rs910121983 Chr.9: 126409123-126409123 on Build GRCh38
rs769888534 Chr.9: 126409116-126409116 on Build GRCh38
rs997854673 Chr.9: 126409228-126409228 on Build GRCh38
rs374326059 Chr.9: 126409119-126409119 on Build GRCh38
rs61211126 Chr.9: 126409337-126409346 on Build GRCh38

View Full Product Details