Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136324264-136324771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKFZP434A062
Entrez Gene ID
26102
Gene Name
uncharacterized LOC26102
Gene Aliases
-
UniGene
Hs.561893
Gene Chromosome Location
Chr.9: 136322302-136327323 on Build GRCh38
Transcripts
NR_026964.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4138890 c.479C>G p.P160R Chr.9: 136324706-136324706 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755746922 Chr.9: 136324592-136324592 on Build GRCh38
rs1000699969 Chr.9: 136324591-136324591 on Build GRCh38
rs116811956 Chr.9: 136324688-136324688 on Build GRCh38
rs370926910 Chr.9: 136324555-136324555 on Build GRCh38
rs889905812 Chr.9: 136324590-136324590 on Build GRCh38
rs79545543 Chr.9: 136324625-136324625 on Build GRCh38
rs140952949 Chr.9: 136324341-136324342 on Build GRCh38
rs562836041 Chr.9: 136324531-136324531 on Build GRCh38
rs971123757 Chr.9: 136324482-136324482 on Build GRCh38
rs531294156 Chr.9: 136324626-136324626 on Build GRCh38
rs998643445 Chr.9: 136324395-136324395 on Build GRCh38
rs534156821 Chr.9: 136324699-136324699 on Build GRCh38
rs979876810 Chr.9: 136324287-136324287 on Build GRCh38
rs527536264 Chr.9: 136324585-136324585 on Build GRCh38
rs189569114 Chr.9: 136324698-136324698 on Build GRCh38
rs544650998 Chr.9: 136324506-136324506 on Build GRCh38
rs573314751 Chr.9: 136324439-136324439 on Build GRCh38
rs1054193680 Chr.9: 136324593-136324593 on Build GRCh38
rs1024217092 Chr.9: 136324630-136324630 on Build GRCh38
rs984542601 Chr.9: 136324678-136324678 on Build GRCh38
rs558831365 Chr.9: 136324475-136324475 on Build GRCh38
rs549385120 Chr.9: 136324640-136324640 on Build GRCh38
rs1037295553 Chr.9: 136324556-136324556 on Build GRCh38
rs955926669 Chr.9: 136324455-136324455 on Build GRCh38
rs752071354 Chr.9: 136324349-136324349 on Build GRCh38
rs891631914 Chr.9: 136324604-136324604 on Build GRCh38
rs980258965 Chr.9: 136324484-136324484 on Build GRCh38
rs984261615 Chr.9: 136324507-136324507 on Build GRCh38
rs34940359 Chr.9: 136324429-136324429 on Build GRCh38
rs782333527 Chr.9: 136324510-136324510 on Build GRCh38
rs1037012460 Chr.9: 136324383-136324383 on Build GRCh38
rs561143876 Chr.9: 136324485-136324485 on Build GRCh38
rs536516878 Chr.9: 136324403-136324403 on Build GRCh38
rs559091956 Chr.9: 136324716-136324716 on Build GRCh38
rs1034446369 Chr.9: 136324655-136324655 on Build GRCh38
rs910070893 Chr.9: 136324553-136324553 on Build GRCh38
rs901176519 Chr.9: 136324394-136324394 on Build GRCh38
rs972977008 Chr.9: 136324748-136324748 on Build GRCh38
rs1000055786 Chr.9: 136324422-136324422 on Build GRCh38
rs938234667 Chr.9: 136324314-136324314 on Build GRCh38
rs1049870929 Chr.9: 136324397-136324397 on Build GRCh38
rs1050252416 Chr.9: 136324589-136324589 on Build GRCh38
rs574806501 Chr.9: 136324541-136324541 on Build GRCh38
rs545977501 Chr.9: 136324588-136324588 on Build GRCh38
rs926779159 Chr.9: 136324292-136324292 on Build GRCh38
rs139830606 Chr.9: 136324642-136324642 on Build GRCh38
rs111799605 Chr.9: 136324320-136324320 on Build GRCh38
rs62579954 Chr.9: 136324456-136324456 on Build GRCh38
rs72775734 Chr.9: 136324569-136324569 on Build GRCh38
rs940269709 Chr.9: 136324343-136324343 on Build GRCh38
rs577390403 Chr.9: 136324501-136324501 on Build GRCh38
rs781790468 Chr.9: 136324722-136324722 on Build GRCh38
rs971453351 Chr.9: 136324641-136324641 on Build GRCh38
rs530335065 Chr.9: 136324656-136324656 on Build GRCh38

View Full Product Details