Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 111521582-111522091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00890
Entrez Gene ID
401613
Gene Name
long intergenic non-protein coding RNA 890
Gene Aliases
DKFZp686D0853
UniGene
Hs.98581
Gene Chromosome Location
Chr.X: 111511662-111522399 on Build GRCh38
Transcripts
NR_033974.1

SNP Detail

SNP ID Chromosome Location
rs192955191 Chr.X: 111521611-111521611 on Build GRCh38
rs201317304 Chr.X: 111521731-111521731 on Build GRCh38
rs777559890 Chr.X: 111521811-111521811 on Build GRCh38
rs1029159482 Chr.X: 111521857-111521857 on Build GRCh38
rs985134570 Chr.X: 111522055-111522055 on Build GRCh38
rs1040898186 Chr.X: 111521607-111521607 on Build GRCh38
rs776432978 Chr.X: 111521996-111521997 on Build GRCh38
rs747490047 Chr.X: 111521668-111521668 on Build GRCh38
rs988574497 Chr.X: 111521974-111521974 on Build GRCh38
rs1038024682 Chr.X: 111521799-111521799 on Build GRCh38
rs956794679 Chr.X: 111521931-111521931 on Build GRCh38
rs1022308084 Chr.X: 111521978-111521978 on Build GRCh38
rs902202812 Chr.X: 111521726-111521726 on Build GRCh38
rs757448524 Chr.X: 111521889-111521889 on Build GRCh38
rs1019025902 Chr.X: 111521925-111521925 on Build GRCh38
rs778000197 Chr.X: 111521867-111521867 on Build GRCh38
rs761539294 Chr.X: 111521990-111521996 on Build GRCh38
rs902011954 Chr.X: 111521608-111521608 on Build GRCh38
rs968430340 Chr.X: 111521993-111521993 on Build GRCh38
rs758720983 Chr.X: 111521824-111521824 on Build GRCh38
rs1021950272 Chr.X: 111521717-111521717 on Build GRCh38
rs893406492 Chr.X: 111521653-111521653 on Build GRCh38
rs189334587 Chr.X: 111521996-111521996 on Build GRCh38
rs1010959158 Chr.X: 111521666-111521666 on Build GRCh38
rs188589451 Chr.X: 111521828-111521828 on Build GRCh38
rs180832241 Chr.X: 111522004-111522004 on Build GRCh38
rs191500909 Chr.X: 111521926-111521926 on Build GRCh38
rs1050595635 Chr.X: 111521638-111521638 on Build GRCh38
rs939723648 Chr.X: 111521784-111521784 on Build GRCh38
rs184234429 Chr.X: 111521639-111521639 on Build GRCh38
rs932156895 Chr.X: 111521610-111521610 on Build GRCh38
rs1034804645 Chr.X: 111521817-111521817 on Build GRCh38
rs926492643 Chr.X: 111522035-111522035 on Build GRCh38
rs999282044 Chr.X: 111521808-111521808 on Build GRCh38
rs1009009747 Chr.X: 111521878-111521878 on Build GRCh38
rs952010930 Chr.X: 111522007-111522007 on Build GRCh38
rs183985299 Chr.X: 111521975-111521975 on Build GRCh38

View Full Product Details